Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39526154-39526710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP8A
Entrez Gene ID
353500
Gene Name
bone morphogenetic protein 8a
Gene Aliases
-
UniGene
Hs.472497
Gene Chromosome Location
Chr.1: 39491626-39529869 on Build GRCh38
Transcripts
NM_181809.3 NR_003929.2

Gene Symbol
PPIEL
Entrez Gene ID
728448
Gene Name
peptidylprolyl isomerase E like pseudogene
Gene Aliases
PPIEP1
UniGene
Hs.472508
Gene Chromosome Location
Chr.1: 39522280-39559698 on Build GRCh38
Transcripts
NM_181809.3 NR_003929.2

SNP Detail

SNP ID Chromosome Location
rs61779339 Chr.1: 39526357-39526357 on Build GRCh38
rs926698877 Chr.1: 39526647-39526647 on Build GRCh38
rs533739663 Chr.1: 39526575-39526575 on Build GRCh38
rs933634965 Chr.1: 39526427-39526427 on Build GRCh38
rs941136766 Chr.1: 39526463-39526463 on Build GRCh38
rs908174406 Chr.1: 39526628-39526628 on Build GRCh38
rs920966357 Chr.1: 39526253-39526253 on Build GRCh38
rs953737254 Chr.1: 39526286-39526286 on Build GRCh38
rs1028038493 Chr.1: 39526519-39526519 on Build GRCh38
rs531718283 Chr.1: 39526452-39526452 on Build GRCh38
rs938028626 Chr.1: 39526672-39526672 on Build GRCh38
rs1030583255 Chr.1: 39526596-39526596 on Build GRCh38
rs865947240 Chr.1: 39526443-39526443 on Build GRCh38
rs181446438 Chr.1: 39526490-39526490 on Build GRCh38
rs79642025 Chr.1: 39526355-39526355 on Build GRCh38
rs34624289 Chr.1: 39526356-39526357 on Build GRCh38
rs1017692685 Chr.1: 39526586-39526586 on Build GRCh38
rs571312413 Chr.1: 39526565-39526565 on Build GRCh38
rs908268625 Chr.1: 39526449-39526449 on Build GRCh38
rs950965897 Chr.1: 39526598-39526598 on Build GRCh38
rs982431136 Chr.1: 39526604-39526604 on Build GRCh38
rs939864618 Chr.1: 39526320-39526320 on Build GRCh38
rs773098705 Chr.1: 39526564-39526564 on Build GRCh38
rs553597247 Chr.1: 39526590-39526590 on Build GRCh38
rs979164391 Chr.1: 39526237-39526237 on Build GRCh38
rs527904515 Chr.1: 39526232-39526232 on Build GRCh38
rs896466288 Chr.1: 39526687-39526687 on Build GRCh38
rs1035238811 Chr.1: 39526194-39526194 on Build GRCh38
rs778377356 Chr.1: 39526589-39526589 on Build GRCh38
rs113902821 Chr.1: 39526340-39526340 on Build GRCh38
rs1045617057 Chr.1: 39526492-39526492 on Build GRCh38
rs575672357 Chr.1: 39526659-39526659 on Build GRCh38
rs759090474 Chr.1: 39526413-39526413 on Build GRCh38
rs1032120278 Chr.1: 39526554-39526554 on Build GRCh38
rs1056492151 Chr.1: 39526686-39526686 on Build GRCh38
rs919746020 Chr.1: 39526326-39526326 on Build GRCh38
rs867931245 Chr.1: 39526448-39526448 on Build GRCh38
rs761533607 Chr.1: 39526534-39526534 on Build GRCh38
rs905193638 Chr.1: 39526388-39526388 on Build GRCh38
rs1038136422 Chr.1: 39526484-39526484 on Build GRCh38
rs1024179590 Chr.1: 39526219-39526219 on Build GRCh38
rs769675574 Chr.1: 39526557-39526557 on Build GRCh38
rs1018162361 Chr.1: 39526325-39526325 on Build GRCh38
rs976290591 Chr.1: 39526400-39526400 on Build GRCh38
rs1000658300 Chr.1: 39526511-39526511 on Build GRCh38
rs904015809 Chr.1: 39526505-39526505 on Build GRCh38
rs1019677938 Chr.1: 39526563-39526563 on Build GRCh38
rs548259506 Chr.1: 39526486-39526486 on Build GRCh38
rs930761048 Chr.1: 39526342-39526342 on Build GRCh38
rs907168590 Chr.1: 39526495-39526495 on Build GRCh38
rs369268367 Chr.1: 39526252-39526252 on Build GRCh38
rs565310320 Chr.1: 39526198-39526198 on Build GRCh38
rs763689462 Chr.1: 39526437-39526437 on Build GRCh38
rs9988612 Chr.1: 39526441-39526441 on Build GRCh38
rs548992905 Chr.1: 39526384-39526384 on Build GRCh38
rs533157824 Chr.1: 39526264-39526264 on Build GRCh38
rs187983712 Chr.1: 39526289-39526289 on Build GRCh38
rs957712745 Chr.1: 39526291-39526291 on Build GRCh38
rs763235527 Chr.1: 39526245-39526246 on Build GRCh38
rs529048091 Chr.1: 39526352-39526352 on Build GRCh38
rs894379489 Chr.1: 39526207-39526207 on Build GRCh38
rs923523641 Chr.1: 39526406-39526406 on Build GRCh38
rs941053857 Chr.1: 39526655-39526655 on Build GRCh38
rs1012756500 Chr.1: 39526217-39526217 on Build GRCh38
rs907179936 Chr.1: 39526230-39526230 on Build GRCh38
rs78176082 Chr.1: 39526339-39526339 on Build GRCh38
rs998794369 Chr.1: 39526528-39526528 on Build GRCh38
rs931108568 Chr.1: 39526618-39526618 on Build GRCh38
rs987127246 Chr.1: 39526287-39526287 on Build GRCh38
rs990323181 Chr.1: 39526297-39526297 on Build GRCh38
rs967815335 Chr.1: 39526231-39526231 on Build GRCh38
rs564221085 Chr.1: 39526193-39526193 on Build GRCh38

View Full Product Details