Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 39528615-39529206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP8A
Entrez Gene ID
353500
Gene Name
bone morphogenetic protein 8a
Gene Aliases
-
UniGene
Hs.472497
Gene Chromosome Location
Chr.1: 39491626-39529869 on Build GRCh38
Transcripts
NM_181809.3 NR_003929.2

Gene Symbol
PPIEL
Entrez Gene ID
728448
Gene Name
peptidylprolyl isomerase E like pseudogene
Gene Aliases
PPIEP1
UniGene
Hs.472508
Gene Chromosome Location
Chr.1: 39522280-39559698 on Build GRCh38
Transcripts
NM_181809.3 NR_003929.2

SNP Detail

SNP ID Chromosome Location
rs1007388985 Chr.1: 39528657-39528666 on Build GRCh38
rs193191945 Chr.1: 39528901-39528901 on Build GRCh38
rs991683142 Chr.1: 39528823-39528823 on Build GRCh38
rs968249688 Chr.1: 39528958-39528958 on Build GRCh38
rs946931088 Chr.1: 39528790-39528790 on Build GRCh38
rs1028039365 Chr.1: 39528664-39528664 on Build GRCh38
rs905168375 Chr.1: 39528915-39528915 on Build GRCh38
rs921550720 Chr.1: 39529053-39529053 on Build GRCh38
rs1039787353 Chr.1: 39528662-39528662 on Build GRCh38
rs944538961 Chr.1: 39528852-39528852 on Build GRCh38
rs890250698 Chr.1: 39528850-39528850 on Build GRCh38
rs1036128472 Chr.1: 39529149-39529149 on Build GRCh38
rs988257306 Chr.1: 39528937-39528937 on Build GRCh38
rs1005800078 Chr.1: 39529065-39529065 on Build GRCh38
rs879501561 Chr.1: 39528873-39528874 on Build GRCh38
rs566034321 Chr.1: 39528943-39528943 on Build GRCh38
rs924245201 Chr.1: 39528854-39528854 on Build GRCh38
rs977136072 Chr.1: 39529124-39529124 on Build GRCh38
rs887073062 Chr.1: 39529017-39529017 on Build GRCh38
rs187903784 Chr.1: 39528831-39528831 on Build GRCh38
rs372991469 Chr.1: 39528923-39528923 on Build GRCh38
rs944423176 Chr.1: 39529127-39529127 on Build GRCh38
rs1035064847 Chr.1: 39528931-39528931 on Build GRCh38
rs548644445 Chr.1: 39528968-39528968 on Build GRCh38
rs960801473 Chr.1: 39528936-39528936 on Build GRCh38
rs1727104 Chr.1: 39528874-39528874 on Build GRCh38
rs925571040 Chr.1: 39528822-39528822 on Build GRCh38
rs781276792 Chr.1: 39529143-39529143 on Build GRCh38
rs969376655 Chr.1: 39528682-39528682 on Build GRCh38
rs34518993 Chr.1: 39528700-39528701 on Build GRCh38
rs988753666 Chr.1: 39528771-39528771 on Build GRCh38
rs1005728033 Chr.1: 39528806-39528806 on Build GRCh38
rs1031345321 Chr.1: 39529137-39529137 on Build GRCh38
rs767918232 Chr.1: 39529111-39529111 on Build GRCh38
rs867991427 Chr.1: 39528882-39528882 on Build GRCh38
rs1013069994 Chr.1: 39529077-39529077 on Build GRCh38
rs897612715 Chr.1: 39529158-39529158 on Build GRCh38
rs12038951 Chr.1: 39528849-39528849 on Build GRCh38
rs537372382 Chr.1: 39529176-39529176 on Build GRCh38
rs941281 Chr.1: 39528818-39528818 on Build GRCh38
rs980059373 Chr.1: 39528984-39528984 on Build GRCh38
rs916994409 Chr.1: 39528826-39528826 on Build GRCh38
rs1002193944 Chr.1: 39528924-39528924 on Build GRCh38
rs570819988 Chr.1: 39528717-39528717 on Build GRCh38
rs930499948 Chr.1: 39529174-39529174 on Build GRCh38
rs534251291 Chr.1: 39528700-39528700 on Build GRCh38
rs990507080 Chr.1: 39529177-39529177 on Build GRCh38
rs56361728 Chr.1: 39528716-39528716 on Build GRCh38
rs755247 Chr.1: 39528873-39528873 on Build GRCh38
rs770872591 Chr.1: 39528778-39528778 on Build GRCh38
rs71497571 Chr.1: 39528873-39528873 on Build GRCh38
rs911632914 Chr.1: 39529102-39529102 on Build GRCh38
rs960853451 Chr.1: 39528764-39528764 on Build GRCh38
rs866299973 Chr.1: 39528690-39528690 on Build GRCh38
rs985820338 Chr.1: 39529078-39529078 on Build GRCh38
rs138801822 Chr.1: 39528946-39528946 on Build GRCh38
rs1040220788 Chr.1: 39528914-39528914 on Build GRCh38
rs12031730 Chr.1: 39528743-39528743 on Build GRCh38
rs568768437 Chr.1: 39529066-39529066 on Build GRCh38
rs958901670 Chr.1: 39528820-39528820 on Build GRCh38
rs957023302 Chr.1: 39529164-39529164 on Build GRCh38
rs931645618 Chr.1: 39528839-39528839 on Build GRCh38
rs201517393 Chr.1: 39528717-39528717 on Build GRCh38

View Full Product Details