Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59672233-59672781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP2
Entrez Gene ID
663
Gene Name
BCL2 interacting protein 2
Gene Aliases
BNIP-2, NIP2
UniGene
Hs.592515
Gene Chromosome Location
Chr.15: 59659146-59689534 on Build GRCh38
Transcripts
NM_001320674.1 NM_001320675.1 NM_004330.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612188 c.512C>T p.A171V Chr.15: 59672700-59672700 on Build GRCh38
6612187 c.875C>T p.A292V Chr.15: 59672700-59672700 on Build GRCh38
4947246 c.938+1G>T p.? Chr.15: 59672636-59672636 on Build GRCh38
159403 c.503T>C p.V168A Chr.15: 59672709-59672709 on Build GRCh38
1708242 c.937A>G p.K313E Chr.15: 59672638-59672638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915810472 Chr.15: 59672332-59672332 on Build GRCh38
rs746618822 Chr.15: 59672738-59672738 on Build GRCh38
rs773585405 Chr.15: 59672614-59672614 on Build GRCh38
rs4774342 Chr.15: 59672599-59672599 on Build GRCh38
rs560657903 Chr.15: 59672265-59672265 on Build GRCh38
rs139971944 Chr.15: 59672699-59672699 on Build GRCh38
rs981372829 Chr.15: 59672603-59672603 on Build GRCh38
rs997726952 Chr.15: 59672447-59672447 on Build GRCh38
rs536151933 Chr.15: 59672613-59672613 on Build GRCh38
rs568505596 Chr.15: 59672611-59672611 on Build GRCh38
rs908950029 Chr.15: 59672679-59672679 on Build GRCh38
rs887561057 Chr.15: 59672408-59672408 on Build GRCh38
rs540282559 Chr.15: 59672311-59672311 on Build GRCh38
rs531974192 Chr.15: 59672363-59672363 on Build GRCh38
rs928940841 Chr.15: 59672661-59672661 on Build GRCh38
rs542574455 Chr.15: 59672264-59672264 on Build GRCh38
rs554506189 Chr.15: 59672620-59672620 on Build GRCh38
rs6151536 Chr.15: 59672476-59672476 on Build GRCh38
rs768314671 Chr.15: 59672752-59672752 on Build GRCh38
rs566284607 Chr.15: 59672456-59672456 on Build GRCh38
rs752070398 Chr.15: 59672636-59672636 on Build GRCh38
rs573303945 Chr.15: 59672397-59672397 on Build GRCh38
rs151090751 Chr.15: 59672469-59672469 on Build GRCh38
rs371054991 Chr.15: 59672724-59672724 on Build GRCh38
rs529930196 Chr.15: 59672424-59672424 on Build GRCh38
rs575164059 Chr.15: 59672263-59672263 on Build GRCh38
rs370271258 Chr.15: 59672757-59672757 on Build GRCh38
rs756321029 Chr.15: 59672655-59672655 on Build GRCh38
rs1032821646 Chr.15: 59672480-59672480 on Build GRCh38
rs918927233 Chr.15: 59672366-59672366 on Build GRCh38
rs188606680 Chr.15: 59672405-59672405 on Build GRCh38
rs201623072 Chr.15: 59672595-59672595 on Build GRCh38
rs991360587 Chr.15: 59672325-59672325 on Build GRCh38
rs957418799 Chr.15: 59672321-59672321 on Build GRCh38
rs778983160 Chr.15: 59672697-59672697 on Build GRCh38
rs112561426 Chr.15: 59672647-59672647 on Build GRCh38
rs554327372 Chr.15: 59672345-59672345 on Build GRCh38
rs1008140812 Chr.15: 59672425-59672425 on Build GRCh38
rs148596531 Chr.15: 59672291-59672291 on Build GRCh38
rs749100497 Chr.15: 59672671-59672671 on Build GRCh38
rs766873046 Chr.15: 59672633-59672633 on Build GRCh38
rs984515241 Chr.15: 59672662-59672662 on Build GRCh38
rs372011900 Chr.15: 59672645-59672645 on Build GRCh38
rs950030060 Chr.15: 59672339-59672339 on Build GRCh38
rs966727386 Chr.15: 59672276-59672276 on Build GRCh38
rs777653663 Chr.15: 59672667-59672667 on Build GRCh38
rs770252466 Chr.15: 59672432-59672432 on Build GRCh38
rs991722508 Chr.15: 59672548-59672548 on Build GRCh38
rs768103949 Chr.15: 59672472-59672472 on Build GRCh38
rs370180810 Chr.15: 59672594-59672594 on Build GRCh38
rs752931638 Chr.15: 59672651-59672651 on Build GRCh38
rs757186206 Chr.15: 59672672-59672672 on Build GRCh38
rs142902479 Chr.15: 59672449-59672449 on Build GRCh38
rs979505392 Chr.15: 59672277-59672277 on Build GRCh38
rs1045389386 Chr.15: 59672352-59672352 on Build GRCh38
rs1057058 Chr.15: 59672705-59672705 on Build GRCh38
rs374057170 Chr.15: 59672660-59672660 on Build GRCh38
rs767541075 Chr.15: 59672646-59672646 on Build GRCh38
rs185418300 Chr.15: 59672327-59672327 on Build GRCh38
rs761422886 Chr.15: 59672509-59672509 on Build GRCh38
rs902060684 Chr.15: 59672438-59672438 on Build GRCh38

View Full Product Details