Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32437294-32437845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFC
Entrez Gene ID
254240
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
UniGene
Hs.372939
Gene Chromosome Location
Chr.22: 32413845-32464465 on Build GRCh38
Transcripts
NM_174932.2 XM_011530088.2 XM_011530089.1 XM_011530090.1 XM_011530091.2 XM_017028740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612384 c.674A>G p.N225S Chr.22: 32437833-32437833 on Build GRCh38
363934 c.720T>C p.T240T Chr.22: 32437787-32437787 on Build GRCh38
5821304 c.686T>A p.L229Q Chr.22: 32437821-32437821 on Build GRCh38
5987303 c.712G>C p.E238Q Chr.22: 32437795-32437795 on Build GRCh38
2151849 c.704G>A p.S235N Chr.22: 32437803-32437803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943765589 Chr.22: 32437368-32437373 on Build GRCh38
rs183083259 Chr.22: 32437567-32437567 on Build GRCh38
rs375143925 Chr.22: 32437727-32437727 on Build GRCh38
rs114522460 Chr.22: 32437525-32437525 on Build GRCh38
rs738262 Chr.22: 32437338-32437338 on Build GRCh38
rs913836371 Chr.22: 32437391-32437391 on Build GRCh38
rs903402357 Chr.22: 32437437-32437437 on Build GRCh38
rs920922491 Chr.22: 32437400-32437400 on Build GRCh38
rs975567100 Chr.22: 32437576-32437576 on Build GRCh38
rs1047892593 Chr.22: 32437760-32437760 on Build GRCh38
rs951223437 Chr.22: 32437334-32437334 on Build GRCh38
rs187642564 Chr.22: 32437581-32437581 on Build GRCh38
rs1044880158 Chr.22: 32437728-32437728 on Build GRCh38
rs777721154 Chr.22: 32437797-32437797 on Build GRCh38
rs945286342 Chr.22: 32437396-32437396 on Build GRCh38
rs933006043 Chr.22: 32437495-32437495 on Build GRCh38
rs906041490 Chr.22: 32437575-32437575 on Build GRCh38
rs117472294 Chr.22: 32437767-32437767 on Build GRCh38
rs184218547 Chr.22: 32437730-32437730 on Build GRCh38
rs899162042 Chr.22: 32437497-32437497 on Build GRCh38
rs550921651 Chr.22: 32437684-32437684 on Build GRCh38
rs988810174 Chr.22: 32437390-32437390 on Build GRCh38
rs750720714 Chr.22: 32437716-32437716 on Build GRCh38
rs1035746826 Chr.22: 32437347-32437347 on Build GRCh38
rs1043332379 Chr.22: 32437436-32437436 on Build GRCh38
rs568464272 Chr.22: 32437358-32437358 on Build GRCh38
rs756628108 Chr.22: 32437720-32437720 on Build GRCh38
rs534563215 Chr.22: 32437504-32437504 on Build GRCh38
rs770971560 Chr.22: 32437803-32437803 on Build GRCh38
rs934883500 Chr.22: 32437669-32437669 on Build GRCh38
rs925173993 Chr.22: 32437707-32437707 on Build GRCh38
rs759137807 Chr.22: 32437812-32437812 on Build GRCh38
rs895926781 Chr.22: 32437472-32437472 on Build GRCh38
rs369213069 Chr.22: 32437654-32437654 on Build GRCh38
rs190948213 Chr.22: 32437414-32437414 on Build GRCh38
rs192779703 Chr.22: 32437681-32437681 on Build GRCh38
rs769542386 Chr.22: 32437420-32437420 on Build GRCh38
rs187497322 Chr.22: 32437505-32437505 on Build GRCh38
rs372145332 Chr.22: 32437724-32437724 on Build GRCh38
rs761356119 Chr.22: 32437808-32437808 on Build GRCh38
rs941646531 Chr.22: 32437787-32437787 on Build GRCh38
rs145861963 Chr.22: 32437645-32437645 on Build GRCh38
rs183197095 Chr.22: 32437498-32437498 on Build GRCh38
rs1001686516 Chr.22: 32437579-32437579 on Build GRCh38
rs748412558 Chr.22: 32437757-32437757 on Build GRCh38
rs565547562 Chr.22: 32437483-32437483 on Build GRCh38
rs879173413 Chr.22: 32437687-32437687 on Build GRCh38
rs113666931 Chr.22: 32437742-32437742 on Build GRCh38
rs767000520 Chr.22: 32437819-32437819 on Build GRCh38
rs191822479 Chr.22: 32437535-32437535 on Build GRCh38
rs539568844 Chr.22: 32437463-32437463 on Build GRCh38
rs1039208796 Chr.22: 32437460-32437460 on Build GRCh38
rs746785490 Chr.22: 32437799-32437799 on Build GRCh38
rs895968778 Chr.22: 32437701-32437701 on Build GRCh38
rs989252311 Chr.22: 32437679-32437679 on Build GRCh38
rs539918991 Chr.22: 32437788-32437788 on Build GRCh38
rs759329238 Chr.22: 32437403-32437403 on Build GRCh38
rs556704420 Chr.22: 32437544-32437544 on Build GRCh38
rs138769365 Chr.22: 32437683-32437683 on Build GRCh38
rs372628550 Chr.22: 32437790-32437790 on Build GRCh38
rs912332439 Chr.22: 32437359-32437359 on Build GRCh38
rs763768001 Chr.22: 32437759-32437759 on Build GRCh38
rs925121694 Chr.22: 32437423-32437423 on Build GRCh38
rs1026254325 Chr.22: 32437772-32437772 on Build GRCh38
rs189196951 Chr.22: 32437413-32437413 on Build GRCh38
rs1039398432 Chr.22: 32437399-32437399 on Build GRCh38
rs1057330874 Chr.22: 32437685-32437685 on Build GRCh38
rs551593388 Chr.22: 32437638-32437638 on Build GRCh38
rs558669943 Chr.22: 32437474-32437474 on Build GRCh38
rs960331298 Chr.22: 32437361-32437361 on Build GRCh38
rs375008064 Chr.22: 32437680-32437680 on Build GRCh38
rs773506493 Chr.22: 32437401-32437402 on Build GRCh38
rs754989246 Chr.22: 32437738-32437738 on Build GRCh38
rs570888963 Chr.22: 32437445-32437445 on Build GRCh38

View Full Product Details