Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220062796-220063393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPNT1
Entrez Gene ID
10380
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Gene Aliases
HEL20, PIP
UniGene
Hs.406134
Gene Chromosome Location
Chr.1: 220057482-220089916 on Build GRCh38
Transcripts
NM_001286149.1 NM_001286150.1 NM_001286151.1 NM_006085.5 XM_005272998.2 XM_005272999.4 XM_005273000.4 XM_005273004.2 XM_005273005.4 XM_006711114.3 XM_011509063.2 XM_011509064.2 XM_011509065.2 XM_017000043.1 XM_017000044.1 XM_017000045.1 XM_017000046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5518249 c.475-10G>A p.? Chr.1: 220062964-220062964 on Build GRCh38
4552154 c.550G>A p.E184K Chr.1: 220062879-220062879 on Build GRCh38
904096 c.488C>T p.A163V Chr.1: 220062941-220062941 on Build GRCh38
1601660 c.522T>G p.G174G Chr.1: 220062907-220062907 on Build GRCh38
140401 c.541C>T p.Q181* Chr.1: 220062888-220062888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376277086 Chr.1: 220062868-220062868 on Build GRCh38
rs529468135 Chr.1: 220063039-220063039 on Build GRCh38
rs755318945 Chr.1: 220062978-220062978 on Build GRCh38
rs747952992 Chr.1: 220062901-220062901 on Build GRCh38
rs760263600 Chr.1: 220062920-220062920 on Build GRCh38
rs998983138 Chr.1: 220063365-220063365 on Build GRCh38
rs887004660 Chr.1: 220063108-220063108 on Build GRCh38
rs890810015 Chr.1: 220063164-220063164 on Build GRCh38
rs780357800 Chr.1: 220062885-220062885 on Build GRCh38
rs111614124 Chr.1: 220062903-220062903 on Build GRCh38
rs41274792 Chr.1: 220063202-220063202 on Build GRCh38
rs977975850 Chr.1: 220063172-220063172 on Build GRCh38
rs774470035 Chr.1: 220062868-220062868 on Build GRCh38
rs968850012 Chr.1: 220063167-220063167 on Build GRCh38
rs979253738 Chr.1: 220062976-220062976 on Build GRCh38
rs772141345 Chr.1: 220062910-220062910 on Build GRCh38
rs751905267 Chr.1: 220062835-220062835 on Build GRCh38
rs746966907 Chr.1: 220063004-220063004 on Build GRCh38
rs765888676 Chr.1: 220063114-220063114 on Build GRCh38
rs750858898 Chr.1: 220062964-220062964 on Build GRCh38
rs953843853 Chr.1: 220063316-220063316 on Build GRCh38
rs535995601 Chr.1: 220063223-220063223 on Build GRCh38
rs1025493234 Chr.1: 220063086-220063086 on Build GRCh38
rs960712096 Chr.1: 220063265-220063265 on Build GRCh38
rs764862721 Chr.1: 220062936-220062936 on Build GRCh38
rs373153863 Chr.1: 220062957-220062957 on Build GRCh38
rs201898401 Chr.1: 220062960-220062960 on Build GRCh38
rs760470365 Chr.1: 220062971-220062973 on Build GRCh38
rs765846743 Chr.1: 220062921-220062921 on Build GRCh38
rs759153119 Chr.1: 220063189-220063189 on Build GRCh38
rs894403854 Chr.1: 220063326-220063326 on Build GRCh38
rs969535183 Chr.1: 220062875-220062875 on Build GRCh38
rs762601460 Chr.1: 220063109-220063109 on Build GRCh38
rs1039402108 Chr.1: 220063205-220063205 on Build GRCh38
rs986568288 Chr.1: 220063338-220063338 on Build GRCh38
rs919645388 Chr.1: 220063209-220063209 on Build GRCh38
rs185174323 Chr.1: 220063289-220063289 on Build GRCh38
rs756616704 Chr.1: 220062965-220062965 on Build GRCh38
rs759241506 Chr.1: 220062926-220062926 on Build GRCh38
rs939678676 Chr.1: 220063343-220063343 on Build GRCh38
rs763775741 Chr.1: 220062984-220062986 on Build GRCh38
rs969195780 Chr.1: 220063359-220063359 on Build GRCh38
rs751921923 Chr.1: 220062943-220062943 on Build GRCh38
rs921129137 Chr.1: 220063304-220063304 on Build GRCh38
rs1047321052 Chr.1: 220063270-220063270 on Build GRCh38
rs749514995 Chr.1: 220062892-220062892 on Build GRCh38
rs757555550 Chr.1: 220062946-220062946 on Build GRCh38
rs566645484 Chr.1: 220063283-220063283 on Build GRCh38
rs779210199 Chr.1: 220062980-220062980 on Build GRCh38
rs931099907 Chr.1: 220063249-220063249 on Build GRCh38
rs575140868 Chr.1: 220063352-220063352 on Build GRCh38
rs991451502 Chr.1: 220063162-220063162 on Build GRCh38
rs995489804 Chr.1: 220063193-220063193 on Build GRCh38
rs1012839516 Chr.1: 220063331-220063331 on Build GRCh38
rs569202809 Chr.1: 220063175-220063175 on Build GRCh38
rs7530481 Chr.1: 220063279-220063279 on Build GRCh38
rs773972399 Chr.1: 220062846-220062846 on Build GRCh38
rs777591805 Chr.1: 220063003-220063003 on Build GRCh38
rs750041214 Chr.1: 220063074-220063074 on Build GRCh38
rs571743830 Chr.1: 220063188-220063188 on Build GRCh38
rs567257978 Chr.1: 220063251-220063251 on Build GRCh38
rs530932883 Chr.1: 220063192-220063192 on Build GRCh38
rs757823489 Chr.1: 220062842-220062842 on Build GRCh38
rs530671710 Chr.1: 220063120-220063120 on Build GRCh38
rs543343412 Chr.1: 220063056-220063056 on Build GRCh38
rs774348278 Chr.1: 220062900-220062900 on Build GRCh38
rs116650800 Chr.1: 220063342-220063342 on Build GRCh38
rs7516345 Chr.1: 220063302-220063302 on Build GRCh38
rs919583480 Chr.1: 220063356-220063356 on Build GRCh38
rs563888320 Chr.1: 220062858-220062858 on Build GRCh38
rs772974713 Chr.1: 220062915-220062915 on Build GRCh38
rs113725800 Chr.1: 220063165-220063165 on Build GRCh38
rs768980706 Chr.1: 220062899-220062899 on Build GRCh38
rs771723467 Chr.1: 220062991-220062991 on Build GRCh38
rs924833938 Chr.1: 220063173-220063173 on Build GRCh38
rs921087504 Chr.1: 220063035-220063035 on Build GRCh38
rs747979127 Chr.1: 220062986-220062986 on Build GRCh38
rs1036735858 Chr.1: 220063355-220063355 on Build GRCh38
rs190930380 Chr.1: 220063124-220063124 on Build GRCh38
rs932427611 Chr.1: 220063098-220063098 on Build GRCh38
rs753955861 Chr.1: 220062975-220062975 on Build GRCh38
rs1002274150 Chr.1: 220063211-220063211 on Build GRCh38
rs377034351 Chr.1: 220063176-220063176 on Build GRCh38
rs180774513 Chr.1: 220063222-220063222 on Build GRCh38
rs368958544 Chr.1: 220062924-220062924 on Build GRCh38

View Full Product Details