Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 28241285-28241834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRE
Entrez Gene ID
9577
Gene Name
brain and reproductive organ-expressed (TNFRSF1A modulator)
Gene Aliases
BRCC4, BRCC45
UniGene
Hs.258314
Gene Chromosome Location
Chr.2: 27890615-28338901 on Build GRCh38
Transcripts
NM_001261840.1 NM_004899.4 NM_199191.2 NM_199192.2 NM_199193.2 NM_199194.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1407557 c.790G>A p.V264M Chr.2: 28241332-28241332 on Build GRCh38
6542471 c.815G>T p.R272I Chr.2: 28241357-28241357 on Build GRCh38
5718882 c.832T>C p.F278L Chr.2: 28241374-28241374 on Build GRCh38
1614744 c.816A>G p.R272R Chr.2: 28241358-28241358 on Build GRCh38
4572692 c.831T>C p.A277A Chr.2: 28241373-28241373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561746143 Chr.2: 28241503-28241503 on Build GRCh38
rs377057013 Chr.2: 28241785-28241785 on Build GRCh38
rs765406615 Chr.2: 28241430-28241430 on Build GRCh38
rs28997579 Chr.2: 28241331-28241331 on Build GRCh38
rs915351039 Chr.2: 28241752-28241752 on Build GRCh38
rs746813255 Chr.2: 28241347-28241347 on Build GRCh38
rs557217675 Chr.2: 28241736-28241736 on Build GRCh38
rs1056673932 Chr.2: 28241640-28241640 on Build GRCh38
rs895265878 Chr.2: 28241644-28241644 on Build GRCh38
rs961356145 Chr.2: 28241701-28241701 on Build GRCh38
rs1007994510 Chr.2: 28241740-28241740 on Build GRCh38
rs988410371 Chr.2: 28241525-28241525 on Build GRCh38
rs756059829 Chr.2: 28241714-28241715 on Build GRCh38
rs764699762 Chr.2: 28241308-28241308 on Build GRCh38
rs902794070 Chr.2: 28241598-28241598 on Build GRCh38
rs566513101 Chr.2: 28241653-28241653 on Build GRCh38
rs758549472 Chr.2: 28241445-28241445 on Build GRCh38
rs914182663 Chr.2: 28241562-28241562 on Build GRCh38
rs569206944 Chr.2: 28241526-28241526 on Build GRCh38
rs746140512 Chr.2: 28241332-28241332 on Build GRCh38
rs775928771 Chr.2: 28241416-28241416 on Build GRCh38
rs376402633 Chr.2: 28241503-28241503 on Build GRCh38
rs535680845 Chr.2: 28241710-28241710 on Build GRCh38
rs995573020 Chr.2: 28241735-28241735 on Build GRCh38
rs896335712 Chr.2: 28241684-28241684 on Build GRCh38
rs79517004 Chr.2: 28241775-28241775 on Build GRCh38
rs185350860 Chr.2: 28241523-28241523 on Build GRCh38
rs932752320 Chr.2: 28241471-28241471 on Build GRCh38
rs1004196177 Chr.2: 28241602-28241602 on Build GRCh38
rs6737817 Chr.2: 28241568-28241568 on Build GRCh38
rs781251587 Chr.2: 28241357-28241357 on Build GRCh38
rs34916548 Chr.2: 28241517-28241518 on Build GRCh38
rs564570842 Chr.2: 28241391-28241391 on Build GRCh38
rs142594193 Chr.2: 28241656-28241656 on Build GRCh38
rs372205006 Chr.2: 28241393-28241393 on Build GRCh38
rs895331772 Chr.2: 28241801-28241801 on Build GRCh38
rs1012629724 Chr.2: 28241652-28241660 on Build GRCh38
rs979804940 Chr.2: 28241788-28241788 on Build GRCh38
rs147449945 Chr.2: 28241325-28241325 on Build GRCh38
rs144070701 Chr.2: 28241346-28241346 on Build GRCh38
rs922424227 Chr.2: 28241455-28241455 on Build GRCh38
rs753768977 Chr.2: 28241407-28241407 on Build GRCh38
rs768523754 Chr.2: 28241350-28241350 on Build GRCh38
rs766915777 Chr.2: 28241597-28241597 on Build GRCh38
rs762848994 Chr.2: 28241772-28241772 on Build GRCh38
rs761768530 Chr.2: 28241424-28241424 on Build GRCh38
rs1019100882 Chr.2: 28241742-28241742 on Build GRCh38
rs574689008 Chr.2: 28241683-28241683 on Build GRCh38
rs113647214 Chr.2: 28241521-28241521 on Build GRCh38
rs984018543 Chr.2: 28241385-28241385 on Build GRCh38
rs977032484 Chr.2: 28241702-28241702 on Build GRCh38
rs750500355 Chr.2: 28241436-28241436 on Build GRCh38
rs551110433 Chr.2: 28241588-28241588 on Build GRCh38
rs114295720 Chr.2: 28241423-28241423 on Build GRCh38
rs764683465 Chr.2: 28241420-28241420 on Build GRCh38
rs375906817 Chr.2: 28241404-28241404 on Build GRCh38
rs372830652 Chr.2: 28241783-28241783 on Build GRCh38
rs926699158 Chr.2: 28241622-28241622 on Build GRCh38
rs926654604 Chr.2: 28241789-28241789 on Build GRCh38
rs898440323 Chr.2: 28241733-28241733 on Build GRCh38
rs879031870 Chr.2: 28241502-28241503 on Build GRCh38
rs938066413 Chr.2: 28241803-28241803 on Build GRCh38
rs13409938 Chr.2: 28241401-28241401 on Build GRCh38
rs953888120 Chr.2: 28241737-28241737 on Build GRCh38
rs1004059870 Chr.2: 28241704-28241704 on Build GRCh38
rs759104636 Chr.2: 28241696-28241696 on Build GRCh38
rs1036889011 Chr.2: 28241707-28241707 on Build GRCh38
rs748390739 Chr.2: 28241417-28241417 on Build GRCh38
rs189773595 Chr.2: 28241533-28241533 on Build GRCh38
rs555478963 Chr.2: 28241669-28241669 on Build GRCh38
rs1015666059 Chr.2: 28241686-28241686 on Build GRCh38
rs181507141 Chr.2: 28241415-28241415 on Build GRCh38
rs753801674 Chr.2: 28241316-28241316 on Build GRCh38
rs985560482 Chr.2: 28241749-28241749 on Build GRCh38
rs747609104 Chr.2: 28241365-28241365 on Build GRCh38
rs758642407 Chr.2: 28241343-28241343 on Build GRCh38

View Full Product Details