Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10125555-10126134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRK1
Entrez Gene ID
55845
Gene Name
BRICK1, SCAR/WAVE actin nucleating complex subunit
Gene Aliases
C3orf10, HSPC300, MDS027, hHBrk1
UniGene
Hs.649307
Gene Chromosome Location
Chr.3: 10115649-10127190 on Build GRCh38
Transcripts
NM_018462.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4188315 c.124T>C p.S42P Chr.3: 10125631-10125631 on Build GRCh38
4188316 c.124T>C p.S42P Chr.3: 10125631-10125631 on Build GRCh38
6855323 c.194A>C p.E65A Chr.3: 10125701-10125701 on Build GRCh38
6835925 c.123G>A p.M41I Chr.3: 10125630-10125630 on Build GRCh38
6855322 c.194A>C p.E65A Chr.3: 10125701-10125701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932501031 Chr.3: 10126086-10126086 on Build GRCh38
rs534956106 Chr.3: 10125907-10125907 on Build GRCh38
rs56274701 Chr.3: 10126106-10126106 on Build GRCh38
rs1038335832 Chr.3: 10126084-10126084 on Build GRCh38
rs77709987 Chr.3: 10125635-10125635 on Build GRCh38
rs945646652 Chr.3: 10125636-10125636 on Build GRCh38
rs1055502617 Chr.3: 10125747-10125747 on Build GRCh38
rs111463051 Chr.3: 10125842-10125842 on Build GRCh38
rs370623683 Chr.3: 10125838-10125838 on Build GRCh38
rs539940424 Chr.3: 10125880-10125880 on Build GRCh38
rs1055257216 Chr.3: 10126054-10126054 on Build GRCh38
rs543974653 Chr.3: 10125955-10125955 on Build GRCh38
rs376431899 Chr.3: 10125688-10125688 on Build GRCh38
rs899703237 Chr.3: 10125847-10125847 on Build GRCh38
rs774183208 Chr.3: 10125644-10125644 on Build GRCh38
rs374159838 Chr.3: 10125867-10125867 on Build GRCh38
rs575874115 Chr.3: 10125698-10125698 on Build GRCh38
rs760115323 Chr.3: 10125748-10125748 on Build GRCh38
rs774284433 Chr.3: 10125756-10125756 on Build GRCh38
rs144305767 Chr.3: 10125837-10125837 on Build GRCh38
rs747437945 Chr.3: 10125706-10125706 on Build GRCh38
rs917011089 Chr.3: 10125995-10125995 on Build GRCh38
rs542290561 Chr.3: 10125828-10125828 on Build GRCh38
rs529993022 Chr.3: 10125983-10125983 on Build GRCh38
rs769551856 Chr.3: 10125724-10125724 on Build GRCh38
rs766384866 Chr.3: 10125638-10125638 on Build GRCh38
rs757358992 Chr.3: 10125676-10125676 on Build GRCh38
rs183664178 Chr.3: 10126071-10126071 on Build GRCh38
rs767226255 Chr.3: 10125654-10125654 on Build GRCh38
rs796089433 Chr.3: 10125816-10125816 on Build GRCh38
rs1002404029 Chr.3: 10125920-10125920 on Build GRCh38
rs938148174 Chr.3: 10125813-10125813 on Build GRCh38
rs998527288 Chr.3: 10125888-10125888 on Build GRCh38
rs1005758749 Chr.3: 10125812-10125812 on Build GRCh38
rs1047393955 Chr.3: 10125801-10125801 on Build GRCh38
rs749992128 Chr.3: 10125893-10125893 on Build GRCh38
rs868334279 Chr.3: 10126030-10126030 on Build GRCh38
rs937099527 Chr.3: 10125730-10125730 on Build GRCh38
rs746376520 Chr.3: 10125683-10125683 on Build GRCh38
rs567655077 Chr.3: 10125989-10125989 on Build GRCh38
rs559866454 Chr.3: 10126048-10126048 on Build GRCh38
rs949750917 Chr.3: 10125773-10125773 on Build GRCh38
rs772839104 Chr.3: 10125623-10125623 on Build GRCh38
rs896802705 Chr.3: 10125886-10125886 on Build GRCh38
rs1009581123 Chr.3: 10125937-10125937 on Build GRCh38
rs373926203 Chr.3: 10125652-10125652 on Build GRCh38
rs773043719 Chr.3: 10125727-10125727 on Build GRCh38
rs570610886 Chr.3: 10125894-10125894 on Build GRCh38
rs770481519 Chr.3: 10125786-10125786 on Build GRCh38
rs953865262 Chr.3: 10125891-10125891 on Build GRCh38
rs557711772 Chr.3: 10125603-10125603 on Build GRCh38
rs748762403 Chr.3: 10125738-10125738 on Build GRCh38
rs765736503 Chr.3: 10125975-10125975 on Build GRCh38
rs779426546 Chr.3: 10125682-10125682 on Build GRCh38
rs772347028 Chr.3: 10125587-10125587 on Build GRCh38
rs562666767 Chr.3: 10125979-10125979 on Build GRCh38
rs1029550444 Chr.3: 10125889-10125889 on Build GRCh38
rs764357880 Chr.3: 10125671-10125671 on Build GRCh38
rs916910206 Chr.3: 10125758-10125758 on Build GRCh38
rs67667957 Chr.3: 10125580-10125580 on Build GRCh38
rs1021598694 Chr.3: 10125951-10125951 on Build GRCh38
rs375307844 Chr.3: 10126039-10126039 on Build GRCh38
rs148406892 Chr.3: 10125987-10125987 on Build GRCh38
rs189298450 Chr.3: 10126080-10126080 on Build GRCh38
rs776290004 Chr.3: 10125591-10125591 on Build GRCh38
rs891232024 Chr.3: 10126104-10126105 on Build GRCh38
rs949728527 Chr.3: 10125996-10125996 on Build GRCh38
rs756323273 Chr.3: 10125660-10125660 on Build GRCh38
rs754355310 Chr.3: 10125614-10125615 on Build GRCh38
rs979974499 Chr.3: 10125966-10125966 on Build GRCh38
rs770366283 Chr.3: 10125752-10125752 on Build GRCh38
rs1051150587 Chr.3: 10126099-10126103 on Build GRCh38
rs764512749 Chr.3: 10125590-10125591 on Build GRCh38
rs762604698 Chr.3: 10125629-10125629 on Build GRCh38
rs56087600 Chr.3: 10125892-10125892 on Build GRCh38
rs938264990 Chr.3: 10125998-10125998 on Build GRCh38
rs748999397 Chr.3: 10125831-10125831 on Build GRCh38
rs369470919 Chr.3: 10125621-10125621 on Build GRCh38
rs928137449 Chr.3: 10125808-10125808 on Build GRCh38
rs1054804942 Chr.3: 10125829-10125829 on Build GRCh38
rs752896650 Chr.3: 10125659-10125659 on Build GRCh38
rs936657258 Chr.3: 10125611-10125611 on Build GRCh38
rs887050084 Chr.3: 10125809-10125809 on Build GRCh38

View Full Product Details