Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93241903-93242452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
UniGene
Hs.525549
Gene Chromosome Location
Chr.14: 93237550-93333092 on Build GRCh38
Transcripts
NM_001002860.3 NM_001289133.1 XM_011536939.2 XM_017021438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4997901 c.3259T>G p.S1087A Chr.14: 93242413-93242413 on Build GRCh38
7416413 c.3275C>T p.A1092V Chr.14: 93242397-93242397 on Build GRCh38
6613405 c.2204G>A p.R735K Chr.14: 93242415-93242415 on Build GRCh38
7416414 c.2222C>T p.A741V Chr.14: 93242397-93242397 on Build GRCh38
1707742 c.3392C>T p.A1131V Chr.14: 93242280-93242280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920287707 Chr.14: 93242210-93242210 on Build GRCh38
rs775432782 Chr.14: 93242393-93242393 on Build GRCh38
rs931590474 Chr.14: 93242321-93242321 on Build GRCh38
rs182953761 Chr.14: 93242033-93242033 on Build GRCh38
rs780432278 Chr.14: 93242086-93242087 on Build GRCh38
rs995952080 Chr.14: 93242056-93242056 on Build GRCh38
rs746372340 Chr.14: 93242254-93242254 on Build GRCh38
rs536674457 Chr.14: 93242330-93242330 on Build GRCh38
rs1009344456 Chr.14: 93242143-93242143 on Build GRCh38
rs898729103 Chr.14: 93242047-93242051 on Build GRCh38
rs905824731 Chr.14: 93242087-93242087 on Build GRCh38
rs767237944 Chr.14: 93242423-93242423 on Build GRCh38
rs79182521 Chr.14: 93242097-93242097 on Build GRCh38
rs960489741 Chr.14: 93241954-93241955 on Build GRCh38
rs961811663 Chr.14: 93242099-93242099 on Build GRCh38
rs139234606 Chr.14: 93242391-93242391 on Build GRCh38
rs773164838 Chr.14: 93242373-93242373 on Build GRCh38
rs550121723 Chr.14: 93242019-93242019 on Build GRCh38
rs141712356 Chr.14: 93241988-93241988 on Build GRCh38
rs200447121 Chr.14: 93242410-93242410 on Build GRCh38
rs878948142 Chr.14: 93242382-93242382 on Build GRCh38
rs781441678 Chr.14: 93242332-93242332 on Build GRCh38
rs150461526 Chr.14: 93242070-93242070 on Build GRCh38
rs774736629 Chr.14: 93242269-93242269 on Build GRCh38
rs967990284 Chr.14: 93242188-93242188 on Build GRCh38
rs958833616 Chr.14: 93242075-93242075 on Build GRCh38
rs904511715 Chr.14: 93242041-93242041 on Build GRCh38
rs565194914 Chr.14: 93242422-93242422 on Build GRCh38
rs769331839 Chr.14: 93242259-93242260 on Build GRCh38
rs376927108 Chr.14: 93242150-93242150 on Build GRCh38
rs756695238 Chr.14: 93242320-93242320 on Build GRCh38
rs115757099 Chr.14: 93242224-93242224 on Build GRCh38
rs375684238 Chr.14: 93242174-93242174 on Build GRCh38
rs36099812 Chr.14: 93242413-93242413 on Build GRCh38
rs397753241 Chr.14: 93242098-93242099 on Build GRCh38
rs891867098 Chr.14: 93242376-93242376 on Build GRCh38
rs780658199 Chr.14: 93242341-93242341 on Build GRCh38
rs780275795 Chr.14: 93242257-93242257 on Build GRCh38
rs1021315936 Chr.14: 93242171-93242171 on Build GRCh38
rs761945122 Chr.14: 93242282-93242282 on Build GRCh38
rs751213966 Chr.14: 93242319-93242319 on Build GRCh38
rs979543752 Chr.14: 93242219-93242219 on Build GRCh38
rs779210366 Chr.14: 93242361-93242361 on Build GRCh38
rs776575899 Chr.14: 93242291-93242291 on Build GRCh38
rs769173156 Chr.14: 93242351-93242351 on Build GRCh38
rs185722120 Chr.14: 93242300-93242300 on Build GRCh38
rs759379749 Chr.14: 93242294-93242294 on Build GRCh38
rs556897574 Chr.14: 93242369-93242369 on Build GRCh38
rs749620723 Chr.14: 93242342-93242342 on Build GRCh38
rs778182264 Chr.14: 93242022-93242022 on Build GRCh38
rs772050304 Chr.14: 93242290-93242290 on Build GRCh38
rs1035713948 Chr.14: 93242098-93242098 on Build GRCh38
rs561007666 Chr.14: 93241974-93241974 on Build GRCh38
rs763885374 Chr.14: 93242411-93242411 on Build GRCh38
rs1020781822 Chr.14: 93242398-93242398 on Build GRCh38
rs911479784 Chr.14: 93242303-93242303 on Build GRCh38
rs986558545 Chr.14: 93242232-93242232 on Build GRCh38
rs768977072 Chr.14: 93242260-93242260 on Build GRCh38
rs907858220 Chr.14: 93242026-93242053 on Build GRCh38
rs750013529 Chr.14: 93242337-93242337 on Build GRCh38
rs762789222 Chr.14: 93242305-93242305 on Build GRCh38
rs921054862 Chr.14: 93242016-93242016 on Build GRCh38
rs752405705 Chr.14: 93242304-93242304 on Build GRCh38
rs151255842 Chr.14: 93242318-93242318 on Build GRCh38
rs756545835 Chr.14: 93242338-93242338 on Build GRCh38
rs376946890 Chr.14: 93242256-93242256 on Build GRCh38
rs538910114 Chr.14: 93242046-93242046 on Build GRCh38
rs547061731 Chr.14: 93242094-93242094 on Build GRCh38
rs748396513 Chr.14: 93242368-93242368 on Build GRCh38
rs370176950 Chr.14: 93242225-93242225 on Build GRCh38
rs932384918 Chr.14: 93242018-93242018 on Build GRCh38
rs901264778 Chr.14: 93241996-93241996 on Build GRCh38
rs971766595 Chr.14: 93242133-93242133 on Build GRCh38

View Full Product Details