Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 92142491-92143044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG1
Entrez Gene ID
694
Gene Name
BTG anti-proliferation factor 1
Gene Aliases
APRO2
UniGene
Hs.255935
Gene Chromosome Location
Chr.12: 92140278-92145897 on Build GRCh38
Transcripts
NM_001731.2 NR_046160.2 NR_132339.1 NR_132340.1 NR_132341.1

Gene Symbol
LINC01619
Entrez Gene ID
256021
Gene Name
long intergenic non-protein coding RNA 1619
Gene Aliases
C12orf79
UniGene
Hs.651357
Gene Chromosome Location
Chr.12: 91984976-92142831 on Build GRCh38
Transcripts
NM_001731.2 NR_046160.2 NR_132339.1 NR_132340.1 NR_132341.1

SNP Detail

SNP ID Chromosome Location
rs1435617 Chr.12: 92142738-92142738 on Build GRCh38
rs904273700 Chr.12: 92142880-92142880 on Build GRCh38
rs763052876 Chr.12: 92142911-92142911 on Build GRCh38
rs552692517 Chr.12: 92142686-92142686 on Build GRCh38
rs769861060 Chr.12: 92142792-92142792 on Build GRCh38
rs1056341890 Chr.12: 92142762-92142762 on Build GRCh38
rs750930530 Chr.12: 92142901-92142903 on Build GRCh38
rs1038982368 Chr.12: 92142525-92142525 on Build GRCh38
rs989895372 Chr.12: 92142938-92142938 on Build GRCh38
rs182299506 Chr.12: 92142564-92142564 on Build GRCh38
rs985418433 Chr.12: 92142848-92142848 on Build GRCh38
rs1030575777 Chr.12: 92142561-92142561 on Build GRCh38
rs921961403 Chr.12: 92142799-92142799 on Build GRCh38
rs543966662 Chr.12: 92142797-92142797 on Build GRCh38
rs746953832 Chr.12: 92142735-92142735 on Build GRCh38
rs190527754 Chr.12: 92142559-92142559 on Build GRCh38
rs997894270 Chr.12: 92142800-92142800 on Build GRCh38
rs555587169 Chr.12: 92142771-92142771 on Build GRCh38
rs1034093841 Chr.12: 92142887-92142887 on Build GRCh38
rs771143486 Chr.12: 92143006-92143006 on Build GRCh38
rs541764824 Chr.12: 92142754-92142754 on Build GRCh38
rs532447480 Chr.12: 92142969-92142969 on Build GRCh38
rs983905629 Chr.12: 92142699-92142699 on Build GRCh38
rs922473545 Chr.12: 92142633-92142633 on Build GRCh38
rs1034389255 Chr.12: 92142669-92142669 on Build GRCh38
rs989928297 Chr.12: 92142683-92142683 on Build GRCh38
rs892037071 Chr.12: 92142838-92142839 on Build GRCh38
rs76835213 Chr.12: 92142585-92142585 on Build GRCh38
rs570546936 Chr.12: 92142631-92142631 on Build GRCh38
rs955328035 Chr.12: 92142645-92142645 on Build GRCh38
rs549648366 Chr.12: 92142611-92142611 on Build GRCh38
rs935718687 Chr.12: 92142673-92142673 on Build GRCh38
rs1010392559 Chr.12: 92142590-92142590 on Build GRCh38
rs1043225853 Chr.12: 92142874-92142874 on Build GRCh38
rs900756670 Chr.12: 92142540-92142540 on Build GRCh38
rs955446693 Chr.12: 92142828-92142828 on Build GRCh38
rs563474957 Chr.12: 92142886-92142886 on Build GRCh38
rs952099348 Chr.12: 92142558-92142558 on Build GRCh38
rs748548933 Chr.12: 92142897-92142910 on Build GRCh38
rs1010424960 Chr.12: 92142867-92142867 on Build GRCh38
rs909313497 Chr.12: 92142708-92142708 on Build GRCh38
rs1030346880 Chr.12: 92142809-92142809 on Build GRCh38
rs139646823 Chr.12: 92142610-92142610 on Build GRCh38
rs951271162 Chr.12: 92142988-92142988 on Build GRCh38
rs928336863 Chr.12: 92143004-92143004 on Build GRCh38
rs1018108436 Chr.12: 92142852-92142852 on Build GRCh38
rs981485389 Chr.12: 92142990-92142990 on Build GRCh38
rs80271339 Chr.12: 92142598-92142598 on Build GRCh38
rs745639535 Chr.12: 92142589-92142589 on Build GRCh38
rs531756260 Chr.12: 92142684-92142684 on Build GRCh38
rs1034531295 Chr.12: 92142522-92142522 on Build GRCh38
rs1006246939 Chr.12: 92142790-92142790 on Build GRCh38
rs143359363 Chr.12: 92142671-92142671 on Build GRCh38
rs564690024 Chr.12: 92142516-92142516 on Build GRCh38
rs77126965 Chr.12: 92142983-92142983 on Build GRCh38
rs912945981 Chr.12: 92142939-92142939 on Build GRCh38
rs756985808 Chr.12: 92142524-92142524 on Build GRCh38
rs1052471826 Chr.12: 92142821-92142821 on Build GRCh38
rs979781046 Chr.12: 92142667-92142667 on Build GRCh38
rs575116654 Chr.12: 92142772-92142772 on Build GRCh38
rs1014858597 Chr.12: 92142581-92142581 on Build GRCh38
rs985670892 Chr.12: 92142647-92142647 on Build GRCh38
rs573134218 Chr.12: 92142753-92142753 on Build GRCh38

View Full Product Details