Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26445666-26446232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
XM_011514268.2 NM_001242803.1 NM_006994.4 NM_197974.2

Gene Symbol
BTN3A3
Entrez Gene ID
10384
Gene Name
butyrophilin subfamily 3 member A3
Gene Aliases
BTF3, BTN3.3
UniGene
Hs.167741
Gene Chromosome Location
Chr.6: 26440472-26453415 on Build GRCh38
Transcripts
XM_011514268.2 NM_001242803.1 NM_006994.4 NM_197974.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3761749 c.711C>T p.I237I Chr.6: 26445981-26445981 on Build GRCh38
4666401 c.682G>A p.G228S Chr.6: 26445952-26445952 on Build GRCh38
1442667 c.697G>A p.A233T Chr.6: 26445967-26445967 on Build GRCh38
5869906 c.478G>A p.G160R Chr.6: 26445748-26445748 on Build GRCh38
4836646 c.563C>T p.P188L Chr.6: 26445833-26445833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761543235 Chr.6: 26445895-26445895 on Build GRCh38
rs200625612 Chr.6: 26445944-26445944 on Build GRCh38
rs770145744 Chr.6: 26445959-26445959 on Build GRCh38
rs45492394 Chr.6: 26446041-26446041 on Build GRCh38
rs373589611 Chr.6: 26445793-26445793 on Build GRCh38
rs752375054 Chr.6: 26445914-26445914 on Build GRCh38
rs1009846197 Chr.6: 26445974-26445974 on Build GRCh38
rs748293471 Chr.6: 26445780-26445780 on Build GRCh38
rs777169027 Chr.6: 26445791-26445791 on Build GRCh38
rs911431072 Chr.6: 26445749-26445749 on Build GRCh38
rs745877467 Chr.6: 26445715-26445715 on Build GRCh38
rs774580341 Chr.6: 26445756-26445756 on Build GRCh38
rs755788691 Chr.6: 26445750-26445751 on Build GRCh38
rs754865821 Chr.6: 26445909-26445909 on Build GRCh38
rs556261031 Chr.6: 26446102-26446102 on Build GRCh38
rs779840810 Chr.6: 26445734-26445734 on Build GRCh38
rs755096337 Chr.6: 26446209-26446209 on Build GRCh38
rs981261979 Chr.6: 26446144-26446144 on Build GRCh38
rs999661296 Chr.6: 26445994-26445994 on Build GRCh38
rs756840647 Chr.6: 26445705-26445705 on Build GRCh38
rs1014799038 Chr.6: 26446084-26446084 on Build GRCh38
rs920188754 Chr.6: 26445794-26445794 on Build GRCh38
rs539561386 Chr.6: 26446078-26446078 on Build GRCh38
rs531383948 Chr.6: 26445983-26445983 on Build GRCh38
rs751265742 Chr.6: 26445822-26445822 on Build GRCh38
rs758331557 Chr.6: 26445974-26445976 on Build GRCh38
rs144615747 Chr.6: 26445908-26445908 on Build GRCh38
rs751053710 Chr.6: 26445703-26445703 on Build GRCh38
rs749042680 Chr.6: 26445742-26445742 on Build GRCh38
rs759929819 Chr.6: 26445877-26445877 on Build GRCh38
rs3846848 Chr.6: 26445981-26445981 on Build GRCh38
rs150636780 Chr.6: 26445786-26445786 on Build GRCh38
rs570459961 Chr.6: 26445858-26445858 on Build GRCh38
rs773420672 Chr.6: 26445784-26445784 on Build GRCh38
rs949370475 Chr.6: 26446080-26446080 on Build GRCh38
rs759530682 Chr.6: 26445995-26445995 on Build GRCh38
rs765015632 Chr.6: 26445996-26445996 on Build GRCh38
rs770264493 Chr.6: 26445884-26445884 on Build GRCh38
rs747213105 Chr.6: 26445934-26445934 on Build GRCh38
rs929963506 Chr.6: 26445797-26445797 on Build GRCh38
rs140844974 Chr.6: 26445902-26445902 on Build GRCh38
rs749879449 Chr.6: 26445833-26445833 on Build GRCh38
rs757820887 Chr.6: 26445920-26445920 on Build GRCh38
rs556808322 Chr.6: 26446160-26446160 on Build GRCh38
rs766138602 Chr.6: 26445819-26445819 on Build GRCh38
rs748073094 Chr.6: 26445967-26445967 on Build GRCh38
rs201388694 Chr.6: 26445838-26445838 on Build GRCh38
rs765497042 Chr.6: 26445810-26445810 on Build GRCh38
rs1055018769 Chr.6: 26445957-26445957 on Build GRCh38
rs775930138 Chr.6: 26445971-26445971 on Build GRCh38
rs763764002 Chr.6: 26445696-26445696 on Build GRCh38
rs749936162 Chr.6: 26445744-26445751 on Build GRCh38
rs746448961 Chr.6: 26445952-26445952 on Build GRCh38
rs1054726466 Chr.6: 26446159-26446159 on Build GRCh38
rs138065289 Chr.6: 26445783-26445783 on Build GRCh38
rs139877036 Chr.6: 26445979-26445979 on Build GRCh38
rs765977692 Chr.6: 26445988-26445988 on Build GRCh38
rs760126544 Chr.6: 26445906-26445906 on Build GRCh38
rs754962015 Chr.6: 26445725-26445725 on Build GRCh38
rs778094119 Chr.6: 26445926-26445926 on Build GRCh38
rs201382168 Chr.6: 26446002-26446002 on Build GRCh38
rs781128072 Chr.6: 26445949-26445949 on Build GRCh38
rs769944320 Chr.6: 26446122-26446122 on Build GRCh38
rs148026276 Chr.6: 26445728-26445728 on Build GRCh38
rs754846329 Chr.6: 26445846-26445846 on Build GRCh38
rs114160412 Chr.6: 26446076-26446076 on Build GRCh38
rs749586779 Chr.6: 26445973-26445973 on Build GRCh38
rs748468450 Chr.6: 26445857-26445857 on Build GRCh38
rs767075667 Chr.6: 26445832-26445832 on Build GRCh38
rs376066083 Chr.6: 26446018-26446018 on Build GRCh38
rs957532336 Chr.6: 26445946-26445946 on Build GRCh38
rs986912157 Chr.6: 26445740-26445740 on Build GRCh38
rs766933059 Chr.6: 26445788-26445788 on Build GRCh38
rs761241434 Chr.6: 26446124-26446124 on Build GRCh38
rs771589705 Chr.6: 26445875-26445875 on Build GRCh38
rs202187130 Chr.6: 26445972-26445972 on Build GRCh38
rs377686214 Chr.6: 26445834-26445834 on Build GRCh38
rs765849446 Chr.6: 26445907-26445907 on Build GRCh38
rs946959587 Chr.6: 26445975-26445975 on Build GRCh38
rs768788537 Chr.6: 26445746-26445746 on Build GRCh38
rs965704959 Chr.6: 26445820-26445820 on Build GRCh38
rs778813530 Chr.6: 26445964-26445965 on Build GRCh38
rs145416805 Chr.6: 26445986-26445986 on Build GRCh38
rs754845628 Chr.6: 26445959-26445959 on Build GRCh38
rs951920502 Chr.6: 26445841-26445841 on Build GRCh38
rs147467780 Chr.6: 26445916-26445916 on Build GRCh38
rs912956996 Chr.6: 26445700-26445700 on Build GRCh38
rs780652944 Chr.6: 26445714-26445714 on Build GRCh38
rs753487688 Chr.6: 26445918-26445919 on Build GRCh38
rs200139388 Chr.6: 26445951-26445951 on Build GRCh38
rs568339258 Chr.6: 26446024-26446024 on Build GRCh38
rs553737662 Chr.6: 26446047-26446047 on Build GRCh38
rs778010971 Chr.6: 26445869-26445869 on Build GRCh38
rs1047092199 Chr.6: 26446055-26446055 on Build GRCh38
rs763951444 Chr.6: 26446015-26446015 on Build GRCh38
rs541415098 Chr.6: 26445870-26445870 on Build GRCh38
rs745871863 Chr.6: 26446033-26446033 on Build GRCh38
rs778825552 Chr.6: 26445911-26445911 on Build GRCh38
rs1045133111 Chr.6: 26446098-26446098 on Build GRCh38
rs920175391 Chr.6: 26446042-26446042 on Build GRCh38
rs986547716 Chr.6: 26445982-26445982 on Build GRCh38
rs778536905 Chr.6: 26445847-26445847 on Build GRCh38
rs767093669 Chr.6: 26445896-26445896 on Build GRCh38
rs777245190 Chr.6: 26445876-26445876 on Build GRCh38
rs781350485 Chr.6: 26446029-26446029 on Build GRCh38
rs374130285 Chr.6: 26445829-26445829 on Build GRCh38
rs775709582 Chr.6: 26445889-26445889 on Build GRCh38
rs753274261 Chr.6: 26445989-26445989 on Build GRCh38

View Full Product Details