Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49708724-49709178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf53
Entrez Gene ID
282966
Gene Name
chromosome 10 open reading frame 53
Gene Aliases
-
UniGene
Hs.131287
Gene Chromosome Location
Chr.10: 49679638-49708910 on Build GRCh38
Transcripts
NM_182554.2

SNP Detail

SNP ID Chromosome Location
rs1016306451 Chr.10: 49708972-49708972 on Build GRCh38
rs1007745195 Chr.10: 49709082-49709082 on Build GRCh38
rs764481521 Chr.10: 49708742-49708742 on Build GRCh38
rs559334484 Chr.10: 49708751-49708751 on Build GRCh38
rs148824190 Chr.10: 49708782-49708782 on Build GRCh38
rs1258237 Chr.10: 49708941-49708941 on Build GRCh38
rs561080561 Chr.10: 49709012-49709012 on Build GRCh38
rs1006227950 Chr.10: 49708961-49708961 on Build GRCh38
rs1017338950 Chr.10: 49709108-49709108 on Build GRCh38
rs78341607 Chr.10: 49708786-49708786 on Build GRCh38
rs537747657 Chr.10: 49709003-49709003 on Build GRCh38
rs916168191 Chr.10: 49709067-49709067 on Build GRCh38
rs867711258 Chr.10: 49709081-49709081 on Build GRCh38
rs992042663 Chr.10: 49709066-49709066 on Build GRCh38
rs774900757 Chr.10: 49708890-49708890 on Build GRCh38
rs971676581 Chr.10: 49708824-49708824 on Build GRCh38
rs969343359 Chr.10: 49709069-49709069 on Build GRCh38
rs143429749 Chr.10: 49708995-49708995 on Build GRCh38
rs35275788 Chr.10: 49708803-49708804 on Build GRCh38
rs576706391 Chr.10: 49708997-49708997 on Build GRCh38
rs368622393 Chr.10: 49709036-49709036 on Build GRCh38
rs916119446 Chr.10: 49708844-49708844 on Build GRCh38
rs567450695 Chr.10: 49708828-49708828 on Build GRCh38
rs1037595847 Chr.10: 49709043-49709043 on Build GRCh38
rs947763663 Chr.10: 49708845-49708845 on Build GRCh38
rs563715193 Chr.10: 49709002-49709002 on Build GRCh38
rs954427093 Chr.10: 49708832-49708832 on Build GRCh38
rs533125529 Chr.10: 49708764-49708764 on Build GRCh38
rs997748526 Chr.10: 49708894-49708894 on Build GRCh38
rs993603028 Chr.10: 49708999-49708999 on Build GRCh38
rs760925131 Chr.10: 49709080-49709080 on Build GRCh38
rs903622357 Chr.10: 49708881-49708881 on Build GRCh38
rs935335951 Chr.10: 49708951-49708951 on Build GRCh38
rs6537550 Chr.10: 49708788-49708788 on Build GRCh38
rs935117069 Chr.10: 49708883-49708883 on Build GRCh38
rs905019359 Chr.10: 49708783-49708783 on Build GRCh38
rs551281256 Chr.10: 49708773-49708773 on Build GRCh38
rs928824833 Chr.10: 49708787-49708787 on Build GRCh38
rs1050379881 Chr.10: 49708940-49708940 on Build GRCh38
rs1270561 Chr.10: 49708885-49708885 on Build GRCh38
rs571278571 Chr.10: 49708925-49708925 on Build GRCh38
rs980119771 Chr.10: 49708927-49708927 on Build GRCh38
rs964224389 Chr.10: 49708886-49708886 on Build GRCh38
rs79606841 Chr.10: 49708862-49708862 on Build GRCh38
rs961716804 Chr.10: 49708792-49708792 on Build GRCh38
rs905048537 Chr.10: 49709035-49709035 on Build GRCh38

View Full Product Details