Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 76472562-76473121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMSY
Entrez Gene ID
56946
Gene Name
EMSY, BRCA2 interacting transcriptional repressor
Gene Aliases
C11orf30, GL002
UniGene
Hs.352588
Gene Chromosome Location
Chr.11: 76444928-76552901 on Build GRCh38
Transcripts
NM_001300942.1 NM_001300943.1 NM_001300944.1 NM_020193.4 XM_005274112.4 XM_006718637.3 XM_006718638.3 XM_006718639.3 XM_011545173.2 XM_011545174.2 XM_011545175.2 XM_011545177.2 XM_011545178.2 XM_011545179.2 XM_017018021.1 XM_017018022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5951761 c.955C>T p.P319S Chr.11: 76472687-76472687 on Build GRCh38
3986474 c.1063G>T p.V355L Chr.11: 76472795-76472795 on Build GRCh38
344072 c.1108G>T p.G370C Chr.11: 76472840-76472840 on Build GRCh38
5958765 c.922delC p.P308fs*16 Chr.11: 76472654-76472654 on Build GRCh38
932074 c.871C>G p.P291A Chr.11: 76472603-76472603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769410924 Chr.11: 76472756-76472756 on Build GRCh38
rs767488407 Chr.11: 76472810-76472810 on Build GRCh38
rs745420267 Chr.11: 76472754-76472754 on Build GRCh38
rs141073024 Chr.11: 76472784-76472784 on Build GRCh38
rs745505392 Chr.11: 76472674-76472674 on Build GRCh38
rs546166223 Chr.11: 76472991-76472991 on Build GRCh38
rs774663871 Chr.11: 76472679-76472679 on Build GRCh38
rs567012258 Chr.11: 76472942-76472942 on Build GRCh38
rs754657014 Chr.11: 76472876-76472876 on Build GRCh38
rs371086579 Chr.11: 76472874-76472874 on Build GRCh38
rs755576897 Chr.11: 76472839-76472839 on Build GRCh38
rs77657510 Chr.11: 76472752-76472752 on Build GRCh38
rs748920929 Chr.11: 76472765-76472765 on Build GRCh38
rs772695634 Chr.11: 76472625-76472625 on Build GRCh38
rs955055880 Chr.11: 76472955-76472955 on Build GRCh38
rs763097746 Chr.11: 76472799-76472799 on Build GRCh38
rs772340945 Chr.11: 76472681-76472681 on Build GRCh38
rs777718429 Chr.11: 76472654-76472654 on Build GRCh38
rs751374681 Chr.11: 76472804-76472804 on Build GRCh38
rs759642236 Chr.11: 76472788-76472788 on Build GRCh38
rs555144519 Chr.11: 76473086-76473086 on Build GRCh38
rs780674990 Chr.11: 76472742-76472742 on Build GRCh38
rs371859985 Chr.11: 76472629-76472629 on Build GRCh38
rs11236760 Chr.11: 76473008-76473008 on Build GRCh38
rs376862886 Chr.11: 76472733-76472733 on Build GRCh38
rs1046963096 Chr.11: 76472675-76472675 on Build GRCh38
rs779629707 Chr.11: 76472757-76472757 on Build GRCh38
rs757199122 Chr.11: 76472737-76472737 on Build GRCh38
rs766555991 Chr.11: 76472700-76472700 on Build GRCh38
rs762301439 Chr.11: 76472588-76472588 on Build GRCh38
rs150297756 Chr.11: 76473072-76473072 on Build GRCh38
rs373783248 Chr.11: 76473033-76473033 on Build GRCh38
rs1053952389 Chr.11: 76472808-76472808 on Build GRCh38
rs946910772 Chr.11: 76472761-76472761 on Build GRCh38
rs762685294 Chr.11: 76472807-76472807 on Build GRCh38
rs770407618 Chr.11: 76472659-76472659 on Build GRCh38
rs760606508 Chr.11: 76472599-76472599 on Build GRCh38
rs766378308 Chr.11: 76472605-76472605 on Build GRCh38
rs551616019 Chr.11: 76472716-76472716 on Build GRCh38
rs201134518 Chr.11: 76472809-76472809 on Build GRCh38
rs559176752 Chr.11: 76473036-76473036 on Build GRCh38
rs780691761 Chr.11: 76472671-76472671 on Build GRCh38
rs761242993 Chr.11: 76472779-76472779 on Build GRCh38
rs754058859 Chr.11: 76472618-76472618 on Build GRCh38
rs750483714 Chr.11: 76472647-76472647 on Build GRCh38
rs146548337 Chr.11: 76472735-76472735 on Build GRCh38
rs982697881 Chr.11: 76473094-76473094 on Build GRCh38
rs763935049 Chr.11: 76472802-76472802 on Build GRCh38
rs754040400 Chr.11: 76472702-76472702 on Build GRCh38
rs769596604 Chr.11: 76472948-76472948 on Build GRCh38
rs4245444 Chr.11: 76472967-76472967 on Build GRCh38
rs756165213 Chr.11: 76472600-76472600 on Build GRCh38
rs367835459 Chr.11: 76472591-76472591 on Build GRCh38
rs773705768 Chr.11: 76472688-76472688 on Build GRCh38
rs374817161 Chr.11: 76472590-76472590 on Build GRCh38
rs761104169 Chr.11: 76472699-76472699 on Build GRCh38
rs765606023 Chr.11: 76472719-76472719 on Build GRCh38
rs765439410 Chr.11: 76472790-76472790 on Build GRCh38
rs772428940 Chr.11: 76472769-76472769 on Build GRCh38
rs199837796 Chr.11: 76472624-76472624 on Build GRCh38
rs773510937 Chr.11: 76472771-76472771 on Build GRCh38
rs567186633 Chr.11: 76472999-76472999 on Build GRCh38
rs4245443 Chr.11: 76472880-76472880 on Build GRCh38
rs575751963 Chr.11: 76473047-76473047 on Build GRCh38
rs746950889 Chr.11: 76472657-76472657 on Build GRCh38
rs778171222 Chr.11: 76472877-76472877 on Build GRCh38
rs1045223588 Chr.11: 76472774-76472774 on Build GRCh38
rs919831517 Chr.11: 76473045-76473045 on Build GRCh38
rs750378586 Chr.11: 76472828-76472828 on Build GRCh38
rs530680307 Chr.11: 76472858-76472858 on Build GRCh38
rs975823444 Chr.11: 76473065-76473065 on Build GRCh38
rs752991597 Chr.11: 76472726-76472726 on Build GRCh38
rs1007615566 Chr.11: 76472961-76472961 on Build GRCh38
rs762282958 Chr.11: 76472680-76472680 on Build GRCh38
rs752582743 Chr.11: 76472631-76472631 on Build GRCh38
rs117625538 Chr.11: 76472678-76472678 on Build GRCh38
rs377404534 Chr.11: 76472836-76472836 on Build GRCh38
rs777434841 Chr.11: 76472734-76472734 on Build GRCh38

View Full Product Details