Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 555660-556203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNTD2
Entrez Gene ID
256329
Gene Name
lamin tail domain containing 2
Gene Aliases
C11orf35
UniGene
Hs.669395
Gene Chromosome Location
Chr.11: 554850-560780 on Build GRCh38
Transcripts
NM_173573.2 XM_011519965.2 XM_011519967.2 XM_017017476.1 XM_017017477.1 XM_017017478.1 XM_017017479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6614012 c.1325C>T p.P442L Chr.11: 556048-556048 on Build GRCh38
4194919 c.1349C>T p.A450V Chr.11: 556024-556024 on Build GRCh38
6391571 c.1415C>A p.P472Q Chr.11: 555893-555893 on Build GRCh38
1604702 c.1402C>T p.R468C Chr.11: 555906-555906 on Build GRCh38
4657456 c.1473T>G p.P491P Chr.11: 555835-555835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754041850 Chr.11: 556032-556032 on Build GRCh38
rs747947128 Chr.11: 555747-555747 on Build GRCh38
rs761789768 Chr.11: 556069-556069 on Build GRCh38
rs773022082 Chr.11: 556138-556138 on Build GRCh38
rs916113064 Chr.11: 555696-555696 on Build GRCh38
rs895953050 Chr.11: 556146-556146 on Build GRCh38
rs764113072 Chr.11: 556087-556087 on Build GRCh38
rs752850359 Chr.11: 556085-556085 on Build GRCh38
rs769534076 Chr.11: 556056-556056 on Build GRCh38
rs968564937 Chr.11: 556146-556147 on Build GRCh38
rs781570848 Chr.11: 555849-555849 on Build GRCh38
rs893224475 Chr.11: 555768-555768 on Build GRCh38
rs749355145 Chr.11: 555858-555858 on Build GRCh38
rs914366507 Chr.11: 555764-555764 on Build GRCh38
rs1028289057 Chr.11: 555715-555715 on Build GRCh38
rs369760457 Chr.11: 555946-555946 on Build GRCh38
rs776163484 Chr.11: 556002-556002 on Build GRCh38
rs559456928 Chr.11: 555734-555734 on Build GRCh38
rs1034135367 Chr.11: 556154-556154 on Build GRCh38
rs764268065 Chr.11: 556176-556176 on Build GRCh38
rs767128294 Chr.11: 555803-555803 on Build GRCh38
rs747952512 Chr.11: 555844-555844 on Build GRCh38
rs751686457 Chr.11: 556185-556185 on Build GRCh38
rs1031851837 Chr.11: 556171-556171 on Build GRCh38
rs752597715 Chr.11: 555962-555962 on Build GRCh38
rs779289251 Chr.11: 555939-555939 on Build GRCh38
rs891678309 Chr.11: 556116-556116 on Build GRCh38
rs898546678 Chr.11: 555841-555841 on Build GRCh38
rs144614541 Chr.11: 556063-556063 on Build GRCh38
rs1037168668 Chr.11: 556027-556027 on Build GRCh38
rs747641994 Chr.11: 556133-556133 on Build GRCh38
rs763317230 Chr.11: 555782-555782 on Build GRCh38
rs774440626 Chr.11: 556066-556066 on Build GRCh38
rs776980952 Chr.11: 555757-555757 on Build GRCh38
rs372220032 Chr.11: 555984-555984 on Build GRCh38
rs769217438 Chr.11: 556009-556009 on Build GRCh38
rs762942330 Chr.11: 555964-555964 on Build GRCh38
rs781579441 Chr.11: 556050-556050 on Build GRCh38
rs995500433 Chr.11: 555714-555714 on Build GRCh38
rs774615777 Chr.11: 555800-555800 on Build GRCh38
rs778243647 Chr.11: 555996-555996 on Build GRCh38
rs750285855 Chr.11: 555995-555999 on Build GRCh38
rs764748102 Chr.11: 555960-555960 on Build GRCh38
rs774991736 Chr.11: 556058-556058 on Build GRCh38
rs779757744 Chr.11: 556123-556123 on Build GRCh38
rs113093997 Chr.11: 556049-556049 on Build GRCh38
rs777661897 Chr.11: 555916-555916 on Build GRCh38
rs771108415 Chr.11: 555755-555755 on Build GRCh38
rs754499209 Chr.11: 555893-555893 on Build GRCh38
rs757820768 Chr.11: 556095-556095 on Build GRCh38
rs369204490 Chr.11: 556014-556014 on Build GRCh38
rs181920428 Chr.11: 555835-555835 on Build GRCh38
rs773110508 Chr.11: 556076-556076 on Build GRCh38
rs756442734 Chr.11: 555929-555929 on Build GRCh38
rs752884321 Chr.11: 556043-556043 on Build GRCh38
rs990276473 Chr.11: 556180-556180 on Build GRCh38
rs767623011 Chr.11: 555949-555950 on Build GRCh38
rs758209006 Chr.11: 555848-555848 on Build GRCh38
rs375199142 Chr.11: 555855-555855 on Build GRCh38
rs758486214 Chr.11: 556086-556086 on Build GRCh38
rs774047600 Chr.11: 556128-556128 on Build GRCh38
rs549422840 Chr.11: 556017-556017 on Build GRCh38
rs1002498488 Chr.11: 556167-556167 on Build GRCh38
rs544702519 Chr.11: 555779-555779 on Build GRCh38
rs765987111 Chr.11: 555890-555890 on Build GRCh38
rs1039069677 Chr.11: 555811-555811 on Build GRCh38
rs754914740 Chr.11: 555840-555840 on Build GRCh38
rs767282735 Chr.11: 556031-556031 on Build GRCh38
rs151247449 Chr.11: 555874-555874 on Build GRCh38
rs746186023 Chr.11: 556053-556053 on Build GRCh38
rs1021447149 Chr.11: 556137-556137 on Build GRCh38
rs765156653 Chr.11: 555898-555898 on Build GRCh38
rs775713053 Chr.11: 556150-556150 on Build GRCh38
rs764092481 Chr.11: 555967-555967 on Build GRCh38
rs762529512 Chr.11: 556012-556012 on Build GRCh38
rs992466909 Chr.11: 556160-556160 on Build GRCh38
rs914771081 Chr.11: 556181-556181 on Build GRCh38
rs934845247 Chr.11: 555738-555738 on Build GRCh38
rs758469682 Chr.11: 556044-556044 on Build GRCh38
rs776344348 Chr.11: 555952-555952 on Build GRCh38
rs769926978 Chr.11: 555765-555765 on Build GRCh38
rs528275466 Chr.11: 555740-555740 on Build GRCh38
rs767799115 Chr.11: 555877-555877 on Build GRCh38
rs898499342 Chr.11: 555702-555702 on Build GRCh38
rs755638244 Chr.11: 555831-555831 on Build GRCh38
rs763123281 Chr.11: 556169-556169 on Build GRCh38
rs201667162 Chr.11: 556093-556093 on Build GRCh38
rs370908143 Chr.11: 556107-556107 on Build GRCh38
rs1001272694 Chr.11: 556153-556153 on Build GRCh38
rs376755050 Chr.11: 555942-555942 on Build GRCh38
rs553041849 Chr.11: 556122-556122 on Build GRCh38
rs573958813 Chr.11: 556118-556118 on Build GRCh38
rs770629458 Chr.11: 555950-555950 on Build GRCh38
rs775105805 Chr.11: 556010-556010 on Build GRCh38
rs776153917 Chr.11: 555771-555771 on Build GRCh38
rs774905680 Chr.11: 555864-555864 on Build GRCh38
rs371311282 Chr.11: 555922-555922 on Build GRCh38
rs201290232 Chr.11: 555992-555992 on Build GRCh38
rs772876961 Chr.11: 555886-555886 on Build GRCh38
rs761703592 Chr.11: 556023-556023 on Build GRCh38
rs766026788 Chr.11: 556151-556164 on Build GRCh38
rs749981586 Chr.11: 555706-555706 on Build GRCh38
rs566272772 Chr.11: 556047-556047 on Build GRCh38
rs760523533 Chr.11: 556139-556139 on Build GRCh38
rs199751579 Chr.11: 556048-556048 on Build GRCh38
rs746028495 Chr.11: 555850-555850 on Build GRCh38
rs772621123 Chr.11: 555993-555993 on Build GRCh38
rs943332404 Chr.11: 555810-555810 on Build GRCh38
rs760285670 Chr.11: 555887-555887 on Build GRCh38
rs746012179 Chr.11: 555760-555760 on Build GRCh38
rs367784358 Chr.11: 555892-555892 on Build GRCh38
rs748889032 Chr.11: 556125-556125 on Build GRCh38
rs1056742363 Chr.11: 555809-555810 on Build GRCh38
rs866202550 Chr.11: 555708-555708 on Build GRCh38
rs936467 Chr.11: 556121-556121 on Build GRCh38
rs757102043 Chr.11: 555980-555980 on Build GRCh38
rs758140960 Chr.11: 555914-555914 on Build GRCh38
rs752614516 Chr.11: 555905-555905 on Build GRCh38
rs768773261 Chr.11: 555937-555937 on Build GRCh38
rs774612819 Chr.11: 555854-555855 on Build GRCh38
rs1036305120 Chr.11: 556141-556141 on Build GRCh38
rs755603542 Chr.11: 555718-555718 on Build GRCh38
rs775937986 Chr.11: 555799-555799 on Build GRCh38
rs530845240 Chr.11: 555724-555724 on Build GRCh38
rs755270456 Chr.11: 556033-556033 on Build GRCh38
rs775669576 Chr.11: 555856-555856 on Build GRCh38
rs770005967 Chr.11: 555798-555798 on Build GRCh38
rs749383860 Chr.11: 555936-555936 on Build GRCh38
rs778888773 Chr.11: 555842-555842 on Build GRCh38
rs764549075 Chr.11: 555789-555789 on Build GRCh38
rs979723878 Chr.11: 555793-555793 on Build GRCh38
rs780397767 Chr.11: 555931-555931 on Build GRCh38
rs372930527 Chr.11: 556000-556000 on Build GRCh38
rs898235211 Chr.11: 556092-556092 on Build GRCh38
rs748643994 Chr.11: 555940-555940 on Build GRCh38
rs753806780 Chr.11: 555838-555838 on Build GRCh38
rs766030791 Chr.11: 556140-556140 on Build GRCh38
rs1004067278 Chr.11: 555888-555888 on Build GRCh38
rs780485752 Chr.11: 555981-555981 on Build GRCh38
rs761857623 Chr.11: 555919-555922 on Build GRCh38
rs376417382 Chr.11: 556007-556007 on Build GRCh38
rs959971866 Chr.11: 556155-556155 on Build GRCh38
rs750067931 Chr.11: 555829-555829 on Build GRCh38
rs899746121 Chr.11: 555812-555812 on Build GRCh38
rs200729988 Chr.11: 555999-555999 on Build GRCh38
rs376070964 Chr.11: 556083-556083 on Build GRCh38
rs955929571 Chr.11: 555753-555753 on Build GRCh38
rs769490400 Chr.11: 556089-556089 on Build GRCh38
rs971066487 Chr.11: 556164-556164 on Build GRCh38
rs758974339 Chr.11: 555958-555958 on Build GRCh38
rs755023856 Chr.11: 555989-555989 on Build GRCh38
rs772460562 Chr.11: 555941-555941 on Build GRCh38
rs764296433 Chr.11: 555842-555842 on Build GRCh38
rs767328607 Chr.11: 556073-556073 on Build GRCh38
rs763018325 Chr.11: 555835-555838 on Build GRCh38
rs751448457 Chr.11: 555973-555973 on Build GRCh38

View Full Product Details