Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81061881-81062433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf46
Entrez Gene ID
123775
Gene Name
chromosome 16 open reading frame 46
Gene Aliases
-
UniGene
Hs.658684
Gene Chromosome Location
Chr.16: 81053497-81077267 on Build GRCh38
Transcripts
NM_001100873.1 NM_152337.2 XM_005255792.2 XM_011522848.2 XM_011522849.2 XM_017022916.1 XM_017022917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5690296 c.362G>T p.G121V Chr.16: 81061987-81061987 on Build GRCh38
5580395 c.431C>T p.S144F Chr.16: 81061918-81061918 on Build GRCh38
5971950 c.439A>G p.I147V Chr.16: 81061910-81061910 on Build GRCh38
4489210 c.344C>A p.T115N Chr.16: 81062005-81062005 on Build GRCh38
7121204 c.255G>T p.P85P Chr.16: 81062094-81062094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770319344 Chr.16: 81062117-81062117 on Build GRCh38
rs536027865 Chr.16: 81062052-81062052 on Build GRCh38
rs906361568 Chr.16: 81061986-81061986 on Build GRCh38
rs961045076 Chr.16: 81062332-81062332 on Build GRCh38
rs999341733 Chr.16: 81062313-81062313 on Build GRCh38
rs369539406 Chr.16: 81062177-81062177 on Build GRCh38
rs745791363 Chr.16: 81062131-81062131 on Build GRCh38
rs774775750 Chr.16: 81062076-81062076 on Build GRCh38
rs144441768 Chr.16: 81062367-81062367 on Build GRCh38
rs1002980905 Chr.16: 81062007-81062007 on Build GRCh38
rs550942710 Chr.16: 81062404-81062404 on Build GRCh38
rs375306034 Chr.16: 81062100-81062100 on Build GRCh38
rs368491503 Chr.16: 81062162-81062162 on Build GRCh38
rs899449115 Chr.16: 81062238-81062238 on Build GRCh38
rs771549687 Chr.16: 81062051-81062051 on Build GRCh38
rs981714390 Chr.16: 81062348-81062348 on Build GRCh38
rs573382071 Chr.16: 81062176-81062176 on Build GRCh38
rs927892859 Chr.16: 81062349-81062349 on Build GRCh38
rs763850964 Chr.16: 81062174-81062174 on Build GRCh38
rs75081512 Chr.16: 81062391-81062391 on Build GRCh38
rs879028808 Chr.16: 81062138-81062138 on Build GRCh38
rs760042318 Chr.16: 81062167-81062167 on Build GRCh38
rs773943085 Chr.16: 81061935-81061935 on Build GRCh38
rs769694619 Chr.16: 81062073-81062073 on Build GRCh38
rs747685632 Chr.16: 81061949-81061949 on Build GRCh38
rs145137165 Chr.16: 81062096-81062096 on Build GRCh38
rs376500530 Chr.16: 81061912-81061912 on Build GRCh38
rs535125163 Chr.16: 81061981-81061981 on Build GRCh38
rs753998253 Chr.16: 81061937-81061937 on Build GRCh38
rs756490402 Chr.16: 81061928-81061928 on Build GRCh38
rs746447976 Chr.16: 81062053-81062053 on Build GRCh38
rs759358077 Chr.16: 81061984-81061984 on Build GRCh38
rs997433409 Chr.16: 81062275-81062275 on Build GRCh38
rs370895120 Chr.16: 81062027-81062027 on Build GRCh38
rs750955250 Chr.16: 81062025-81062025 on Build GRCh38
rs908476644 Chr.16: 81061954-81061954 on Build GRCh38
rs531128657 Chr.16: 81062208-81062208 on Build GRCh38
rs547136317 Chr.16: 81062368-81062368 on Build GRCh38
rs548573737 Chr.16: 81062043-81062043 on Build GRCh38
rs746561469 Chr.16: 81062113-81062113 on Build GRCh38
rs200088357 Chr.16: 81061988-81061988 on Build GRCh38
rs200929292 Chr.16: 81062150-81062150 on Build GRCh38
rs757375444 Chr.16: 81061918-81061918 on Build GRCh38
rs987641321 Chr.16: 81062204-81062204 on Build GRCh38
rs753151457 Chr.16: 81062258-81062258 on Build GRCh38
rs145959785 Chr.16: 81062111-81062111 on Build GRCh38
rs781594740 Chr.16: 81061922-81061922 on Build GRCh38
rs751802794 Chr.16: 81061993-81061993 on Build GRCh38
rs867119077 Chr.16: 81062393-81062393 on Build GRCh38
rs540126191 Chr.16: 81062095-81062095 on Build GRCh38
rs762118775 Chr.16: 81062086-81062086 on Build GRCh38
rs139640227 Chr.16: 81061956-81061956 on Build GRCh38
rs894553525 Chr.16: 81062168-81062168 on Build GRCh38
rs372482557 Chr.16: 81062171-81062171 on Build GRCh38
rs940810823 Chr.16: 81062064-81062064 on Build GRCh38
rs770924767 Chr.16: 81062059-81062059 on Build GRCh38
rs550734784 Chr.16: 81062257-81062258 on Build GRCh38
rs778576934 Chr.16: 81062049-81062049 on Build GRCh38
rs377690413 Chr.16: 81061965-81061965 on Build GRCh38
rs965520073 Chr.16: 81062083-81062083 on Build GRCh38
rs903256454 Chr.16: 81062309-81062309 on Build GRCh38
rs757385777 Chr.16: 81062379-81062379 on Build GRCh38
rs767565356 Chr.16: 81062087-81062087 on Build GRCh38
rs780125351 Chr.16: 81061904-81061904 on Build GRCh38
rs142581816 Chr.16: 81062048-81062048 on Build GRCh38
rs138473797 Chr.16: 81062206-81062206 on Build GRCh38
rs758651996 Chr.16: 81062173-81062173 on Build GRCh38
rs539680019 Chr.16: 81062344-81062345 on Build GRCh38
rs747775575 Chr.16: 81062050-81062050 on Build GRCh38
rs34336262 Chr.16: 81062114-81062115 on Build GRCh38
rs766163636 Chr.16: 81061978-81061978 on Build GRCh38
rs758084167 Chr.16: 81062108-81062108 on Build GRCh38
rs939935320 Chr.16: 81061961-81061961 on Build GRCh38
rs185945609 Chr.16: 81062279-81062279 on Build GRCh38
rs552328735 Chr.16: 81061931-81061931 on Build GRCh38
rs150847199 Chr.16: 81061930-81061930 on Build GRCh38
rs879543236 Chr.16: 81062193-81062193 on Build GRCh38
rs535935042 Chr.16: 81062397-81062397 on Build GRCh38
rs764444536 Chr.16: 81061992-81061992 on Build GRCh38
rs756646141 Chr.16: 81062039-81062039 on Build GRCh38
rs781329941 Chr.16: 81062125-81062125 on Build GRCh38
rs975584668 Chr.16: 81062091-81062091 on Build GRCh38
rs114440744 Chr.16: 81062063-81062063 on Build GRCh38
rs758700250 Chr.16: 81061908-81061908 on Build GRCh38
rs564331174 Chr.16: 81062366-81062366 on Build GRCh38
rs953181551 Chr.16: 81062374-81062374 on Build GRCh38
rs137998021 Chr.16: 81062023-81062023 on Build GRCh38
rs930888341 Chr.16: 81062240-81062240 on Build GRCh38
rs533122983 Chr.16: 81062267-81062267 on Build GRCh38
rs201312033 Chr.16: 81062310-81062310 on Build GRCh38
rs773071105 Chr.16: 81061957-81061957 on Build GRCh38
rs983820384 Chr.16: 81062310-81062312 on Build GRCh38
rs894474749 Chr.16: 81062270-81062270 on Build GRCh38
rs201160160 Chr.16: 81062152-81062152 on Build GRCh38
rs754222687 Chr.16: 81062044-81062044 on Build GRCh38
rs930992894 Chr.16: 81061905-81061905 on Build GRCh38
rs149233140 Chr.16: 81062330-81062330 on Build GRCh38
rs574719697 Chr.16: 81061982-81061982 on Build GRCh38
rs368168951 Chr.16: 81062151-81062151 on Build GRCh38
rs17855893 Chr.16: 81062120-81062120 on Build GRCh38
rs149945062 Chr.16: 81062094-81062094 on Build GRCh38
rs746367231 Chr.16: 81061924-81061924 on Build GRCh38
rs1014225418 Chr.16: 81062272-81062272 on Build GRCh38
rs764356197 Chr.16: 81061910-81061910 on Build GRCh38
rs757465481 Chr.16: 81061997-81061997 on Build GRCh38
rs772357799 Chr.16: 81062147-81062147 on Build GRCh38
rs34238619 Chr.16: 81062196-81062197 on Build GRCh38
rs753085120 Chr.16: 81062172-81062172 on Build GRCh38
rs544037105 Chr.16: 81062198-81062198 on Build GRCh38
rs148661197 Chr.16: 81062099-81062099 on Build GRCh38
rs568594148 Chr.16: 81062394-81062394 on Build GRCh38
rs775426030 Chr.16: 81062137-81062137 on Build GRCh38
rs1036520487 Chr.16: 81062227-81062227 on Build GRCh38
rs769775408 Chr.16: 81062136-81062136 on Build GRCh38
rs752993396 Chr.16: 81062106-81062106 on Build GRCh38
rs989531596 Chr.16: 81062006-81062006 on Build GRCh38
rs917739173 Chr.16: 81061920-81061920 on Build GRCh38
rs952602423 Chr.16: 81062309-81062310 on Build GRCh38
rs1019898100 Chr.16: 81062246-81062246 on Build GRCh38
rs762594199 Chr.16: 81062145-81062145 on Build GRCh38

View Full Product Details