Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 413933-414483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf97
Entrez Gene ID
400566
Gene Name
chromosome 17 open reading frame 97
Gene Aliases
CK20, LIAT1
UniGene
Hs.499607
Gene Chromosome Location
Chr.17: 410327-414666 on Build GRCh38
Transcripts
NM_001013672.4 NR_136407.1

Gene Symbol
LOC105371430
Entrez Gene ID
105371430
Gene Name
uncharacterized LOC105371430
Gene Aliases
-
UniGene
Hs.616427
Gene Chromosome Location
Chr.17: 404535-414023 on Build GRCh38
Transcripts
NM_001013672.4 NR_136407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6231107 c.1114G>A p.E372K Chr.17: 413957-413957 on Build GRCh38
6614790 c.1225A>G p.T409A Chr.17: 414068-414068 on Build GRCh38
6614785 c.1128G>T p.E376D Chr.17: 413971-413971 on Build GRCh38
436220 c.1113C>T p.A371A Chr.17: 413956-413956 on Build GRCh38
6312839 c.1182G>T p.T394T Chr.17: 414025-414025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190578696 Chr.17: 414130-414130 on Build GRCh38
rs782698850 Chr.17: 414122-414122 on Build GRCh38
rs563855864 Chr.17: 414177-414177 on Build GRCh38
rs782649545 Chr.17: 414163-414163 on Build GRCh38
rs376542072 Chr.17: 413986-413986 on Build GRCh38
rs376498016 Chr.17: 414127-414127 on Build GRCh38
rs990777408 Chr.17: 414195-414195 on Build GRCh38
rs782015239 Chr.17: 414228-414228 on Build GRCh38
rs139203462 Chr.17: 414305-414305 on Build GRCh38
rs781935648 Chr.17: 414354-414354 on Build GRCh38
rs782250521 Chr.17: 413956-413956 on Build GRCh38
rs782780272 Chr.17: 414013-414013 on Build GRCh38
rs143312241 Chr.17: 413967-413967 on Build GRCh38
rs999991239 Chr.17: 414414-414414 on Build GRCh38
rs571644556 Chr.17: 414272-414272 on Build GRCh38
rs782347758 Chr.17: 414053-414053 on Build GRCh38
rs942213743 Chr.17: 414009-414009 on Build GRCh38
rs782222381 Chr.17: 414151-414151 on Build GRCh38
rs781895652 Chr.17: 414015-414015 on Build GRCh38
rs1010186048 Chr.17: 414106-414106 on Build GRCh38
rs782085753 Chr.17: 414421-414421 on Build GRCh38
rs573093685 Chr.17: 414028-414028 on Build GRCh38
rs181768176 Chr.17: 414167-414167 on Build GRCh38
rs782198369 Chr.17: 414051-414051 on Build GRCh38
rs554349869 Chr.17: 414446-414446 on Build GRCh38
rs921678899 Chr.17: 414255-414255 on Build GRCh38
rs782082319 Chr.17: 413988-413988 on Build GRCh38
rs185673051 Chr.17: 414083-414083 on Build GRCh38
rs949674747 Chr.17: 413962-413962 on Build GRCh38
rs782542903 Chr.17: 414132-414132 on Build GRCh38
rs557076695 Chr.17: 414424-414424 on Build GRCh38
rs782427003 Chr.17: 414018-414018 on Build GRCh38
rs374601645 Chr.17: 414025-414025 on Build GRCh38
rs782381526 Chr.17: 414206-414206 on Build GRCh38
rs367550079 Chr.17: 414136-414136 on Build GRCh38
rs782392734 Chr.17: 413957-413957 on Build GRCh38
rs904445781 Chr.17: 414388-414388 on Build GRCh38
rs1020518051 Chr.17: 414120-414120 on Build GRCh38
rs531176470 Chr.17: 414205-414205 on Build GRCh38
rs782561616 Chr.17: 414042-414042 on Build GRCh38
rs781853699 Chr.17: 413989-413989 on Build GRCh38
rs983700234 Chr.17: 414235-414235 on Build GRCh38
rs908061292 Chr.17: 414003-414003 on Build GRCh38
rs1032392584 Chr.17: 414158-414158 on Build GRCh38
rs549602714 Chr.17: 414229-414229 on Build GRCh38
rs1041605351 Chr.17: 414323-414323 on Build GRCh38
rs149967281 Chr.17: 414442-414442 on Build GRCh38
rs1034183616 Chr.17: 414425-414425 on Build GRCh38
rs973983917 Chr.17: 414243-414243 on Build GRCh38
rs1051415707 Chr.17: 414271-414271 on Build GRCh38
rs79767178 Chr.17: 414005-414005 on Build GRCh38
rs782154320 Chr.17: 414115-414115 on Build GRCh38
rs908135012 Chr.17: 414239-414239 on Build GRCh38
rs782314729 Chr.17: 413979-413979 on Build GRCh38
rs782500692 Chr.17: 414159-414159 on Build GRCh38
rs528797803 Chr.17: 414256-414256 on Build GRCh38
rs782313685 Chr.17: 414351-414351 on Build GRCh38
rs782016918 Chr.17: 414086-414086 on Build GRCh38
rs374213304 Chr.17: 413966-413966 on Build GRCh38
rs781971351 Chr.17: 414076-414076 on Build GRCh38
rs782611489 Chr.17: 414149-414149 on Build GRCh38
rs568704805 Chr.17: 414426-414426 on Build GRCh38
rs546651078 Chr.17: 414261-414261 on Build GRCh38
rs371022573 Chr.17: 414131-414131 on Build GRCh38
rs377181273 Chr.17: 414128-414128 on Build GRCh38
rs374369462 Chr.17: 414134-414134 on Build GRCh38
rs568625990 Chr.17: 414240-414240 on Build GRCh38

View Full Product Details