Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41443985-41444542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5SL
Entrez Gene ID
55101
Gene Name
ATP5S like
Gene Aliases
-
UniGene
Hs.351099
Gene Chromosome Location
Chr.19: 41431318-41447822 on Build GRCh38
Transcripts
XM_011527065.2 NM_001130514.1 XM_017026119.1

Gene Symbol
ERICH4
Entrez Gene ID
100170765
Gene Name
glutamate rich 4
Gene Aliases
C19orf69
UniGene
Hs.649452
Gene Chromosome Location
Chr.19: 41442426-41444765 on Build GRCh38
Transcripts
XM_011527065.2 NM_001130514.1 XM_017026119.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4389002 c.219C>T p.C73C Chr.19: 41444050-41444050 on Build GRCh38
7260535 c.269_270insGGA p.E95_S96insE Chr.19: 41444100-41444101 on Build GRCh38
5389855 c.295G>A p.E99K Chr.19: 41444126-41444126 on Build GRCh38
4487516 c.319C>T p.Q107* Chr.19: 41444150-41444150 on Build GRCh38
3227150 c.184C>T p.R62C Chr.19: 41444015-41444015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111840261 Chr.19: 41444078-41444078 on Build GRCh38
rs896182658 Chr.19: 41444500-41444500 on Build GRCh38
rs765074669 Chr.19: 41444294-41444294 on Build GRCh38
rs782797466 Chr.19: 41444027-41444027 on Build GRCh38
rs782441153 Chr.19: 41444036-41444036 on Build GRCh38
rs376618280 Chr.19: 41444185-41444185 on Build GRCh38
rs941271703 Chr.19: 41444019-41444019 on Build GRCh38
rs1001058310 Chr.19: 41444151-41444151 on Build GRCh38
rs956984191 Chr.19: 41444204-41444204 on Build GRCh38
rs782744564 Chr.19: 41444009-41444009 on Build GRCh38
rs369926537 Chr.19: 41444227-41444227 on Build GRCh38
rs549466784 Chr.19: 41444015-41444015 on Build GRCh38
rs560077124 Chr.19: 41444285-41444285 on Build GRCh38
rs139707196 Chr.19: 41444144-41444144 on Build GRCh38
rs782085682 Chr.19: 41444260-41444260 on Build GRCh38
rs782319177 Chr.19: 41444263-41444263 on Build GRCh38
rs3745285 Chr.19: 41444392-41444392 on Build GRCh38
rs150607510 Chr.19: 41444073-41444073 on Build GRCh38
rs116061667 Chr.19: 41444304-41444304 on Build GRCh38
rs923311408 Chr.19: 41444241-41444241 on Build GRCh38
rs896230142 Chr.19: 41444074-41444074 on Build GRCh38
rs782174108 Chr.19: 41444225-41444225 on Build GRCh38
rs976478703 Chr.19: 41444230-41444230 on Build GRCh38
rs556738553 Chr.19: 41444480-41444480 on Build GRCh38
rs1023795725 Chr.19: 41444119-41444119 on Build GRCh38
rs782540249 Chr.19: 41444173-41444173 on Build GRCh38
rs188198596 Chr.19: 41444032-41444032 on Build GRCh38
rs782111142 Chr.19: 41444016-41444016 on Build GRCh38
rs79685372 Chr.19: 41444515-41444515 on Build GRCh38
rs782657551 Chr.19: 41444505-41444505 on Build GRCh38
rs782715914 Chr.19: 41444048-41444048 on Build GRCh38
rs782497144 Chr.19: 41444439-41444439 on Build GRCh38
rs375945214 Chr.19: 41444079-41444079 on Build GRCh38
rs1038666379 Chr.19: 41444023-41444023 on Build GRCh38
rs941175448 Chr.19: 41444354-41444354 on Build GRCh38
rs77815988 Chr.19: 41444430-41444430 on Build GRCh38
rs17265865 Chr.19: 41444361-41444361 on Build GRCh38

View Full Product Details