Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119158568-119159120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QL2
Entrez Gene ID
165257
Gene Name
complement C1q like 2
Gene Aliases
C1QTNF10, CTRP10
UniGene
Hs.433493
Gene Chromosome Location
Chr.2: 119156243-119158895 on Build GRCh38
Transcripts
NM_182528.3

SNP Detail

SNP ID Chromosome Location
rs867133638 Chr.2: 119158856-119158856 on Build GRCh38
rs909214620 Chr.2: 119158935-119158935 on Build GRCh38
rs868130348 Chr.2: 119158678-119158678 on Build GRCh38
rs7600045 Chr.2: 119159040-119159040 on Build GRCh38
rs570031866 Chr.2: 119158793-119158793 on Build GRCh38
rs1023722822 Chr.2: 119158782-119158782 on Build GRCh38
rs956495772 Chr.2: 119158657-119158657 on Build GRCh38
rs866421668 Chr.2: 119158951-119158951 on Build GRCh38
rs997398244 Chr.2: 119159037-119159038 on Build GRCh38
rs948992580 Chr.2: 119158995-119158995 on Build GRCh38
rs1040547657 Chr.2: 119158727-119158727 on Build GRCh38
rs967471027 Chr.2: 119158925-119158925 on Build GRCh38
rs975762799 Chr.2: 119158997-119158997 on Build GRCh38
rs751737091 Chr.2: 119158993-119158993 on Build GRCh38
rs986015824 Chr.2: 119158979-119158979 on Build GRCh38
rs1026331493 Chr.2: 119158591-119158591 on Build GRCh38
rs892211970 Chr.2: 119159036-119159036 on Build GRCh38
rs986467204 Chr.2: 119159077-119159077 on Build GRCh38
rs977269948 Chr.2: 119158669-119158669 on Build GRCh38
rs1024353758 Chr.2: 119158660-119158660 on Build GRCh38
rs550486519 Chr.2: 119158602-119158602 on Build GRCh38
rs537079179 Chr.2: 119158902-119158902 on Build GRCh38
rs966116226 Chr.2: 119158996-119158996 on Build GRCh38
rs1043284126 Chr.2: 119158999-119158999 on Build GRCh38
rs999134241 Chr.2: 119158937-119158937 on Build GRCh38
rs1034346485 Chr.2: 119158628-119158628 on Build GRCh38
rs932030365 Chr.2: 119158703-119158703 on Build GRCh38
rs1005797311 Chr.2: 119158781-119158781 on Build GRCh38
rs1030732058 Chr.2: 119158824-119158824 on Build GRCh38
rs1047207090 Chr.2: 119158778-119158778 on Build GRCh38
rs925895980 Chr.2: 119158695-119158695 on Build GRCh38
rs935919174 Chr.2: 119158696-119158696 on Build GRCh38
rs376301684 Chr.2: 119158841-119158841 on Build GRCh38
rs910227284 Chr.2: 119158712-119158712 on Build GRCh38
rs919326732 Chr.2: 119158944-119158944 on Build GRCh38
rs547983632 Chr.2: 119158707-119158707 on Build GRCh38
rs192850219 Chr.2: 119158764-119158764 on Build GRCh38
rs147085666 Chr.2: 119158732-119158732 on Build GRCh38
rs765831711 Chr.2: 119159008-119159008 on Build GRCh38
rs958810699 Chr.2: 119158641-119158641 on Build GRCh38
rs967634976 Chr.2: 119158661-119158661 on Build GRCh38
rs372145189 Chr.2: 119158790-119158790 on Build GRCh38
rs923247833 Chr.2: 119158876-119158876 on Build GRCh38
rs910059235 Chr.2: 119159039-119159039 on Build GRCh38
rs756560589 Chr.2: 119158726-119158726 on Build GRCh38
rs529489078 Chr.2: 119158699-119158699 on Build GRCh38
rs866371334 Chr.2: 119159080-119159080 on Build GRCh38
rs929066270 Chr.2: 119159019-119159019 on Build GRCh38
rs1032800049 Chr.2: 119158949-119158949 on Build GRCh38
rs1030784424 Chr.2: 119159035-119159035 on Build GRCh38
rs553200094 Chr.2: 119158981-119158981 on Build GRCh38
rs1012310939 Chr.2: 119158623-119158623 on Build GRCh38
rs775731869 Chr.2: 119158679-119158679 on Build GRCh38
rs2121217 Chr.2: 119158737-119158737 on Build GRCh38
rs534777100 Chr.2: 119158611-119158611 on Build GRCh38
rs888738379 Chr.2: 119158780-119158780 on Build GRCh38
rs1047293313 Chr.2: 119159037-119159037 on Build GRCh38
rs953415183 Chr.2: 119158883-119158883 on Build GRCh38
rs993328927 Chr.2: 119158651-119158651 on Build GRCh38
rs967670008 Chr.2: 119158789-119158789 on Build GRCh38
rs942122740 Chr.2: 119158728-119158728 on Build GRCh38
rs1036326265 Chr.2: 119158588-119158588 on Build GRCh38
rs975900567 Chr.2: 119158853-119158853 on Build GRCh38

View Full Product Details