Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7089477-7090029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1R
Entrez Gene ID
715
Gene Name
complement C1r subcomponent
Gene Aliases
-
UniGene
Hs.524224
Gene Chromosome Location
Chr.12: 7080209-7092447 on Build GRCh38
Transcripts
NM_001733.4 XM_017019915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7358355 c.595G>A p.D199N Chr.12: 7089605-7089605 on Build GRCh38
3872462 c.577G>A p.G193S Chr.12: 7089623-7089623 on Build GRCh38
6880407 c.585G>A p.E195E Chr.12: 7089615-7089615 on Build GRCh38
3872463 c.511C>T p.P171S Chr.12: 7089689-7089689 on Build GRCh38
6452516 c.546C>T p.Y182Y Chr.12: 7089654-7089654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144141261 Chr.12: 7089671-7089671 on Build GRCh38
rs147288661 Chr.12: 7089732-7089732 on Build GRCh38
rs1046146760 Chr.12: 7089816-7089816 on Build GRCh38
rs775225161 Chr.12: 7089818-7089818 on Build GRCh38
rs774435623 Chr.12: 7089610-7089610 on Build GRCh38
rs756745476 Chr.12: 7089545-7089545 on Build GRCh38
rs984374350 Chr.12: 7089644-7089644 on Build GRCh38
rs934242210 Chr.12: 7089717-7089717 on Build GRCh38
rs972670373 Chr.12: 7089967-7089967 on Build GRCh38
rs769139608 Chr.12: 7089549-7089549 on Build GRCh38
rs778291125 Chr.12: 7089633-7089633 on Build GRCh38
rs767512802 Chr.12: 7089622-7089622 on Build GRCh38
rs891887068 Chr.12: 7089811-7089813 on Build GRCh38
rs752220136 Chr.12: 7089639-7089639 on Build GRCh38
rs192795036 Chr.12: 7089863-7089863 on Build GRCh38
rs916076749 Chr.12: 7089616-7089616 on Build GRCh38
rs1023163412 Chr.12: 7089881-7089881 on Build GRCh38
rs911499963 Chr.12: 7089964-7089964 on Build GRCh38
rs1010679695 Chr.12: 7089614-7089614 on Build GRCh38
rs985550787 Chr.12: 7089982-7089982 on Build GRCh38
rs970233366 Chr.12: 7089900-7089900 on Build GRCh38
rs773610671 Chr.12: 7089578-7089578 on Build GRCh38
rs779673453 Chr.12: 7089733-7089733 on Build GRCh38
rs763446444 Chr.12: 7089554-7089554 on Build GRCh38
rs753847112 Chr.12: 7089938-7089938 on Build GRCh38
rs772158595 Chr.12: 7089813-7089813 on Build GRCh38
rs750020707 Chr.12: 7089731-7089731 on Build GRCh38
rs746343842 Chr.12: 7089916-7089916 on Build GRCh38
rs922603955 Chr.12: 7089974-7089974 on Build GRCh38
rs771121230 Chr.12: 7089692-7089692 on Build GRCh38
rs368895945 Chr.12: 7089546-7089546 on Build GRCh38
rs781112846 Chr.12: 7089701-7089701 on Build GRCh38
rs768646357 Chr.12: 7089834-7089834 on Build GRCh38
rs775687119 Chr.12: 7089972-7089972 on Build GRCh38
rs764995825 Chr.12: 7089723-7089723 on Build GRCh38
rs1017111744 Chr.12: 7089933-7089933 on Build GRCh38
rs768951004 Chr.12: 7089536-7089536 on Build GRCh38
rs572440545 Chr.12: 7089508-7089508 on Build GRCh38
rs779970479 Chr.12: 7089769-7089769 on Build GRCh38
rs757414596 Chr.12: 7089880-7089880 on Build GRCh38
rs767184697 Chr.12: 7089740-7089740 on Build GRCh38
rs111937645 Chr.12: 7089514-7089514 on Build GRCh38
rs745450078 Chr.12: 7089636-7089636 on Build GRCh38
rs377259241 Chr.12: 7089729-7089729 on Build GRCh38
rs200018198 Chr.12: 7089628-7089628 on Build GRCh38
rs774434691 Chr.12: 7089694-7089694 on Build GRCh38
rs766984810 Chr.12: 7089573-7089574 on Build GRCh38
rs990294606 Chr.12: 7089991-7089991 on Build GRCh38
rs746034917 Chr.12: 7089810-7089810 on Build GRCh38
rs758521733 Chr.12: 7089851-7089851 on Build GRCh38
rs563648384 Chr.12: 7089831-7089831 on Build GRCh38
rs771189824 Chr.12: 7089778-7089778 on Build GRCh38
rs747484081 Chr.12: 7089548-7089548 on Build GRCh38
rs762993517 Chr.12: 7089513-7089513 on Build GRCh38
rs985676031 Chr.12: 7089949-7089949 on Build GRCh38
rs374722868 Chr.12: 7089763-7089763 on Build GRCh38
rs757966852 Chr.12: 7089641-7089641 on Build GRCh38
rs34054348 Chr.12: 7089889-7089890 on Build GRCh38
rs759722177 Chr.12: 7089699-7089699 on Build GRCh38
rs934019766 Chr.12: 7089979-7089979 on Build GRCh38
rs1801046 Chr.12: 7089703-7089703 on Build GRCh38
rs1013995041 Chr.12: 7089877-7089877 on Build GRCh38
rs773663621 Chr.12: 7089655-7089655 on Build GRCh38
rs748580260 Chr.12: 7089571-7089571 on Build GRCh38
rs752287162 Chr.12: 7089533-7089533 on Build GRCh38
rs777447841 Chr.12: 7089535-7089535 on Build GRCh38
rs73046147 Chr.12: 7089712-7089712 on Build GRCh38
rs1033519837 Chr.12: 7089652-7089652 on Build GRCh38
rs760553459 Chr.12: 7089708-7089708 on Build GRCh38
rs748635646 Chr.12: 7089654-7089654 on Build GRCh38
rs751089506 Chr.12: 7089625-7089625 on Build GRCh38
rs756873175 Chr.12: 7089630-7089630 on Build GRCh38
rs759400396 Chr.12: 7089621-7089621 on Build GRCh38
rs777271496 Chr.12: 7089562-7089562 on Build GRCh38
rs765619313 Chr.12: 7089504-7089504 on Build GRCh38
rs1126605 Chr.12: 7089608-7089608 on Build GRCh38
rs267603660 Chr.12: 7089605-7089605 on Build GRCh38
rs763194636 Chr.12: 7089584-7089584 on Build GRCh38
rs998548080 Chr.12: 7089570-7089570 on Build GRCh38
rs756632580 Chr.12: 7089755-7089755 on Build GRCh38
rs375378142 Chr.12: 7089517-7089517 on Build GRCh38
rs904461071 Chr.12: 7089828-7089828 on Build GRCh38
rs757198922 Chr.12: 7089964-7089964 on Build GRCh38
rs767238087 Chr.12: 7089575-7089575 on Build GRCh38
rs772646819 Chr.12: 7089898-7089898 on Build GRCh38
rs373462345 Chr.12: 7089713-7089713 on Build GRCh38
rs770372363 Chr.12: 7089653-7089653 on Build GRCh38
rs751484583 Chr.12: 7089516-7089516 on Build GRCh38
rs752425153 Chr.12: 7089522-7089522 on Build GRCh38
rs4519167 Chr.12: 7089601-7089601 on Build GRCh38
rs1000247449 Chr.12: 7089582-7089582 on Build GRCh38
rs750747014 Chr.12: 7089510-7089510 on Build GRCh38
rs779247472 Chr.12: 7089805-7089805 on Build GRCh38
rs747039205 Chr.12: 7089869-7089869 on Build GRCh38
rs761308082 Chr.12: 7089920-7089920 on Build GRCh38
rs1029753488 Chr.12: 7089937-7089937 on Build GRCh38
rs73046140 Chr.12: 7089518-7089518 on Build GRCh38

View Full Product Details