Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26951921-26952467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDF1
Entrez Gene ID
126695
Gene Name
keratinocyte differentiation factor 1
Gene Aliases
C1orf172
UniGene
Hs.188881
Gene Chromosome Location
Chr.1: 26949556-26960484 on Build GRCh38
Transcripts
NM_152365.2 XM_005245735.2 XM_011540622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
126580 c.58G>T p.E20* Chr.1: 26952323-26952323 on Build GRCh38
6615135 c.119G>A p.R40H Chr.1: 26952262-26952262 on Build GRCh38
3487624 c.411C>T p.S137S Chr.1: 26951970-26951970 on Build GRCh38
4856095 c.82G>T p.E28* Chr.1: 26952299-26952299 on Build GRCh38
3419086 c.126C>A p.T42T Chr.1: 26952255-26952255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768659946 Chr.1: 26952137-26952137 on Build GRCh38
rs541799014 Chr.1: 26952100-26952100 on Build GRCh38
rs375412273 Chr.1: 26952410-26952410 on Build GRCh38
rs765887283 Chr.1: 26952095-26952095 on Build GRCh38
rs768274676 Chr.1: 26952352-26952352 on Build GRCh38
rs745948877 Chr.1: 26952133-26952133 on Build GRCh38
rs768749901 Chr.1: 26952081-26952081 on Build GRCh38
rs566133911 Chr.1: 26951976-26951976 on Build GRCh38
rs767081810 Chr.1: 26952179-26952179 on Build GRCh38
rs550041376 Chr.1: 26951947-26951947 on Build GRCh38
rs759227084 Chr.1: 26952263-26952263 on Build GRCh38
rs148853297 Chr.1: 26952098-26952098 on Build GRCh38
rs759059288 Chr.1: 26952176-26952176 on Build GRCh38
rs550689765 Chr.1: 26952042-26952042 on Build GRCh38
rs565917289 Chr.1: 26952359-26952359 on Build GRCh38
rs770558931 Chr.1: 26952260-26952260 on Build GRCh38
rs1001504049 Chr.1: 26952142-26952142 on Build GRCh38
rs371561424 Chr.1: 26952060-26952060 on Build GRCh38
rs145806681 Chr.1: 26952328-26952328 on Build GRCh38
rs770975636 Chr.1: 26951994-26951994 on Build GRCh38
rs375203089 Chr.1: 26952344-26952344 on Build GRCh38
rs3010109 Chr.1: 26952062-26952062 on Build GRCh38
rs368517120 Chr.1: 26952163-26952163 on Build GRCh38
rs199965062 Chr.1: 26952254-26952254 on Build GRCh38
rs144535348 Chr.1: 26952188-26952188 on Build GRCh38
rs753510550 Chr.1: 26951958-26951958 on Build GRCh38
rs749541622 Chr.1: 26952070-26952070 on Build GRCh38
rs765760178 Chr.1: 26952306-26952306 on Build GRCh38
rs756057910 Chr.1: 26952164-26952165 on Build GRCh38
rs371974787 Chr.1: 26951995-26951995 on Build GRCh38
rs548929772 Chr.1: 26952358-26952358 on Build GRCh38
rs17360994 Chr.1: 26952082-26952082 on Build GRCh38
rs762075123 Chr.1: 26952291-26952291 on Build GRCh38
rs151306106 Chr.1: 26952121-26952121 on Build GRCh38
rs377413091 Chr.1: 26951962-26951962 on Build GRCh38
rs373271588 Chr.1: 26952342-26952342 on Build GRCh38
rs150246438 Chr.1: 26951948-26951948 on Build GRCh38
rs372393245 Chr.1: 26952201-26952201 on Build GRCh38
rs778640487 Chr.1: 26952431-26952431 on Build GRCh38
rs770601582 Chr.1: 26952332-26952332 on Build GRCh38
rs368075916 Chr.1: 26951963-26951963 on Build GRCh38
rs756324322 Chr.1: 26952416-26952416 on Build GRCh38
rs1033950363 Chr.1: 26952146-26952146 on Build GRCh38
rs779015336 Chr.1: 26952128-26952128 on Build GRCh38
rs369366004 Chr.1: 26951957-26951957 on Build GRCh38
rs144301593 Chr.1: 26952086-26952086 on Build GRCh38
rs775005166 Chr.1: 26952269-26952269 on Build GRCh38
rs776692680 Chr.1: 26952140-26952140 on Build GRCh38
rs749842745 Chr.1: 26952037-26952037 on Build GRCh38
rs750638599 Chr.1: 26952309-26952309 on Build GRCh38
rs748346371 Chr.1: 26951985-26951985 on Build GRCh38
rs745671012 Chr.1: 26952432-26952432 on Build GRCh38
rs752071068 Chr.1: 26952059-26952059 on Build GRCh38
rs758545597 Chr.1: 26952231-26952231 on Build GRCh38
rs145915640 Chr.1: 26952122-26952122 on Build GRCh38
rs139183337 Chr.1: 26952079-26952079 on Build GRCh38
rs139077715 Chr.1: 26952327-26952327 on Build GRCh38
rs200247502 Chr.1: 26952373-26952373 on Build GRCh38
rs534821230 Chr.1: 26951988-26951988 on Build GRCh38
rs1034227152 Chr.1: 26951992-26951992 on Build GRCh38
rs768999890 Chr.1: 26951980-26951980 on Build GRCh38
rs781573827 Chr.1: 26951981-26951981 on Build GRCh38
rs377299520 Chr.1: 26952126-26952126 on Build GRCh38
rs758768470 Chr.1: 26951971-26951971 on Build GRCh38
rs758740877 Chr.1: 26952313-26952313 on Build GRCh38
rs771012694 Chr.1: 26952077-26952077 on Build GRCh38
rs903857906 Chr.1: 26952141-26952141 on Build GRCh38
rs752617278 Chr.1: 26951972-26951973 on Build GRCh38
rs369041942 Chr.1: 26952421-26952421 on Build GRCh38
rs762549436 Chr.1: 26952094-26952094 on Build GRCh38
rs780162178 Chr.1: 26952438-26952438 on Build GRCh38
rs756011181 Chr.1: 26952320-26952320 on Build GRCh38
rs761208100 Chr.1: 26952275-26952275 on Build GRCh38
rs919668124 Chr.1: 26952199-26952199 on Build GRCh38
rs757023061 Chr.1: 26951960-26951960 on Build GRCh38
rs879093596 Chr.1: 26952120-26952120 on Build GRCh38
rs773781837 Chr.1: 26952167-26952167 on Build GRCh38
rs1049224185 Chr.1: 26952227-26952227 on Build GRCh38
rs982502162 Chr.1: 26952187-26952187 on Build GRCh38
rs932471700 Chr.1: 26952292-26952292 on Build GRCh38
rs369334382 Chr.1: 26952281-26952281 on Build GRCh38
rs202145629 Chr.1: 26952198-26952198 on Build GRCh38
rs780230453 Chr.1: 26952314-26952314 on Build GRCh38
rs746153605 Chr.1: 26952223-26952223 on Build GRCh38
rs773018618 Chr.1: 26952258-26952258 on Build GRCh38
rs779019147 Chr.1: 26952222-26952222 on Build GRCh38
rs766711238 Chr.1: 26952408-26952408 on Build GRCh38
rs763224813 Chr.1: 26952406-26952406 on Build GRCh38
rs750837066 Chr.1: 26952391-26952391 on Build GRCh38
rs143721921 Chr.1: 26952259-26952259 on Build GRCh38
rs919593203 Chr.1: 26952390-26952390 on Build GRCh38
rs753001226 Chr.1: 26952103-26952103 on Build GRCh38
rs753712013 Chr.1: 26952019-26952019 on Build GRCh38
rs773978155 Chr.1: 26952262-26952262 on Build GRCh38
rs763108668 Chr.1: 26951991-26951991 on Build GRCh38
rs754178127 Chr.1: 26952282-26952282 on Build GRCh38
rs761519934 Chr.1: 26952023-26952023 on Build GRCh38
rs144505487 Chr.1: 26952337-26952337 on Build GRCh38
rs757208209 Chr.1: 26952428-26952428 on Build GRCh38
rs769637271 Chr.1: 26952255-26952255 on Build GRCh38
rs535449942 Chr.1: 26951952-26951952 on Build GRCh38
rs747525165 Chr.1: 26951978-26951978 on Build GRCh38
rs982450139 Chr.1: 26952076-26952076 on Build GRCh38
rs769596979 Chr.1: 26952153-26952153 on Build GRCh38
rs772911827 Chr.1: 26952093-26952093 on Build GRCh38
rs769935791 Chr.1: 26952067-26952067 on Build GRCh38
rs373738003 Chr.1: 26952012-26952012 on Build GRCh38
rs17162368 Chr.1: 26952180-26952180 on Build GRCh38
rs561551802 Chr.1: 26952108-26952108 on Build GRCh38
rs772925863 Chr.1: 26952160-26952160 on Build GRCh38
rs770025779 Chr.1: 26951986-26951986 on Build GRCh38
rs79374331 Chr.1: 26952319-26952319 on Build GRCh38
rs1009596067 Chr.1: 26951954-26951954 on Build GRCh38
rs755438296 Chr.1: 26952099-26952099 on Build GRCh38
rs1011626133 Chr.1: 26952311-26952311 on Build GRCh38
rs939725374 Chr.1: 26951974-26951974 on Build GRCh38
rs765977534 Chr.1: 26952165-26952165 on Build GRCh38
rs748161190 Chr.1: 26952145-26952145 on Build GRCh38
rs199725807 Chr.1: 26951996-26951996 on Build GRCh38
rs780318215 Chr.1: 26952242-26952242 on Build GRCh38
rs367617835 Chr.1: 26952050-26952050 on Build GRCh38
rs141234946 Chr.1: 26952068-26952068 on Build GRCh38
rs765268716 Chr.1: 26951944-26951944 on Build GRCh38
rs761717950 Chr.1: 26952083-26952083 on Build GRCh38
rs760507136 Chr.1: 26951956-26951956 on Build GRCh38
rs779492462 Chr.1: 26952058-26952058 on Build GRCh38
rs757910529 Chr.1: 26952057-26952057 on Build GRCh38
rs529561188 Chr.1: 26952338-26952338 on Build GRCh38
rs138248574 Chr.1: 26952061-26952061 on Build GRCh38
rs760644538 Chr.1: 26952080-26952080 on Build GRCh38
rs140672463 Chr.1: 26952181-26952181 on Build GRCh38
rs3010110 Chr.1: 26952030-26952030 on Build GRCh38

View Full Product Details