Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1599884-1600408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC10
Entrez Gene ID
643988
Gene Name
fibronectin type III domain containing 10
Gene Aliases
C1orf233
UniGene
Hs.528816
Gene Chromosome Location
Chr.1: 1598008-1600096 on Build GRCh38
Transcripts
NM_001242659.1

SNP Detail

SNP ID Chromosome Location
rs1024081296 Chr.1: 1600382-1600382 on Build GRCh38
rs1007059835 Chr.1: 1600152-1600152 on Build GRCh38
rs575088735 Chr.1: 1600253-1600253 on Build GRCh38
rs537102883 Chr.1: 1600259-1600259 on Build GRCh38
rs374342563 Chr.1: 1600312-1600312 on Build GRCh38
rs1033126554 Chr.1: 1600378-1600378 on Build GRCh38
rs994304789 Chr.1: 1599963-1599963 on Build GRCh38
rs868051545 Chr.1: 1600368-1600368 on Build GRCh38
rs949390948 Chr.1: 1599943-1599943 on Build GRCh38
rs918557058 Chr.1: 1600262-1600262 on Build GRCh38
rs1055729485 Chr.1: 1599920-1599920 on Build GRCh38
rs888652457 Chr.1: 1599985-1599986 on Build GRCh38
rs1027716011 Chr.1: 1600103-1600117 on Build GRCh38
rs888586242 Chr.1: 1600148-1600148 on Build GRCh38
rs967099336 Chr.1: 1600188-1600188 on Build GRCh38
rs978558259 Chr.1: 1600090-1600090 on Build GRCh38
rs72897847 Chr.1: 1600379-1600379 on Build GRCh38
rs917173191 Chr.1: 1600111-1600111 on Build GRCh38
rs925764763 Chr.1: 1600095-1600095 on Build GRCh38
rs937236806 Chr.1: 1599920-1599920 on Build GRCh38
rs1032766173 Chr.1: 1600200-1600201 on Build GRCh38
rs200752460 Chr.1: 1600081-1600082 on Build GRCh38
rs1007523099 Chr.1: 1599987-1599987 on Build GRCh38
rs930009221 Chr.1: 1600283-1600283 on Build GRCh38
rs899409327 Chr.1: 1600252-1600252 on Build GRCh38
rs937208445 Chr.1: 1600096-1600096 on Build GRCh38
rs967528756 Chr.1: 1600069-1600069 on Build GRCh38
rs1039891849 Chr.1: 1600319-1600319 on Build GRCh38
rs1048916978 Chr.1: 1600303-1600303 on Build GRCh38
rs1018554379 Chr.1: 1600016-1600016 on Build GRCh38
rs866586459 Chr.1: 1600134-1600134 on Build GRCh38
rs940836738 Chr.1: 1600207-1600207 on Build GRCh38
rs1019994652 Chr.1: 1600044-1600044 on Build GRCh38
rs991275089 Chr.1: 1600208-1600208 on Build GRCh38
rs902809360 Chr.1: 1600345-1600345 on Build GRCh38
rs112683308 Chr.1: 1600205-1600205 on Build GRCh38
rs573171520 Chr.1: 1600381-1600381 on Build GRCh38
rs917120901 Chr.1: 1600215-1600215 on Build GRCh38
rs895128330 Chr.1: 1599938-1599938 on Build GRCh38
rs942794873 Chr.1: 1600314-1600314 on Build GRCh38
rs999873251 Chr.1: 1600347-1600347 on Build GRCh38
rs998425371 Chr.1: 1600340-1600340 on Build GRCh38
rs1012886515 Chr.1: 1600185-1600186 on Build GRCh38
rs971985762 Chr.1: 1600255-1600255 on Build GRCh38
rs907963609 Chr.1: 1600193-1600193 on Build GRCh38
rs1027551207 Chr.1: 1599974-1599974 on Build GRCh38
rs901415330 Chr.1: 1600156-1600156 on Build GRCh38
rs556702454 Chr.1: 1600308-1600308 on Build GRCh38
rs1012644955 Chr.1: 1600380-1600380 on Build GRCh38
rs950005621 Chr.1: 1600130-1600130 on Build GRCh38
rs897239408 Chr.1: 1599957-1599957 on Build GRCh38
rs998476321 Chr.1: 1600185-1600185 on Build GRCh38
rs116788278 Chr.1: 1600320-1600320 on Build GRCh38
rs868850591 Chr.1: 1600189-1600189 on Build GRCh38
rs965002261 Chr.1: 1600027-1600027 on Build GRCh38
rs1050514392 Chr.1: 1600325-1600325 on Build GRCh38
rs1037855210 Chr.1: 1600243-1600243 on Build GRCh38
rs897179622 Chr.1: 1600131-1600131 on Build GRCh38
rs552519502 Chr.1: 1599986-1599987 on Build GRCh38
rs1018502059 Chr.1: 1600155-1600155 on Build GRCh38
rs897943550 Chr.1: 1599953-1599965 on Build GRCh38
rs976421385 Chr.1: 1600037-1600037 on Build GRCh38

View Full Product Details