Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154212440-154213031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf43
Entrez Gene ID
25912
Gene Name
chromosome 1 open reading frame 43
Gene Aliases
HSPC012, NICE-3, NS5ATP4, S863-3
UniGene
Hs.287471
Gene Chromosome Location
Chr.1: 154206701-154220797 on Build GRCh38
Transcripts
NM_001098616.2 NM_001297717.1 NM_001297718.1 NM_001297720.1 NM_001297721.1 NM_001297723.1 NM_015449.3 NM_138740.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817904 c.419G>T p.R140L Chr.1: 154212546-154212546 on Build GRCh38
1472686 c.370C>T p.P124S Chr.1: 154212595-154212595 on Build GRCh38
1334392 c.330G>A p.T110T Chr.1: 154212533-154212533 on Build GRCh38
5775981 c.391A>G p.N131D Chr.1: 154212574-154212574 on Build GRCh38
5817905 c.317G>T p.R106L Chr.1: 154212546-154212546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146574729 Chr.1: 154212561-154212561 on Build GRCh38
rs946812541 Chr.1: 154212600-154212600 on Build GRCh38
rs749597940 Chr.1: 154212581-154212581 on Build GRCh38
rs1039758683 Chr.1: 154212834-154212834 on Build GRCh38
rs200726498 Chr.1: 154212836-154212836 on Build GRCh38
rs941147693 Chr.1: 154212710-154212710 on Build GRCh38
rs768270379 Chr.1: 154212498-154212498 on Build GRCh38
rs771075228 Chr.1: 154212516-154212516 on Build GRCh38
rs886755251 Chr.1: 154212911-154212911 on Build GRCh38
rs766248276 Chr.1: 154212637-154212637 on Build GRCh38
rs144345546 Chr.1: 154212541-154212541 on Build GRCh38
rs143492793 Chr.1: 154212647-154212648 on Build GRCh38
rs918441325 Chr.1: 154212754-154212754 on Build GRCh38
rs11265299 Chr.1: 154213000-154213000 on Build GRCh38
rs759676897 Chr.1: 154212526-154212526 on Build GRCh38
rs746459466 Chr.1: 154212497-154212497 on Build GRCh38
rs189173750 Chr.1: 154212703-154212703 on Build GRCh38
rs757114970 Chr.1: 154212472-154212472 on Build GRCh38
rs780448587 Chr.1: 154212680-154212680 on Build GRCh38
rs765023656 Chr.1: 154212617-154212617 on Build GRCh38
rs764075230 Chr.1: 154212468-154212468 on Build GRCh38
rs1000972075 Chr.1: 154212874-154212874 on Build GRCh38
rs750358991 Chr.1: 154212627-154212627 on Build GRCh38
rs751309561 Chr.1: 154212641-154212641 on Build GRCh38
rs1013802540 Chr.1: 154212938-154212938 on Build GRCh38
rs779908623 Chr.1: 154212554-154212554 on Build GRCh38
rs891099582 Chr.1: 154212824-154212824 on Build GRCh38
rs775388750 Chr.1: 154212603-154212603 on Build GRCh38
rs772578611 Chr.1: 154212964-154212964 on Build GRCh38
rs144385115 Chr.1: 154212517-154212517 on Build GRCh38
rs916892647 Chr.1: 154212693-154212693 on Build GRCh38
rs1047461585 Chr.1: 154212885-154212885 on Build GRCh38
rs770248018 Chr.1: 154212503-154212503 on Build GRCh38
rs529144689 Chr.1: 154212950-154212950 on Build GRCh38
rs763268372 Chr.1: 154212506-154212506 on Build GRCh38
rs779712469 Chr.1: 154212492-154212492 on Build GRCh38
rs759731768 Chr.1: 154212601-154212601 on Build GRCh38
rs565667137 Chr.1: 154212836-154212836 on Build GRCh38
rs537167939 Chr.1: 154212592-154212592 on Build GRCh38
rs979419058 Chr.1: 154212982-154212982 on Build GRCh38
rs564126082 Chr.1: 154212761-154212761 on Build GRCh38
rs747351828 Chr.1: 154212670-154212670 on Build GRCh38
rs917116479 Chr.1: 154212880-154212880 on Build GRCh38
rs763460932 Chr.1: 154212534-154212534 on Build GRCh38
rs531304728 Chr.1: 154212781-154212781 on Build GRCh38
rs559840807 Chr.1: 154212699-154212699 on Build GRCh38
rs772160636 Chr.1: 154212602-154212602 on Build GRCh38
rs756051751 Chr.1: 154212467-154212467 on Build GRCh38
rs886807848 Chr.1: 154212681-154212681 on Build GRCh38
rs761607078 Chr.1: 154212536-154212536 on Build GRCh38
rs780549431 Chr.1: 154212567-154212567 on Build GRCh38
rs927094724 Chr.1: 154212509-154212509 on Build GRCh38
rs769179333 Chr.1: 154212669-154212670 on Build GRCh38
rs111727677 Chr.1: 154212482-154212482 on Build GRCh38
rs916922935 Chr.1: 154212488-154212488 on Build GRCh38
rs897062308 Chr.1: 154212533-154212533 on Build GRCh38
rs773400484 Chr.1: 154212504-154212504 on Build GRCh38
rs781017184 Chr.1: 154212746-154212746 on Build GRCh38
rs16835874 Chr.1: 154212646-154212646 on Build GRCh38
rs778550771 Chr.1: 154212477-154212477 on Build GRCh38
rs555392838 Chr.1: 154212636-154212636 on Build GRCh38
rs770161723 Chr.1: 154212568-154212568 on Build GRCh38
rs968009937 Chr.1: 154212978-154212978 on Build GRCh38
rs182361299 Chr.1: 154212965-154212965 on Build GRCh38
rs956958075 Chr.1: 154212667-154212667 on Build GRCh38
rs988177331 Chr.1: 154212546-154212546 on Build GRCh38
rs200843446 Chr.1: 154212649-154212649 on Build GRCh38
rs553346355 Chr.1: 154212719-154212720 on Build GRCh38
rs756366734 Chr.1: 154212662-154212662 on Build GRCh38
rs149819448 Chr.1: 154212591-154212591 on Build GRCh38
rs979782523 Chr.1: 154212702-154212702 on Build GRCh38
rs891325080 Chr.1: 154212955-154212955 on Build GRCh38
rs748575878 Chr.1: 154212502-154212502 on Build GRCh38
rs541603280 Chr.1: 154212683-154212683 on Build GRCh38
rs376339258 Chr.1: 154212512-154212512 on Build GRCh38
rs926991101 Chr.1: 154213007-154213007 on Build GRCh38
rs778205904 Chr.1: 154212569-154212569 on Build GRCh38
rs370615805 Chr.1: 154212675-154212675 on Build GRCh38
rs543546165 Chr.1: 154212918-154212918 on Build GRCh38
rs758081183 Chr.1: 154212491-154212491 on Build GRCh38
rs761811000 Chr.1: 154212605-154212605 on Build GRCh38
rs112267415 Chr.1: 154212759-154212759 on Build GRCh38
rs750114569 Chr.1: 154212551-154212551 on Build GRCh38
rs949989990 Chr.1: 154212883-154212883 on Build GRCh38
rs575991961 Chr.1: 154212565-154212565 on Build GRCh38
rs779211626 Chr.1: 154212924-154212924 on Build GRCh38
rs1033684523 Chr.1: 154212999-154212999 on Build GRCh38
rs755339704 Chr.1: 154212884-154212884 on Build GRCh38
rs576205561 Chr.1: 154213005-154213005 on Build GRCh38
rs776392549 Chr.1: 154212483-154212483 on Build GRCh38
rs764986749 Chr.1: 154212501-154212501 on Build GRCh38
rs565865829 Chr.1: 154212977-154212977 on Build GRCh38
rs561729230 Chr.1: 154212932-154212932 on Build GRCh38
rs1006900523 Chr.1: 154212831-154212831 on Build GRCh38
rs114974392 Chr.1: 154212553-154212553 on Build GRCh38

View Full Product Details