Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2192617-2193186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAP20
Entrez Gene ID
199990
Gene Name
Fanconi anemia core complex associated protein 20
Gene Aliases
C1orf86, FP7162
UniGene
Hs.107101
Gene Chromosome Location
Chr.1: 2184460-2212720 on Build GRCh38
Transcripts
NM_001146310.1 NM_001256945.1 NM_001256946.1 NM_001256947.1 NM_001282670.1 NM_001282671.1 NM_001282672.1 NM_001282673.1 NM_182533.2 NR_046427.1 XM_006710419.3 XM_006710421.3 XM_011540914.2 XM_011540921.2 XM_011540922.1 XM_017000553.1 XM_017000554.1 XM_017000557.1

SNP Detail

SNP ID Chromosome Location
rs922285877 Chr.1: 2192835-2192835 on Build GRCh38
rs955250989 Chr.1: 2193098-2193098 on Build GRCh38
rs952206423 Chr.1: 2192758-2192758 on Build GRCh38
rs951444359 Chr.1: 2192732-2192732 on Build GRCh38
rs144522721 Chr.1: 2192953-2192953 on Build GRCh38
rs894625424 Chr.1: 2192920-2192920 on Build GRCh38
rs77158046 Chr.1: 2192905-2192905 on Build GRCh38
rs949024772 Chr.1: 2192925-2192925 on Build GRCh38
rs1045949826 Chr.1: 2192926-2192926 on Build GRCh38
rs935707839 Chr.1: 2192889-2192889 on Build GRCh38
rs902946331 Chr.1: 2192890-2192890 on Build GRCh38
rs1029598855 Chr.1: 2193083-2193083 on Build GRCh38
rs1016808675 Chr.1: 2193021-2193021 on Build GRCh38
rs142041547 Chr.1: 2192906-2192907 on Build GRCh38
rs1017145432 Chr.1: 2192763-2192763 on Build GRCh38
rs918674188 Chr.1: 2192721-2192721 on Build GRCh38
rs990301600 Chr.1: 2193129-2193129 on Build GRCh38
rs774561727 Chr.1: 2192762-2192762 on Build GRCh38
rs774911040 Chr.1: 2192944-2192944 on Build GRCh38
rs935777325 Chr.1: 2193140-2193140 on Build GRCh38
rs577550357 Chr.1: 2192677-2192677 on Build GRCh38
rs924236030 Chr.1: 2193108-2193108 on Build GRCh38
rs1039946922 Chr.1: 2192833-2192833 on Build GRCh38
rs115162458 Chr.1: 2193035-2193035 on Build GRCh38
rs866210297 Chr.1: 2192907-2192907 on Build GRCh38
rs887875695 Chr.1: 2192999-2192999 on Build GRCh38
rs929527195 Chr.1: 2193149-2193149 on Build GRCh38
rs935797370 Chr.1: 2193113-2193113 on Build GRCh38
rs1012819082 Chr.1: 2192646-2192647 on Build GRCh38
rs1047755714 Chr.1: 2193150-2193150 on Build GRCh38
rs559230284 Chr.1: 2192983-2192983 on Build GRCh38
rs571837250 Chr.1: 2193107-2193107 on Build GRCh38
rs975778434 Chr.1: 2193097-2193097 on Build GRCh38
rs757323268 Chr.1: 2193092-2193092 on Build GRCh38
rs528172736 Chr.1: 2193009-2193009 on Build GRCh38
rs775716114 Chr.1: 2193019-2193019 on Build GRCh38
rs543276998 Chr.1: 2192761-2192761 on Build GRCh38
rs934176205 Chr.1: 2192879-2192879 on Build GRCh38
rs769285784 Chr.1: 2192872-2192872 on Build GRCh38
rs542599272 Chr.1: 2192908-2192908 on Build GRCh38
rs72925890 Chr.1: 2193142-2193142 on Build GRCh38
rs984114925 Chr.1: 2192760-2192760 on Build GRCh38
rs563316537 Chr.1: 2192776-2192776 on Build GRCh38
rs113874353 Chr.1: 2192943-2192943 on Build GRCh38
rs754855519 Chr.1: 2192843-2192843 on Build GRCh38
rs752983330 Chr.1: 2192907-2192909 on Build GRCh38
rs1026233816 Chr.1: 2192977-2192977 on Build GRCh38
rs746228536 Chr.1: 2192779-2192779 on Build GRCh38
rs775822613 Chr.1: 2192909-2192910 on Build GRCh38
rs1024599550 Chr.1: 2192646-2192646 on Build GRCh38
rs562868733 Chr.1: 2192780-2192780 on Build GRCh38
rs878870861 Chr.1: 2193079-2193079 on Build GRCh38
rs916112400 Chr.1: 2193130-2193130 on Build GRCh38
rs746106653 Chr.1: 2192952-2192952 on Build GRCh38
rs770133643 Chr.1: 2192960-2192960 on Build GRCh38
rs948914840 Chr.1: 2193135-2193135 on Build GRCh38
rs972274564 Chr.1: 2192699-2192699 on Build GRCh38
rs933631281 Chr.1: 2192858-2192858 on Build GRCh38
rs1045855820 Chr.1: 2193139-2193139 on Build GRCh38
rs1026075273 Chr.1: 2192990-2192990 on Build GRCh38
rs975354732 Chr.1: 2193039-2193039 on Build GRCh38
rs1064455 Chr.1: 2192933-2192933 on Build GRCh38
rs984167195 Chr.1: 2193001-2193001 on Build GRCh38
rs749172105 Chr.1: 2192812-2192812 on Build GRCh38
rs778867523 Chr.1: 2192853-2192853 on Build GRCh38
rs964083886 Chr.1: 2193056-2193057 on Build GRCh38
rs547846697 Chr.1: 2192803-2192803 on Build GRCh38
rs971248214 Chr.1: 2192661-2192661 on Build GRCh38
rs747057779 Chr.1: 2192880-2192880 on Build GRCh38

View Full Product Details