Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37101200-37101785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH8
Entrez Gene ID
140699
Gene Name
maestro heat like repeat family member 8
Gene Aliases
C20orf131, C20orf132, dJ621N11.3, dJ621N11.4
UniGene
Hs.349125
Gene Chromosome Location
Chr.20: 37101226-37179588 on Build GRCh38
Transcripts
NM_152503.5

SNP Detail

SNP ID Chromosome Location
rs919135519 Chr.20: 37101644-37101644 on Build GRCh38
rs912679113 Chr.20: 37101617-37101617 on Build GRCh38
rs1008480083 Chr.20: 37101648-37101648 on Build GRCh38
rs535018280 Chr.20: 37101717-37101717 on Build GRCh38
rs887497590 Chr.20: 37101470-37101470 on Build GRCh38
rs986621824 Chr.20: 37101325-37101325 on Build GRCh38
rs971545192 Chr.20: 37101495-37101495 on Build GRCh38
rs996511203 Chr.20: 37101598-37101598 on Build GRCh38
rs181009694 Chr.20: 37101317-37101317 on Build GRCh38
rs926698908 Chr.20: 37101429-37101429 on Build GRCh38
rs1044163005 Chr.20: 37101692-37101693 on Build GRCh38
rs961186236 Chr.20: 37101697-37101697 on Build GRCh38
rs908472217 Chr.20: 37101354-37101354 on Build GRCh38
rs111716163 Chr.20: 37101541-37101541 on Build GRCh38
rs929252878 Chr.20: 37101645-37101645 on Build GRCh38
rs939534998 Chr.20: 37101441-37101441 on Build GRCh38
rs192355939 Chr.20: 37101602-37101602 on Build GRCh38
rs1056599206 Chr.20: 37101455-37101455 on Build GRCh38
rs956322273 Chr.20: 37101533-37101533 on Build GRCh38
rs972046137 Chr.20: 37101393-37101393 on Build GRCh38
rs574858450 Chr.20: 37101755-37101755 on Build GRCh38
rs528101826 Chr.20: 37101395-37101395 on Build GRCh38
rs568143112 Chr.20: 37101440-37101440 on Build GRCh38
rs1050191829 Chr.20: 37101320-37101320 on Build GRCh38
rs1015346273 Chr.20: 37101688-37101688 on Build GRCh38
rs1013055874 Chr.20: 37101231-37101231 on Build GRCh38
rs1021936379 Chr.20: 37101311-37101311 on Build GRCh38
rs955411579 Chr.20: 37101321-37101321 on Build GRCh38
rs78235784 Chr.20: 37101437-37101438 on Build GRCh38
rs6017272 Chr.20: 37101712-37101712 on Build GRCh38
rs973986035 Chr.20: 37101714-37101714 on Build GRCh38
rs961237671 Chr.20: 37101370-37101370 on Build GRCh38
rs553135914 Chr.20: 37101749-37101749 on Build GRCh38
rs149935918 Chr.20: 37101508-37101508 on Build GRCh38
rs147267250 Chr.20: 37101313-37101313 on Build GRCh38
rs530800346 Chr.20: 37101312-37101312 on Build GRCh38
rs982085556 Chr.20: 37101260-37101260 on Build GRCh38
rs1031851322 Chr.20: 37101738-37101738 on Build GRCh38
rs1040209925 Chr.20: 37101644-37101645 on Build GRCh38
rs1040685788 Chr.20: 37101354-37101354 on Build GRCh38
rs187070538 Chr.20: 37101536-37101536 on Build GRCh38
rs1023153153 Chr.20: 37101238-37101239 on Build GRCh38
rs1012690519 Chr.20: 37101472-37101472 on Build GRCh38
rs374738329 Chr.20: 37101743-37101743 on Build GRCh38
rs971618668 Chr.20: 37101240-37101240 on Build GRCh38
rs1003078372 Chr.20: 37101504-37101504 on Build GRCh38
rs74417908 Chr.20: 37101414-37101414 on Build GRCh38
rs920636299 Chr.20: 37101394-37101394 on Build GRCh38
rs887425686 Chr.20: 37101675-37101675 on Build GRCh38
rs555023896 Chr.20: 37101479-37101479 on Build GRCh38
rs546701579 Chr.20: 37101401-37101401 on Build GRCh38
rs548743758 Chr.20: 37101515-37101515 on Build GRCh38
rs944539704 Chr.20: 37101621-37101621 on Build GRCh38
rs1780705 Chr.20: 37101547-37101547 on Build GRCh38
rs112684688 Chr.20: 37101276-37101276 on Build GRCh38
rs924919191 Chr.20: 37101270-37101270 on Build GRCh38
rs955127813 Chr.20: 37101625-37101625 on Build GRCh38

View Full Product Details