Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19847604-19848185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf29
Entrez Gene ID
79680
Gene Name
chromosome 22 open reading frame 29
Gene Aliases
BOP
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19846138-19854848 on Build GRCh38
Transcripts
NM_024627.5 NM_053004.2

Gene Symbol
GNB1L
Entrez Gene ID
54584
Gene Name
G protein subunit beta 1 like
Gene Aliases
DGCRK3, FKSG1, GY2, WDR14, WDVCF
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19788411-19854939 on Build GRCh38
Transcripts
NM_024627.5 NM_053004.2

SNP Detail

SNP ID Chromosome Location
rs763812481 Chr.22: 19847701-19847701 on Build GRCh38
rs966354996 Chr.22: 19848111-19848111 on Build GRCh38
rs550430162 Chr.22: 19847671-19847671 on Build GRCh38
rs148256752 Chr.22: 19848135-19848139 on Build GRCh38
rs1046464240 Chr.22: 19847737-19847737 on Build GRCh38
rs796279064 Chr.22: 19847805-19847806 on Build GRCh38
rs531784946 Chr.22: 19847711-19847711 on Build GRCh38
rs71186631 Chr.22: 19847805-19847806 on Build GRCh38
rs370925254 Chr.22: 19848033-19848033 on Build GRCh38
rs888417016 Chr.22: 19847804-19847806 on Build GRCh38
rs911445455 Chr.22: 19847630-19847630 on Build GRCh38
rs538551374 Chr.22: 19847981-19847981 on Build GRCh38
rs897143868 Chr.22: 19847741-19847741 on Build GRCh38
rs527756180 Chr.22: 19847865-19847865 on Build GRCh38
rs753396913 Chr.22: 19847748-19847748 on Build GRCh38
rs945689599 Chr.22: 19847744-19847744 on Build GRCh38
rs888515165 Chr.22: 19848025-19848025 on Build GRCh38
rs1016094439 Chr.22: 19847804-19847810 on Build GRCh38
rs980901970 Chr.22: 19847833-19847833 on Build GRCh38
rs888625438 Chr.22: 19847770-19847770 on Build GRCh38
rs990642077 Chr.22: 19847910-19847910 on Build GRCh38
rs1027064734 Chr.22: 19847760-19847760 on Build GRCh38
rs970874897 Chr.22: 19847673-19847673 on Build GRCh38
rs929927964 Chr.22: 19847978-19847978 on Build GRCh38
rs922736779 Chr.22: 19847918-19847918 on Build GRCh38
rs936503169 Chr.22: 19847894-19847894 on Build GRCh38
rs73379999 Chr.22: 19848026-19848026 on Build GRCh38
rs925155802 Chr.22: 19847680-19847680 on Build GRCh38
rs770537435 Chr.22: 19847806-19847807 on Build GRCh38
rs932708538 Chr.22: 19847764-19847764 on Build GRCh38
rs189463496 Chr.22: 19847690-19847690 on Build GRCh38
rs114499579 Chr.22: 19847956-19847956 on Build GRCh38
rs34331843 Chr.22: 19847804-19847805 on Build GRCh38
rs1002046543 Chr.22: 19848061-19848061 on Build GRCh38
rs1025214634 Chr.22: 19847827-19847827 on Build GRCh38
rs924997489 Chr.22: 19847893-19847893 on Build GRCh38
rs998484070 Chr.22: 19848070-19848070 on Build GRCh38
rs945608676 Chr.22: 19847866-19847866 on Build GRCh38
rs1042101182 Chr.22: 19847672-19847672 on Build GRCh38
rs1049764670 Chr.22: 19847932-19847932 on Build GRCh38
rs756799717 Chr.22: 19847928-19847928 on Build GRCh38
rs977843273 Chr.22: 19847873-19847873 on Build GRCh38
rs80122683 Chr.22: 19847995-19847995 on Build GRCh38
rs1020615941 Chr.22: 19848158-19848158 on Build GRCh38
rs1005514045 Chr.22: 19847804-19847808 on Build GRCh38
rs548280989 Chr.22: 19847771-19847771 on Build GRCh38
rs73877361 Chr.22: 19847804-19847804 on Build GRCh38
rs975749735 Chr.22: 19847654-19847654 on Build GRCh38
rs977686468 Chr.22: 19848138-19848138 on Build GRCh38
rs1018959510 Chr.22: 19847790-19847790 on Build GRCh38
rs546789419 Chr.22: 19847940-19847940 on Build GRCh38
rs901319361 Chr.22: 19848036-19848036 on Build GRCh38
rs537966586 Chr.22: 19847693-19847693 on Build GRCh38
rs964733038 Chr.22: 19847651-19847651 on Build GRCh38
rs888532933 Chr.22: 19847942-19847942 on Build GRCh38
rs554328494 Chr.22: 19848106-19848106 on Build GRCh38
rs914162075 Chr.22: 19847714-19847714 on Build GRCh38
rs977053946 Chr.22: 19847888-19847888 on Build GRCh38
rs989259341 Chr.22: 19848143-19848143 on Build GRCh38
rs895040629 Chr.22: 19847694-19847694 on Build GRCh38
rs1020484313 Chr.22: 19848110-19848110 on Build GRCh38
rs977395384 Chr.22: 19847858-19847858 on Build GRCh38
rs993461197 Chr.22: 19847807-19847807 on Build GRCh38
rs916387985 Chr.22: 19847919-19847919 on Build GRCh38
rs970606452 Chr.22: 19847833-19847833 on Build GRCh38
rs1033554116 Chr.22: 19848068-19848068 on Build GRCh38
rs141316334 Chr.22: 19847862-19847862 on Build GRCh38
rs1033437735 Chr.22: 19847851-19847851 on Build GRCh38
rs945157024 Chr.22: 19847660-19847660 on Build GRCh38
rs150981910 Chr.22: 19847656-19847656 on Build GRCh38
rs897743993 Chr.22: 19847805-19847805 on Build GRCh38
rs932846146 Chr.22: 19847920-19847920 on Build GRCh38
rs897194079 Chr.22: 19847974-19847974 on Build GRCh38

View Full Product Details