Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49270324-49270883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf62
Entrez Gene ID
375341
Gene Name
chromosome 3 open reading frame 62
Gene Aliases
-
UniGene
Hs.403828
Gene Chromosome Location
Chr.3: 49268597-49277075 on Build GRCh38
Transcripts
NM_198562.2

SNP Detail

SNP ID Chromosome Location
rs555401706 Chr.3: 49270592-49270592 on Build GRCh38
rs527446074 Chr.3: 49270846-49270846 on Build GRCh38
rs903097465 Chr.3: 49270650-49270650 on Build GRCh38
rs112370052 Chr.3: 49270599-49270599 on Build GRCh38
rs577250090 Chr.3: 49270612-49270612 on Build GRCh38
rs953555108 Chr.3: 49270419-49270419 on Build GRCh38
rs953259126 Chr.3: 49270715-49270715 on Build GRCh38
rs771064466 Chr.3: 49270355-49270357 on Build GRCh38
rs534146492 Chr.3: 49270589-49270589 on Build GRCh38
rs1011233177 Chr.3: 49270623-49270623 on Build GRCh38
rs916382877 Chr.3: 49270740-49270740 on Build GRCh38
rs982398711 Chr.3: 49270847-49270847 on Build GRCh38
rs1016290696 Chr.3: 49270724-49270724 on Build GRCh38
rs573358669 Chr.3: 49270582-49270582 on Build GRCh38
rs919502523 Chr.3: 49270576-49270576 on Build GRCh38
rs180688401 Chr.3: 49270463-49270463 on Build GRCh38
rs185733885 Chr.3: 49270610-49270610 on Build GRCh38
rs997442420 Chr.3: 49270701-49270701 on Build GRCh38
rs916454006 Chr.3: 49270411-49270411 on Build GRCh38
rs545266548 Chr.3: 49270682-49270682 on Build GRCh38
rs923848550 Chr.3: 49270824-49270824 on Build GRCh38
rs754166845 Chr.3: 49270803-49270803 on Build GRCh38
rs560208865 Chr.3: 49270786-49270786 on Build GRCh38
rs879389086 Chr.3: 49270365-49270365 on Build GRCh38
rs972673796 Chr.3: 49270669-49270669 on Build GRCh38
rs1019994484 Chr.3: 49270630-49270630 on Build GRCh38
rs963466432 Chr.3: 49270802-49270802 on Build GRCh38
rs934984701 Chr.3: 49270469-49270469 on Build GRCh38
rs144694808 Chr.3: 49270577-49270577 on Build GRCh38
rs368066415 Chr.3: 49270501-49270501 on Build GRCh38
rs566276166 Chr.3: 49270534-49270535 on Build GRCh38
rs551648533 Chr.3: 49270354-49270355 on Build GRCh38
rs1030552569 Chr.3: 49270714-49270714 on Build GRCh38
rs543948087 Chr.3: 49270609-49270609 on Build GRCh38
rs961941421 Chr.3: 49270624-49270624 on Build GRCh38
rs1030201241 Chr.3: 49270392-49270392 on Build GRCh38
rs141330633 Chr.3: 49270573-49270573 on Build GRCh38
rs990893811 Chr.3: 49270737-49270737 on Build GRCh38
rs11130187 Chr.3: 49270439-49270439 on Build GRCh38
rs146603295 Chr.3: 49270507-49270507 on Build GRCh38
rs952795315 Chr.3: 49270732-49270732 on Build GRCh38
rs986395012 Chr.3: 49270719-49270719 on Build GRCh38
rs919263353 Chr.3: 49270804-49270804 on Build GRCh38
rs893522906 Chr.3: 49270508-49270508 on Build GRCh38
rs905367842 Chr.3: 49270593-49270593 on Build GRCh38
rs982367079 Chr.3: 49270810-49270810 on Build GRCh38
rs990573077 Chr.3: 49270355-49270358 on Build GRCh38

View Full Product Details