Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37591841-37592410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf19
Entrez Gene ID
55286
Gene Name
chromosome 4 open reading frame 19
Gene Aliases
-
UniGene
Hs.107527
Gene Chromosome Location
Chr.4: 37453454-37596146 on Build GRCh38
Transcripts
NM_001104629.1 NM_018302.2 XM_011513712.2 XM_011513713.2 XM_017008361.1 NM_001085399.1

Gene Symbol
RELL1
Entrez Gene ID
768211
Gene Name
RELT like 1
Gene Aliases
-
UniGene
Hs.283378
Gene Chromosome Location
Chr.4: 37590800-37686377 on Build GRCh38
Transcripts
NM_001104629.1 NM_018302.2 XM_011513712.2 XM_011513713.2 XM_017008361.1 NM_001085399.1

SNP Detail

SNP ID Chromosome Location
rs564405245 Chr.4: 37592075-37592075 on Build GRCh38
rs972811338 Chr.4: 37592102-37592102 on Build GRCh38
rs796661728 Chr.4: 37592228-37592228 on Build GRCh38
rs1056830956 Chr.4: 37592098-37592098 on Build GRCh38
rs1020522176 Chr.4: 37591935-37591935 on Build GRCh38
rs973225707 Chr.4: 37592070-37592070 on Build GRCh38
rs1000344575 Chr.4: 37591968-37591968 on Build GRCh38
rs180678683 Chr.4: 37592202-37592202 on Build GRCh38
rs1018442360 Chr.4: 37592285-37592285 on Build GRCh38
rs941946341 Chr.4: 37592065-37592065 on Build GRCh38
rs148549235 Chr.4: 37592076-37592076 on Build GRCh38
rs560066864 Chr.4: 37592080-37592080 on Build GRCh38
rs569257883 Chr.4: 37592220-37592220 on Build GRCh38
rs755772926 Chr.4: 37591931-37591931 on Build GRCh38
rs963315971 Chr.4: 37592380-37592380 on Build GRCh38
rs945686791 Chr.4: 37592173-37592173 on Build GRCh38
rs904784724 Chr.4: 37591948-37591948 on Build GRCh38
rs78397437 Chr.4: 37592212-37592212 on Build GRCh38
rs985440439 Chr.4: 37592190-37592190 on Build GRCh38
rs764456904 Chr.4: 37592210-37592211 on Build GRCh38
rs540392992 Chr.4: 37592077-37592077 on Build GRCh38
rs553138711 Chr.4: 37592337-37592337 on Build GRCh38
rs575504185 Chr.4: 37592040-37592040 on Build GRCh38
rs572966792 Chr.4: 37592041-37592041 on Build GRCh38
rs553929132 Chr.4: 37592257-37592257 on Build GRCh38
rs905570784 Chr.4: 37591876-37591876 on Build GRCh38
rs369293188 Chr.4: 37592352-37592353 on Build GRCh38
rs1025136413 Chr.4: 37592050-37592050 on Build GRCh38
rs11403816 Chr.4: 37592372-37592373 on Build GRCh38
rs895522512 Chr.4: 37591930-37591930 on Build GRCh38
rs943272714 Chr.4: 37592213-37592213 on Build GRCh38
rs188371592 Chr.4: 37591977-37591977 on Build GRCh38
rs66819746 Chr.4: 37592354-37592354 on Build GRCh38
rs534566517 Chr.4: 37592354-37592355 on Build GRCh38
rs1027488858 Chr.4: 37592110-37592110 on Build GRCh38
rs63456163 Chr.4: 37592211-37592211 on Build GRCh38
rs752716264 Chr.4: 37591974-37591974 on Build GRCh38
rs112219673 Chr.4: 37592354-37592354 on Build GRCh38
rs998260225 Chr.4: 37592326-37592326 on Build GRCh38
rs1041793792 Chr.4: 37592254-37592254 on Build GRCh38
rs35282918 Chr.4: 37592353-37592354 on Build GRCh38
rs964154773 Chr.4: 37591975-37591975 on Build GRCh38
rs945144546 Chr.4: 37591936-37591936 on Build GRCh38
rs544421798 Chr.4: 37592064-37592064 on Build GRCh38
rs536955651 Chr.4: 37592033-37592033 on Build GRCh38
rs184410654 Chr.4: 37592378-37592378 on Build GRCh38
rs539051130 Chr.4: 37591894-37591894 on Build GRCh38
rs976445350 Chr.4: 37592235-37592235 on Build GRCh38
rs397881399 Chr.4: 37592210-37592210 on Build GRCh38
rs1029725183 Chr.4: 37592336-37592336 on Build GRCh38
rs1045553340 Chr.4: 37592287-37592287 on Build GRCh38
rs1000890200 Chr.4: 37592280-37592280 on Build GRCh38
rs576984055 Chr.4: 37592164-37592164 on Build GRCh38
rs900022727 Chr.4: 37592313-37592313 on Build GRCh38
rs367544040 Chr.4: 37592263-37592263 on Build GRCh38
rs535409733 Chr.4: 37592011-37592011 on Build GRCh38
rs754518539 Chr.4: 37592209-37592211 on Build GRCh38
rs895473211 Chr.4: 37592103-37592103 on Build GRCh38
rs982743178 Chr.4: 37592358-37592358 on Build GRCh38
rs34307749 Chr.4: 37592228-37592228 on Build GRCh38
rs76722421 Chr.4: 37591934-37591934 on Build GRCh38
rs951391996 Chr.4: 37592035-37592035 on Build GRCh38
rs1056715522 Chr.4: 37591904-37591904 on Build GRCh38
rs923094278 Chr.4: 37592247-37592247 on Build GRCh38
rs529260178 Chr.4: 37592195-37592195 on Build GRCh38

View Full Product Details