Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133955742-133956291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf15
Entrez Gene ID
56951
Gene Name
chromosome 5 open reading frame 15
Gene Aliases
HTGN29, KCT2
UniGene
Hs.744034
Gene Chromosome Location
Chr.5: 133955507-133968715 on Build GRCh38
Transcripts
NM_020199.2

SNP Detail

SNP ID Chromosome Location
rs1801366 Chr.5: 133955940-133955940 on Build GRCh38
rs951164649 Chr.5: 133955870-133955870 on Build GRCh38
rs949985720 Chr.5: 133956023-133956023 on Build GRCh38
rs576139132 Chr.5: 133956266-133956266 on Build GRCh38
rs943504738 Chr.5: 133956177-133956177 on Build GRCh38
rs778894019 Chr.5: 133955824-133955824 on Build GRCh38
rs934769095 Chr.5: 133956061-133956061 on Build GRCh38
rs991386369 Chr.5: 133955962-133955962 on Build GRCh38
rs902110736 Chr.5: 133956207-133956207 on Build GRCh38
rs962513987 Chr.5: 133955960-133955960 on Build GRCh38
rs570612814 Chr.5: 133955777-133955777 on Build GRCh38
rs146658599 Chr.5: 133956255-133956255 on Build GRCh38
rs1017851877 Chr.5: 133955775-133955775 on Build GRCh38
rs998136373 Chr.5: 133956213-133956213 on Build GRCh38
rs1029717793 Chr.5: 133955832-133955832 on Build GRCh38
rs1029338086 Chr.5: 133956246-133956246 on Build GRCh38
rs772028637 Chr.5: 133956071-133956071 on Build GRCh38
rs10422 Chr.5: 133955951-133955951 on Build GRCh38
rs915814862 Chr.5: 133955991-133955991 on Build GRCh38
rs886767524 Chr.5: 133956251-133956251 on Build GRCh38
rs772946447 Chr.5: 133956134-133956134 on Build GRCh38
rs144708470 Chr.5: 133955938-133955938 on Build GRCh38
rs997875400 Chr.5: 133955822-133955822 on Build GRCh38
rs187244711 Chr.5: 133956130-133956130 on Build GRCh38
rs190474658 Chr.5: 133956247-133956247 on Build GRCh38
rs1039585131 Chr.5: 133956203-133956203 on Build GRCh38
rs909553856 Chr.5: 133955771-133955771 on Build GRCh38
rs981470016 Chr.5: 133956024-133956024 on Build GRCh38
rs1016618300 Chr.5: 133956262-133956262 on Build GRCh38
rs924575908 Chr.5: 133956047-133956047 on Build GRCh38
rs1036939628 Chr.5: 133955778-133955778 on Build GRCh38
rs182902801 Chr.5: 133956099-133956099 on Build GRCh38
rs893333628 Chr.5: 133956083-133956083 on Build GRCh38
rs559234726 Chr.5: 133956069-133956069 on Build GRCh38
rs1054542362 Chr.5: 133956070-133956070 on Build GRCh38
rs1044089598 Chr.5: 133955770-133955770 on Build GRCh38
rs575428655 Chr.5: 133956172-133956172 on Build GRCh38

View Full Product Details