Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90273565-90274120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP69
Entrez Gene ID
79846
Gene Name
cilia and flagella associated protein 69
Gene Aliases
C7orf63, FAP69
UniGene
Hs.521014
Gene Chromosome Location
Chr.7: 90245174-90320964 on Build GRCh38
Transcripts
NM_001039706.2 NM_001160138.1 XM_005250600.3 XM_011516577.2 XM_011516578.2 XM_011516579.1 XM_017012624.1 XM_017012625.1 XM_017012626.1 XM_017012627.1 XM_017012628.1 XM_017012629.1 XM_017012630.1 XM_017012631.1 XM_017012632.1 XM_017012633.1 XM_017012634.1 XM_017012635.1 XM_017012636.1 XM_017012637.1 XM_017012638.1 XM_017012639.1 XM_017012640.1 XM_017012641.1 XM_017012642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5498653 c.903C>T p.F301F Chr.7: 90274029-90274029 on Build GRCh38
1728601 c.898G>A p.G300S Chr.7: 90274024-90274024 on Build GRCh38
6420121 c.973G>A p.A325T Chr.7: 90274099-90274099 on Build GRCh38
5498655 c.952A>G p.I318V Chr.7: 90274078-90274078 on Build GRCh38
3431843 c.882A>C p.K294N Chr.7: 90274008-90274008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051542921 Chr.7: 90273761-90273761 on Build GRCh38
rs756616968 Chr.7: 90274025-90274025 on Build GRCh38
rs759945748 Chr.7: 90273783-90273783 on Build GRCh38
rs185909119 Chr.7: 90273979-90273979 on Build GRCh38
rs992312539 Chr.7: 90273635-90273635 on Build GRCh38
rs575397914 Chr.7: 90273933-90273933 on Build GRCh38
rs984407200 Chr.7: 90273600-90273600 on Build GRCh38
rs750199884 Chr.7: 90274006-90274006 on Build GRCh38
rs190904333 Chr.7: 90273877-90273877 on Build GRCh38
rs1046216070 Chr.7: 90273742-90273742 on Build GRCh38
rs1007400115 Chr.7: 90273779-90273779 on Build GRCh38
rs892440744 Chr.7: 90274081-90274081 on Build GRCh38
rs755155695 Chr.7: 90273617-90273617 on Build GRCh38
rs968419087 Chr.7: 90273609-90273609 on Build GRCh38
rs776560830 Chr.7: 90273980-90273980 on Build GRCh38
rs749682826 Chr.7: 90274030-90274030 on Build GRCh38
rs776236775 Chr.7: 90273920-90273920 on Build GRCh38
rs992207702 Chr.7: 90273872-90273872 on Build GRCh38
rs904455479 Chr.7: 90273795-90273795 on Build GRCh38
rs28945134 Chr.7: 90273916-90273916 on Build GRCh38
rs969517879 Chr.7: 90273897-90273897 on Build GRCh38
rs771240711 Chr.7: 90274034-90274034 on Build GRCh38
rs753213245 Chr.7: 90274024-90274024 on Build GRCh38
rs17865126 Chr.7: 90273660-90273660 on Build GRCh38
rs780839211 Chr.7: 90274022-90274022 on Build GRCh38
rs1005793830 Chr.7: 90273770-90273770 on Build GRCh38
rs909480863 Chr.7: 90273671-90273671 on Build GRCh38
rs1025073673 Chr.7: 90273894-90273894 on Build GRCh38
rs761794797 Chr.7: 90273995-90273995 on Build GRCh38
rs182618567 Chr.7: 90273932-90273932 on Build GRCh38
rs776368188 Chr.7: 90274043-90274043 on Build GRCh38
rs17863974 Chr.7: 90273759-90273759 on Build GRCh38
rs1053656287 Chr.7: 90273719-90273719 on Build GRCh38
rs1031028010 Chr.7: 90273866-90273866 on Build GRCh38
rs925991977 Chr.7: 90273675-90273675 on Build GRCh38
rs1013606432 Chr.7: 90274092-90274092 on Build GRCh38
rs778052150 Chr.7: 90274029-90274029 on Build GRCh38
rs937739000 Chr.7: 90273973-90273973 on Build GRCh38
rs17863973 Chr.7: 90273643-90273643 on Build GRCh38
rs979468994 Chr.7: 90273610-90273610 on Build GRCh38
rs749665355 Chr.7: 90273758-90273758 on Build GRCh38
rs943171927 Chr.7: 90273971-90273971 on Build GRCh38
rs145440500 Chr.7: 90273905-90273905 on Build GRCh38
rs775633552 Chr.7: 90274095-90274095 on Build GRCh38
rs1036053447 Chr.7: 90273744-90273744 on Build GRCh38
rs542429356 Chr.7: 90273934-90273934 on Build GRCh38
rs956766144 Chr.7: 90273871-90273871 on Build GRCh38
rs888101722 Chr.7: 90274033-90274033 on Build GRCh38
rs772196992 Chr.7: 90274060-90274060 on Build GRCh38
rs17866223 Chr.7: 90274042-90274042 on Build GRCh38
rs888877582 Chr.7: 90273955-90273955 on Build GRCh38
rs892288129 Chr.7: 90273724-90273724 on Build GRCh38
rs1033514586 Chr.7: 90273786-90273812 on Build GRCh38
rs754551096 Chr.7: 90274017-90274017 on Build GRCh38
rs906238229 Chr.7: 90273754-90273754 on Build GRCh38
rs927837083 Chr.7: 90273941-90273941 on Build GRCh38
rs369150743 Chr.7: 90273890-90273890 on Build GRCh38
rs762608119 Chr.7: 90274014-90274014 on Build GRCh38
rs572884625 Chr.7: 90273962-90273962 on Build GRCh38
rs551651658 Chr.7: 90273645-90273645 on Build GRCh38
rs186499470 Chr.7: 90273865-90273865 on Build GRCh38
rs765007571 Chr.7: 90274004-90274004 on Build GRCh38
rs751192811 Chr.7: 90274016-90274016 on Build GRCh38
rs774504302 Chr.7: 90273957-90273957 on Build GRCh38
rs17863975 Chr.7: 90273936-90273936 on Build GRCh38
rs948207623 Chr.7: 90273644-90273644 on Build GRCh38
rs1051427598 Chr.7: 90273954-90273954 on Build GRCh38
rs139514912 Chr.7: 90273729-90273729 on Build GRCh38
rs932755574 Chr.7: 90273659-90273659 on Build GRCh38
rs17863972 Chr.7: 90273631-90273631 on Build GRCh38
rs936564582 Chr.7: 90273680-90273680 on Build GRCh38
rs980983284 Chr.7: 90273900-90273900 on Build GRCh38
rs113972709 Chr.7: 90273768-90273768 on Build GRCh38

View Full Product Details