Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56885528-56886089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8A
Entrez Gene ID
731
Gene Name
complement component 8 alpha subunit
Gene Aliases
-
UniGene
Hs.93210
Gene Chromosome Location
Chr.1: 56854770-56918221 on Build GRCh38
Transcripts
NM_000562.2 XM_011542079.2 XM_017002234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6246888 c.856-1G>T p.? Chr.1: 56885926-56885926 on Build GRCh38
6063631 c.879C>T p.F293F Chr.1: 56885950-56885950 on Build GRCh38
6991537 c.962T>C p.L321P Chr.1: 56886033-56886033 on Build GRCh38
3491187 c.950C>T p.S317L Chr.1: 56886021-56886021 on Build GRCh38
5037743 c.867C>T p.F289F Chr.1: 56885938-56885938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201414278 Chr.1: 56885556-56885557 on Build GRCh38
rs377378202 Chr.1: 56885977-56885977 on Build GRCh38
rs34914563 Chr.1: 56885580-56885581 on Build GRCh38
rs748788541 Chr.1: 56886047-56886047 on Build GRCh38
rs563439122 Chr.1: 56886021-56886021 on Build GRCh38
rs944851416 Chr.1: 56885703-56885703 on Build GRCh38
rs74075638 Chr.1: 56885915-56885915 on Build GRCh38
rs887095569 Chr.1: 56885735-56885735 on Build GRCh38
rs766398440 Chr.1: 56885983-56885983 on Build GRCh38
rs931218315 Chr.1: 56885717-56885717 on Build GRCh38
rs779669335 Chr.1: 56886041-56886041 on Build GRCh38
rs375634050 Chr.1: 56885914-56885914 on Build GRCh38
rs897049063 Chr.1: 56885804-56885804 on Build GRCh38
rs140024291 Chr.1: 56885665-56885665 on Build GRCh38
rs139789526 Chr.1: 56885772-56885772 on Build GRCh38
rs856847 Chr.1: 56885815-56885815 on Build GRCh38
rs140619602 Chr.1: 56885908-56885908 on Build GRCh38
rs888577267 Chr.1: 56885569-56885569 on Build GRCh38
rs1049423801 Chr.1: 56885742-56885742 on Build GRCh38
rs77289345 Chr.1: 56885938-56885938 on Build GRCh38
rs761659751 Chr.1: 56885953-56885953 on Build GRCh38
rs188323392 Chr.1: 56885565-56885565 on Build GRCh38
rs949347512 Chr.1: 56886013-56886013 on Build GRCh38
rs1036934457 Chr.1: 56885798-56885798 on Build GRCh38
rs1012314913 Chr.1: 56886028-56886028 on Build GRCh38
rs1032556082 Chr.1: 56885564-56885564 on Build GRCh38
rs769359160 Chr.1: 56885913-56885913 on Build GRCh38
rs894081772 Chr.1: 56885816-56885816 on Build GRCh38
rs1003801837 Chr.1: 56885805-56885805 on Build GRCh38
rs759999228 Chr.1: 56885994-56885994 on Build GRCh38
rs772254769 Chr.1: 56885959-56885959 on Build GRCh38
rs369233365 Chr.1: 56885881-56885881 on Build GRCh38
rs570673430 Chr.1: 56885555-56885556 on Build GRCh38
rs570532832 Chr.1: 56885676-56885676 on Build GRCh38
rs372529913 Chr.1: 56885912-56885912 on Build GRCh38
rs555688426 Chr.1: 56885880-56885880 on Build GRCh38
rs1019976407 Chr.1: 56885859-56885859 on Build GRCh38
rs966289684 Chr.1: 56885843-56885843 on Build GRCh38
rs918509649 Chr.1: 56885974-56885974 on Build GRCh38
rs536090836 Chr.1: 56885832-56885832 on Build GRCh38
rs888108683 Chr.1: 56885764-56885764 on Build GRCh38
rs1047080125 Chr.1: 56885732-56885732 on Build GRCh38
rs542666529 Chr.1: 56886051-56886051 on Build GRCh38
rs999495257 Chr.1: 56886067-56886067 on Build GRCh38
rs758786260 Chr.1: 56886009-56886009 on Build GRCh38
rs565821004 Chr.1: 56885575-56885575 on Build GRCh38
rs755705114 Chr.1: 56886038-56886038 on Build GRCh38
rs139867818 Chr.1: 56885968-56885968 on Build GRCh38
rs147513844 Chr.1: 56886031-56886031 on Build GRCh38
rs112132683 Chr.1: 56885629-56885629 on Build GRCh38
rs778909267 Chr.1: 56886063-56886063 on Build GRCh38
rs562380219 Chr.1: 56886016-56886016 on Build GRCh38
rs866770672 Chr.1: 56885980-56885980 on Build GRCh38
rs752607032 Chr.1: 56885586-56885586 on Build GRCh38
rs1034827080 Chr.1: 56885810-56885810 on Build GRCh38
rs545622749 Chr.1: 56885620-56885620 on Build GRCh38
rs765750603 Chr.1: 56885996-56885996 on Build GRCh38
rs753059924 Chr.1: 56886003-56886003 on Build GRCh38
rs922357058 Chr.1: 56885738-56885738 on Build GRCh38
rs555772990 Chr.1: 56885835-56885835 on Build GRCh38
rs756908882 Chr.1: 56885905-56885905 on Build GRCh38
rs779105028 Chr.1: 56885884-56885884 on Build GRCh38
rs1056741518 Chr.1: 56885997-56885997 on Build GRCh38
rs968008323 Chr.1: 56885831-56885831 on Build GRCh38
rs1031688594 Chr.1: 56885840-56885840 on Build GRCh38
rs976253422 Chr.1: 56885708-56885708 on Build GRCh38
rs781320253 Chr.1: 56885884-56885885 on Build GRCh38
rs114723546 Chr.1: 56885886-56885886 on Build GRCh38
rs112987054 Chr.1: 56885739-56885739 on Build GRCh38
rs181097681 Chr.1: 56885573-56885573 on Build GRCh38
rs942472981 Chr.1: 56885698-56885698 on Build GRCh38
rs773356663 Chr.1: 56885963-56885963 on Build GRCh38
rs967149031 Chr.1: 56885634-56885634 on Build GRCh38
rs1022286732 Chr.1: 56885827-56885827 on Build GRCh38
rs768609847 Chr.1: 56885923-56885923 on Build GRCh38
rs754376076 Chr.1: 56885991-56885991 on Build GRCh38
rs376936908 Chr.1: 56885812-56885812 on Build GRCh38
rs912419514 Chr.1: 56885684-56885684 on Build GRCh38

View Full Product Details