Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 544552-545101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRP
Entrez Gene ID
157695
Gene Name
testis development related protein
Gene Aliases
C8orf42, Inm01, TDRP1, TDRP2
UniGene
Hs.289293
Gene Chromosome Location
Chr.8: 489790-545781 on Build GRCh38
Transcripts
NM_001256113.1 NM_175075.4 XM_006716232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3786009 c.21C>A p.G7G Chr.8: 544737-544737 on Build GRCh38
128722 c.94_99delGCCGCC p.A32_A33delAA Chr.8: 544659-544664 on Build GRCh38
4959211 c.14G>A p.G5D Chr.8: 544744-544744 on Build GRCh38
1199036 c.59G>T p.G20V Chr.8: 544699-544699 on Build GRCh38
5445771 c.82_87delGCCGCC p.A32_A33delAA Chr.8: 544671-544676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007820438 Chr.8: 544816-544816 on Build GRCh38
rs372797729 Chr.8: 544625-544625 on Build GRCh38
rs1055788694 Chr.8: 545076-545076 on Build GRCh38
rs539456513 Chr.8: 544665-544665 on Build GRCh38
rs749571609 Chr.8: 544637-544637 on Build GRCh38
rs909358682 Chr.8: 544846-544846 on Build GRCh38
rs1017677260 Chr.8: 544580-544580 on Build GRCh38
rs761824588 Chr.8: 544672-544672 on Build GRCh38
rs993569610 Chr.8: 544835-544835 on Build GRCh38
rs975687229 Chr.8: 544832-544832 on Build GRCh38
rs557298028 Chr.8: 544806-544806 on Build GRCh38
rs1017332450 Chr.8: 544817-544817 on Build GRCh38
rs1025970151 Chr.8: 544836-544836 on Build GRCh38
rs985781370 Chr.8: 544989-544989 on Build GRCh38
rs946400643 Chr.8: 544977-544977 on Build GRCh38
rs927794615 Chr.8: 544871-544871 on Build GRCh38
rs903066233 Chr.8: 544803-544803 on Build GRCh38
rs893063323 Chr.8: 544874-544874 on Build GRCh38
rs938162738 Chr.8: 544849-544849 on Build GRCh38
rs1050705820 Chr.8: 544811-544811 on Build GRCh38
rs934553671 Chr.8: 544863-544863 on Build GRCh38
rs572880491 Chr.8: 545032-545032 on Build GRCh38
rs576869074 Chr.8: 544830-544830 on Build GRCh38
rs559466526 Chr.8: 544980-544980 on Build GRCh38
rs764365534 Chr.8: 544721-544721 on Build GRCh38
rs780222845 Chr.8: 544788-544788 on Build GRCh38
rs982376720 Chr.8: 544862-544862 on Build GRCh38
rs566646227 Chr.8: 544691-544691 on Build GRCh38
rs1042401896 Chr.8: 544857-544857 on Build GRCh38
rs907206391 Chr.8: 545047-545047 on Build GRCh38
rs1040957711 Chr.8: 545074-545074 on Build GRCh38
rs910820448 Chr.8: 545007-545007 on Build GRCh38
rs757632990 Chr.8: 544737-544737 on Build GRCh38
rs949455304 Chr.8: 544856-544856 on Build GRCh38
rs760219453 Chr.8: 544688-544688 on Build GRCh38
rs906278005 Chr.8: 544611-544611 on Build GRCh38
rs780088725 Chr.8: 544631-544631 on Build GRCh38
rs769868971 Chr.8: 544603-544603 on Build GRCh38
rs909421641 Chr.8: 544767-544767 on Build GRCh38
rs938075837 Chr.8: 544773-544773 on Build GRCh38
rs189573614 Chr.8: 545080-545080 on Build GRCh38
rs866391923 Chr.8: 544671-544671 on Build GRCh38
rs1009065910 Chr.8: 544680-544680 on Build GRCh38
rs553014928 Chr.8: 544674-544674 on Build GRCh38
rs965714031 Chr.8: 544588-544588 on Build GRCh38
rs892016016 Chr.8: 544910-544910 on Build GRCh38
rs927731302 Chr.8: 544792-544792 on Build GRCh38
rs1050596073 Chr.8: 544668-544668 on Build GRCh38
rs375431855 Chr.8: 544801-544801 on Build GRCh38
rs947811875 Chr.8: 544578-544578 on Build GRCh38
rs984916973 Chr.8: 544765-544765 on Build GRCh38
rs991413570 Chr.8: 544851-544851 on Build GRCh38
rs574664098 Chr.8: 544802-544802 on Build GRCh38
rs554500161 Chr.8: 544795-544795 on Build GRCh38
rs772020843 Chr.8: 544805-544805 on Build GRCh38
rs531921590 Chr.8: 544664-544665 on Build GRCh38
rs1001603560 Chr.8: 545053-545053 on Build GRCh38
rs894085737 Chr.8: 544662-544662 on Build GRCh38
rs952646562 Chr.8: 544728-544728 on Build GRCh38
rs949339735 Chr.8: 544793-544793 on Build GRCh38
rs923975333 Chr.8: 544861-544861 on Build GRCh38
rs10105131 Chr.8: 544804-544804 on Build GRCh38
rs756124621 Chr.8: 544769-544769 on Build GRCh38
rs916416376 Chr.8: 544763-544763 on Build GRCh38
rs1045750832 Chr.8: 544920-544920 on Build GRCh38
rs62485665 Chr.8: 544600-544600 on Build GRCh38
rs534792489 Chr.8: 544718-544718 on Build GRCh38
rs763222363 Chr.8: 544659-544661 on Build GRCh38
rs753413202 Chr.8: 544713-544713 on Build GRCh38
rs545833550 Chr.8: 544833-544833 on Build GRCh38
rs983966233 Chr.8: 544743-544743 on Build GRCh38
rs1055243079 Chr.8: 544774-544774 on Build GRCh38
rs985032197 Chr.8: 544843-544843 on Build GRCh38
rs750961863 Chr.8: 544757-544757 on Build GRCh38
rs975741005 Chr.8: 544614-544614 on Build GRCh38
rs893001858 Chr.8: 544812-544812 on Build GRCh38
rs987854636 Chr.8: 544967-544967 on Build GRCh38
rs896574857 Chr.8: 544782-544782 on Build GRCh38
rs773283807 Chr.8: 544677-544677 on Build GRCh38
rs950966977 Chr.8: 544838-544838 on Build GRCh38
rs753058565 Chr.8: 544658-544659 on Build GRCh38
rs879427057 Chr.8: 544700-544700 on Build GRCh38
rs1055132576 Chr.8: 544896-544896 on Build GRCh38
rs866240667 Chr.8: 545015-545015 on Build GRCh38
rs184711936 Chr.8: 544779-544779 on Build GRCh38
rs897962334 Chr.8: 545081-545081 on Build GRCh38
rs71202629 Chr.8: 544659-544664 on Build GRCh38
rs981957628 Chr.8: 544771-544771 on Build GRCh38
rs917967441 Chr.8: 544852-544852 on Build GRCh38
rs914992162 Chr.8: 544975-544975 on Build GRCh38
rs966190599 Chr.8: 544823-544823 on Build GRCh38
rs935024253 Chr.8: 544658-544658 on Build GRCh38
rs751979167 Chr.8: 544733-544733 on Build GRCh38
rs936162658 Chr.8: 544890-544890 on Build GRCh38
rs1054352914 Chr.8: 544872-544872 on Build GRCh38
rs1009510092 Chr.8: 544820-544820 on Build GRCh38
rs781614919 Chr.8: 544742-544742 on Build GRCh38
rs768304012 Chr.8: 544642-544642 on Build GRCh38
rs946043681 Chr.8: 544914-544914 on Build GRCh38

View Full Product Details