Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 13567713-13568112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf48
Entrez Gene ID
157773
Gene Name
chromosome 8 open reading frame 48
Gene Aliases
-
UniGene
Hs.104941
Gene Chromosome Location
Chr.8: 13566843-13568288 on Build GRCh38
Transcripts
NM_001007090.2 XM_017012951.1

Gene Symbol
DLC1
Entrez Gene ID
10395
Gene Name
DLC1 Rho GTPase activating protein
Gene Aliases
ARHGAP7, HP, STARD12, p122-RhoGAP
UniGene
Hs.134296
Gene Chromosome Location
Chr.8: 13083361-13604606 on Build GRCh38
Transcripts
NM_001007090.2 XM_017012951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5636194 c.727C>T p.R243C Chr.8: 13567718-13567718 on Build GRCh38
3432148 c.754C>T p.P252S Chr.8: 13567745-13567745 on Build GRCh38
6353119 c.859C>A p.R287R Chr.8: 13567850-13567850 on Build GRCh38
3318496 c.761T>G p.L254R Chr.8: 13567752-13567752 on Build GRCh38
673725 c.793C>G p.L265V Chr.8: 13567784-13567784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182572686 Chr.8: 13568032-13568032 on Build GRCh38
rs11203497 Chr.8: 13567844-13567844 on Build GRCh38
rs751198675 Chr.8: 13568046-13568046 on Build GRCh38
rs943692701 Chr.8: 13567813-13567813 on Build GRCh38
rs769617734 Chr.8: 13567884-13567884 on Build GRCh38
rs547029149 Chr.8: 13568001-13568001 on Build GRCh38
rs757766184 Chr.8: 13567802-13567802 on Build GRCh38
rs558942676 Chr.8: 13568007-13568007 on Build GRCh38
rs931046677 Chr.8: 13567845-13567845 on Build GRCh38
rs921151972 Chr.8: 13567990-13567990 on Build GRCh38
rs542176418 Chr.8: 13567871-13567871 on Build GRCh38
rs374857532 Chr.8: 13567774-13567774 on Build GRCh38
rs551603600 Chr.8: 13567891-13567891 on Build GRCh38
rs969528956 Chr.8: 13567906-13567906 on Build GRCh38
rs368655522 Chr.8: 13567947-13567947 on Build GRCh38
rs757714808 Chr.8: 13567984-13567984 on Build GRCh38
rs1021004817 Chr.8: 13567902-13567902 on Build GRCh38
rs189908929 Chr.8: 13567953-13567953 on Build GRCh38
rs987937638 Chr.8: 13567925-13567925 on Build GRCh38
rs746547308 Chr.8: 13567827-13567827 on Build GRCh38
rs939776976 Chr.8: 13568065-13568065 on Build GRCh38
rs547918239 Chr.8: 13568051-13568051 on Build GRCh38
rs140504541 Chr.8: 13567850-13567850 on Build GRCh38
rs766448217 Chr.8: 13567901-13567901 on Build GRCh38
rs921128564 Chr.8: 13567744-13567744 on Build GRCh38
rs908268209 Chr.8: 13568053-13568053 on Build GRCh38
rs1039737134 Chr.8: 13567820-13567820 on Build GRCh38
rs764376441 Chr.8: 13567961-13567961 on Build GRCh38
rs769933681 Chr.8: 13567998-13567998 on Build GRCh38
rs920913836 Chr.8: 13567968-13567968 on Build GRCh38
rs536934802 Chr.8: 13567740-13567740 on Build GRCh38
rs180788033 Chr.8: 13567919-13567919 on Build GRCh38
rs200907430 Chr.8: 13567892-13567892 on Build GRCh38
rs373263421 Chr.8: 13567957-13567957 on Build GRCh38
rs1046075131 Chr.8: 13567836-13567836 on Build GRCh38
rs1045097104 Chr.8: 13568084-13568084 on Build GRCh38
rs543114869 Chr.8: 13567969-13567969 on Build GRCh38
rs1048620835 Chr.8: 13567772-13567772 on Build GRCh38
rs763750008 Chr.8: 13567804-13567804 on Build GRCh38
rs758451818 Chr.8: 13567893-13567893 on Build GRCh38
rs1049256774 Chr.8: 13567848-13567848 on Build GRCh38
rs767759297 Chr.8: 13567763-13567763 on Build GRCh38
rs561451447 Chr.8: 13567899-13567899 on Build GRCh38
rs931099912 Chr.8: 13567974-13567974 on Build GRCh38
rs558609269 Chr.8: 13567770-13567770 on Build GRCh38
rs896778856 Chr.8: 13568071-13568072 on Build GRCh38
rs1036582454 Chr.8: 13567897-13567897 on Build GRCh38
rs146773735 Chr.8: 13567793-13567793 on Build GRCh38
rs906180487 Chr.8: 13567837-13567837 on Build GRCh38
rs573459343 Chr.8: 13567870-13567870 on Build GRCh38
rs145664837 Chr.8: 13568060-13568060 on Build GRCh38
rs767563531 Chr.8: 13567912-13567912 on Build GRCh38
rs554147664 Chr.8: 13567812-13567812 on Build GRCh38
rs905211465 Chr.8: 13568085-13568085 on Build GRCh38
rs75464306 Chr.8: 13568082-13568082 on Build GRCh38
rs778874736 Chr.8: 13567838-13567838 on Build GRCh38
rs749543123 Chr.8: 13568041-13568041 on Build GRCh38
rs536897309 Chr.8: 13568061-13568061 on Build GRCh38
rs185450144 Chr.8: 13567928-13567928 on Build GRCh38
rs754546798 Chr.8: 13567810-13567810 on Build GRCh38
rs887927420 Chr.8: 13567856-13567856 on Build GRCh38
rs981845779 Chr.8: 13568071-13568071 on Build GRCh38
rs149798700 Chr.8: 13567758-13567758 on Build GRCh38
rs774481502 Chr.8: 13567907-13567907 on Build GRCh38
rs764683239 Chr.8: 13568049-13568049 on Build GRCh38
rs1014242184 Chr.8: 13568072-13568072 on Build GRCh38
rs964731705 Chr.8: 13567955-13567955 on Build GRCh38
rs190827878 Chr.8: 13567908-13567908 on Build GRCh38
rs747821007 Chr.8: 13567864-13567864 on Build GRCh38
rs756304478 Chr.8: 13567939-13567939 on Build GRCh38
rs745996247 Chr.8: 13567920-13567920 on Build GRCh38
rs1011037834 Chr.8: 13567894-13567894 on Build GRCh38
rs779936142 Chr.8: 13567999-13567999 on Build GRCh38
rs939680236 Chr.8: 13567800-13567800 on Build GRCh38
rs956633330 Chr.8: 13567767-13567767 on Build GRCh38
rs1000845574 Chr.8: 13568086-13568086 on Build GRCh38

View Full Product Details