Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136845075-136845657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf172
Entrez Gene ID
389813
Gene Name
chromosome 9 open reading frame 172
Gene Aliases
-
UniGene
Hs.495485
Gene Chromosome Location
Chr.9: 136844415-136847345 on Build GRCh38
Transcripts
NM_001080482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3905854 c.886G>A p.A296T Chr.9: 136845300-136845300 on Build GRCh38
7293721 c.909C>T p.D303D Chr.9: 136845323-136845323 on Build GRCh38
6248015 c.873G>T p.R291R Chr.9: 136845287-136845287 on Build GRCh38
4998250 c.1075C>T p.R359C Chr.9: 136845489-136845489 on Build GRCh38
1247013 c.716C>G p.P239R Chr.9: 136845130-136845130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770970200 Chr.9: 136845588-136845588 on Build GRCh38
rs773101030 Chr.9: 136845566-136845566 on Build GRCh38
rs755149242 Chr.9: 136845362-136845362 on Build GRCh38
rs755684073 Chr.9: 136845177-136845177 on Build GRCh38
rs752437179 Chr.9: 136845293-136845293 on Build GRCh38
rs751060368 Chr.9: 136845102-136845102 on Build GRCh38
rs776897694 Chr.9: 136845158-136845158 on Build GRCh38
rs773156184 Chr.9: 136845515-136845515 on Build GRCh38
rs771347157 Chr.9: 136845341-136845341 on Build GRCh38
rs745910968 Chr.9: 136845247-136845247 on Build GRCh38
rs755097987 Chr.9: 136845422-136845422 on Build GRCh38
rs764134799 Chr.9: 136845619-136845619 on Build GRCh38
rs758258222 Chr.9: 136845297-136845297 on Build GRCh38
rs760284089 Chr.9: 136845221-136845221 on Build GRCh38
rs530989554 Chr.9: 136845092-136845092 on Build GRCh38
rs763688079 Chr.9: 136845389-136845389 on Build GRCh38
rs748775805 Chr.9: 136845481-136845481 on Build GRCh38
rs756566872 Chr.9: 136845551-136845551 on Build GRCh38
rs768533933 Chr.9: 136845128-136845128 on Build GRCh38
rs747059901 Chr.9: 136845135-136845135 on Build GRCh38
rs1033950941 Chr.9: 136845165-136845165 on Build GRCh38
rs774820563 Chr.9: 136845272-136845272 on Build GRCh38
rs768336016 Chr.9: 136845482-136845482 on Build GRCh38
rs565642539 Chr.9: 136845308-136845308 on Build GRCh38
rs749140080 Chr.9: 136845124-136845124 on Build GRCh38
rs752773831 Chr.9: 136845528-136845528 on Build GRCh38
rs767752100 Chr.9: 136845638-136845638 on Build GRCh38
rs759065608 Chr.9: 136845346-136845346 on Build GRCh38
rs911393490 Chr.9: 136845421-136845421 on Build GRCh38
rs761664074 Chr.9: 136845500-136845500 on Build GRCh38
rs746429035 Chr.9: 136845461-136845461 on Build GRCh38
rs752492393 Chr.9: 136845236-136845236 on Build GRCh38
rs765917776 Chr.9: 136845224-136845224 on Build GRCh38
rs769953917 Chr.9: 136845254-136845254 on Build GRCh38
rs755806557 Chr.9: 136845119-136845119 on Build GRCh38
rs764899714 Chr.9: 136845287-136845287 on Build GRCh38
rs769357773 Chr.9: 136845379-136845379 on Build GRCh38
rs759122525 Chr.9: 136845286-136845286 on Build GRCh38
rs191760762 Chr.9: 136845394-136845394 on Build GRCh38
rs751791871 Chr.9: 136845542-136845542 on Build GRCh38
rs747373398 Chr.9: 136845439-136845439 on Build GRCh38
rs774321155 Chr.9: 136845134-136845134 on Build GRCh38
rs753357668 Chr.9: 136845285-136845285 on Build GRCh38
rs767080219 Chr.9: 136845174-136845174 on Build GRCh38
rs959298819 Chr.9: 136845167-136845167 on Build GRCh38
rs772051769 Chr.9: 136845501-136845501 on Build GRCh38
rs757602551 Chr.9: 136845546-136845546 on Build GRCh38
rs547443739 Chr.9: 136845185-136845185 on Build GRCh38
rs556778623 Chr.9: 136845402-136845402 on Build GRCh38
rs750821040 Chr.9: 136845550-136845550 on Build GRCh38
rs774873860 Chr.9: 136845216-136845216 on Build GRCh38
rs10870132 Chr.9: 136845191-136845191 on Build GRCh38
rs758604679 Chr.9: 136845433-136845433 on Build GRCh38
rs1047400352 Chr.9: 136845290-136845290 on Build GRCh38
rs780116796 Chr.9: 136845311-136845311 on Build GRCh38
rs749956494 Chr.9: 136845176-136845176 on Build GRCh38
rs536849229 Chr.9: 136845273-136845273 on Build GRCh38
rs763374907 Chr.9: 136845213-136845213 on Build GRCh38
rs200347553 Chr.9: 136845324-136845324 on Build GRCh38
rs767531845 Chr.9: 136845411-136845411 on Build GRCh38
rs756873636 Chr.9: 136845105-136845105 on Build GRCh38
rs771857283 Chr.9: 136845564-136845564 on Build GRCh38
rs757182652 Chr.9: 136845354-136845354 on Build GRCh38
rs765498387 Chr.9: 136845160-136845160 on Build GRCh38
rs778816389 Chr.9: 136845187-136845187 on Build GRCh38
rs770292381 Chr.9: 136845473-136845473 on Build GRCh38
rs778619824 Chr.9: 136845234-136845234 on Build GRCh38
rs777879549 Chr.9: 136845437-136845437 on Build GRCh38
rs768103235 Chr.9: 136845220-136845220 on Build GRCh38
rs752182340 Chr.9: 136845166-136845166 on Build GRCh38
rs760566982 Chr.9: 136845579-136845579 on Build GRCh38
rs779650178 Chr.9: 136845122-136845122 on Build GRCh38
rs779780463 Chr.9: 136845179-136845179 on Build GRCh38
rs775674999 Chr.9: 136845162-136845162 on Build GRCh38
rs377124253 Chr.9: 136845366-136845366 on Build GRCh38
rs762603558 Chr.9: 136845386-136845386 on Build GRCh38
rs771419371 Chr.9: 136845373-136845373 on Build GRCh38
rs749015039 Chr.9: 136845180-136845180 on Build GRCh38
rs954854261 Chr.9: 136845107-136845118 on Build GRCh38
rs773811813 Chr.9: 136845275-136845275 on Build GRCh38
rs552865442 Chr.9: 136845553-136845553 on Build GRCh38
rs760621878 Chr.9: 136845519-136845519 on Build GRCh38
rs753945927 Chr.9: 136845361-136845361 on Build GRCh38
rs771064957 Chr.9: 136845154-136845154 on Build GRCh38
rs775825952 Chr.9: 136845212-136845212 on Build GRCh38
rs781660782 Chr.9: 136845452-136845452 on Build GRCh38
rs754754523 Chr.9: 136845231-136845231 on Build GRCh38
rs569890181 Chr.9: 136845347-136845347 on Build GRCh38
rs372798221 Chr.9: 136845335-136845335 on Build GRCh38
rs765971426 Chr.9: 136845279-136845279 on Build GRCh38
rs777786024 Chr.9: 136845245-136845245 on Build GRCh38
rs773915795 Chr.9: 136845483-136845483 on Build GRCh38
rs573595505 Chr.9: 136845420-136845420 on Build GRCh38
rs936751983 Chr.9: 136845240-136845240 on Build GRCh38
rs745346813 Chr.9: 136845377-136845377 on Build GRCh38
rs762345829 Chr.9: 136845217-136845217 on Build GRCh38
rs759617108 Chr.9: 136845159-136845159 on Build GRCh38
rs551188501 Chr.9: 136845146-136845146 on Build GRCh38
rs757653827 Chr.9: 136845451-136845451 on Build GRCh38
rs778130595 Chr.9: 136845369-136845369 on Build GRCh38
rs769856493 Chr.9: 136845209-136845209 on Build GRCh38
rs56267514 Chr.9: 136845246-136845246 on Build GRCh38
rs571349112 Chr.9: 136845368-136845368 on Build GRCh38
rs747363781 Chr.9: 136845370-136845370 on Build GRCh38
rs773167137 Chr.9: 136845210-136845210 on Build GRCh38
rs774943768 Chr.9: 136845476-136845476 on Build GRCh38
rs201654604 Chr.9: 136845558-136845558 on Build GRCh38
rs370877404 Chr.9: 136845525-136845525 on Build GRCh38
rs761162532 Chr.9: 136845498-136845498 on Build GRCh38
rs767589006 Chr.9: 136845357-136845357 on Build GRCh38
rs745988535 Chr.9: 136845207-136845207 on Build GRCh38
rs756818565 Chr.9: 136845173-136845173 on Build GRCh38
rs774868497 Chr.9: 136845443-136845445 on Build GRCh38
rs752722561 Chr.9: 136845432-136845432 on Build GRCh38
rs542467230 Chr.9: 136845549-136845549 on Build GRCh38
rs761986244 Chr.9: 136845632-136845632 on Build GRCh38
rs557541546 Chr.9: 136845353-136845353 on Build GRCh38
rs776469636 Chr.9: 136845589-136845589 on Build GRCh38
rs199919460 Chr.9: 136845327-136845328 on Build GRCh38
rs866435517 Chr.9: 136845310-136845310 on Build GRCh38
rs762991487 Chr.9: 136845532-136845532 on Build GRCh38
rs753594723 Chr.9: 136845225-136845225 on Build GRCh38
rs758289193 Chr.9: 136845244-136845244 on Build GRCh38
rs764615747 Chr.9: 136845214-136845214 on Build GRCh38
rs764342617 Chr.9: 136845533-136845533 on Build GRCh38
rs530462305 Chr.9: 136845260-136845260 on Build GRCh38
rs747747733 Chr.9: 136845561-136845561 on Build GRCh38
rs749415108 Chr.9: 136845317-136845317 on Build GRCh38
rs751670859 Chr.9: 136845629-136845629 on Build GRCh38
rs763080457 Chr.9: 136845610-136845610 on Build GRCh38
rs780990690 Chr.9: 136845117-136845117 on Build GRCh38
rs748141801 Chr.9: 136845190-136845190 on Build GRCh38
rs749466349 Chr.9: 136845269-136845269 on Build GRCh38
rs930248957 Chr.9: 136845518-136845518 on Build GRCh38
rs768954361 Chr.9: 136845320-136845320 on Build GRCh38
rs764726447 Chr.9: 136845093-136845093 on Build GRCh38
rs777538201 Chr.9: 136845304-136845304 on Build GRCh38
rs778614881 Chr.9: 136845560-136845560 on Build GRCh38
rs941778698 Chr.9: 136845557-136845557 on Build GRCh38
rs776078721 Chr.9: 136845345-136845345 on Build GRCh38
rs775107430 Chr.9: 136845383-136845383 on Build GRCh38
rs748441211 Chr.9: 136845365-136845365 on Build GRCh38
rs748803802 Chr.9: 136845555-136845555 on Build GRCh38
rs925322578 Chr.9: 136845227-136845227 on Build GRCh38
rs764673024 Chr.9: 136845164-136845164 on Build GRCh38
rs773580400 Chr.9: 136845338-136845338 on Build GRCh38

View Full Product Details