Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36564336-36564868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG2
Entrez Gene ID
10369
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 2
Gene Aliases
MRD10
UniGene
Hs.146766
Gene Chromosome Location
Chr.22: 36560869-36702645 on Build GRCh38
Transcripts
NM_006078.3 XM_017028531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1130440 c.606C>T p.I202I Chr.22: 36564717-36564717 on Build GRCh38
1199387 c.590C>T p.A197V Chr.22: 36564733-36564733 on Build GRCh38
3553980 c.888C>T p.F296F Chr.22: 36564435-36564435 on Build GRCh38
6618694 c.567C>T p.I189I Chr.22: 36564756-36564756 on Build GRCh38
4103845 c.627G>A p.R209R Chr.22: 36564696-36564696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755200898 Chr.22: 36564629-36564629 on Build GRCh38
rs144838444 Chr.22: 36564831-36564831 on Build GRCh38
rs751747375 Chr.22: 36564774-36564774 on Build GRCh38
rs768099799 Chr.22: 36564685-36564685 on Build GRCh38
rs748221864 Chr.22: 36564525-36564525 on Build GRCh38
rs113927105 Chr.22: 36564829-36564829 on Build GRCh38
rs776372258 Chr.22: 36564382-36564382 on Build GRCh38
rs113130432 Chr.22: 36564777-36564777 on Build GRCh38
rs756431865 Chr.22: 36564476-36564476 on Build GRCh38
rs368237033 Chr.22: 36564495-36564495 on Build GRCh38
rs745850264 Chr.22: 36564792-36564792 on Build GRCh38
rs748055470 Chr.22: 36564355-36564355 on Build GRCh38
rs779033735 Chr.22: 36564507-36564507 on Build GRCh38
rs1023763061 Chr.22: 36564650-36564650 on Build GRCh38
rs868300937 Chr.22: 36564673-36564673 on Build GRCh38
rs141608078 Chr.22: 36564564-36564564 on Build GRCh38
rs866575214 Chr.22: 36564733-36564733 on Build GRCh38
rs201847633 Chr.22: 36564368-36564368 on Build GRCh38
rs774312249 Chr.22: 36564354-36564354 on Build GRCh38
rs139052540 Chr.22: 36564683-36564683 on Build GRCh38
rs146183978 Chr.22: 36564665-36564665 on Build GRCh38
rs753303377 Chr.22: 36564839-36564839 on Build GRCh38
rs71314996 Chr.22: 36564481-36564481 on Build GRCh38
rs759775990 Chr.22: 36564697-36564697 on Build GRCh38
rs756630130 Chr.22: 36564591-36564591 on Build GRCh38
rs989827080 Chr.22: 36564725-36564725 on Build GRCh38
rs761169033 Chr.22: 36564686-36564686 on Build GRCh38
rs377137598 Chr.22: 36564467-36564467 on Build GRCh38
rs559126345 Chr.22: 36564691-36564691 on Build GRCh38
rs547867640 Chr.22: 36564408-36564408 on Build GRCh38
rs750884147 Chr.22: 36564587-36564587 on Build GRCh38
rs111500782 Chr.22: 36564736-36564736 on Build GRCh38
rs773842942 Chr.22: 36564830-36564830 on Build GRCh38
rs748431364 Chr.22: 36564636-36564636 on Build GRCh38
rs762691427 Chr.22: 36564684-36564684 on Build GRCh38
rs574213269 Chr.22: 36564622-36564622 on Build GRCh38
rs779039681 Chr.22: 36564634-36564634 on Build GRCh38
rs763627504 Chr.22: 36564571-36564571 on Build GRCh38
rs752215474 Chr.22: 36564409-36564409 on Build GRCh38
rs759983832 Chr.22: 36564508-36564508 on Build GRCh38
rs750757628 Chr.22: 36564472-36564472 on Build GRCh38
rs781032119 Chr.22: 36564788-36564788 on Build GRCh38
rs111933816 Chr.22: 36564820-36564820 on Build GRCh38
rs764857508 Chr.22: 36564560-36564560 on Build GRCh38
rs923823706 Chr.22: 36564689-36564689 on Build GRCh38
rs766305433 Chr.22: 36564381-36564381 on Build GRCh38
rs773240271 Chr.22: 36564534-36564534 on Build GRCh38
rs771958374 Chr.22: 36564371-36564371 on Build GRCh38
rs149442040 Chr.22: 36564744-36564744 on Build GRCh38
rs559004347 Chr.22: 36564362-36564362 on Build GRCh38
rs758357916 Chr.22: 36564654-36564654 on Build GRCh38
rs952887410 Chr.22: 36564582-36564582 on Build GRCh38
rs376065197 Chr.22: 36564573-36564573 on Build GRCh38
rs944342384 Chr.22: 36564840-36564840 on Build GRCh38
rs867189579 Chr.22: 36564674-36564674 on Build GRCh38
rs1019562662 Chr.22: 36564756-36564756 on Build GRCh38
rs748829000 Chr.22: 36564817-36564817 on Build GRCh38
rs372295505 Chr.22: 36564606-36564606 on Build GRCh38
rs775042681 Chr.22: 36564561-36564561 on Build GRCh38
rs190739501 Chr.22: 36564711-36564711 on Build GRCh38
rs931411438 Chr.22: 36564688-36564688 on Build GRCh38
rs200227696 Chr.22: 36564542-36564542 on Build GRCh38
rs759221759 Chr.22: 36564556-36564556 on Build GRCh38
rs757960446 Chr.22: 36564460-36564460 on Build GRCh38
rs777995011 Chr.22: 36564765-36564765 on Build GRCh38
rs779915321 Chr.22: 36564814-36564814 on Build GRCh38
rs113704375 Chr.22: 36564597-36564597 on Build GRCh38
rs776572206 Chr.22: 36564554-36564554 on Build GRCh38
rs915061917 Chr.22: 36564734-36564734 on Build GRCh38
rs899662674 Chr.22: 36564816-36564816 on Build GRCh38
rs759121913 Chr.22: 36564395-36564395 on Build GRCh38
rs567997419 Chr.22: 36564535-36564535 on Build GRCh38
rs201512121 Chr.22: 36564678-36564678 on Build GRCh38
rs1065089 Chr.22: 36564576-36564576 on Build GRCh38
rs752802266 Chr.22: 36564716-36564716 on Build GRCh38
rs754196562 Chr.22: 36564694-36564694 on Build GRCh38
rs1021475125 Chr.22: 36564841-36564841 on Build GRCh38
rs887134997 Chr.22: 36564706-36564706 on Build GRCh38
rs755164929 Chr.22: 36564499-36564499 on Build GRCh38
rs769458037 Chr.22: 36564681-36564681 on Build GRCh38
rs911651890 Chr.22: 36564825-36564825 on Build GRCh38
rs535062605 Chr.22: 36564798-36564798 on Build GRCh38
rs768367873 Chr.22: 36564822-36564822 on Build GRCh38
rs970675754 Chr.22: 36564663-36564663 on Build GRCh38
rs200149890 Chr.22: 36564372-36564372 on Build GRCh38
rs781121207 Chr.22: 36564672-36564672 on Build GRCh38
rs780421828 Chr.22: 36564482-36564482 on Build GRCh38

View Full Product Details