Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179709698-179710105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CANX
Entrez Gene ID
821
Gene Name
calnexin
Gene Aliases
CNX, IP90, P90
UniGene
Hs.567968
Gene Chromosome Location
Chr.5: 179678631-179731641 on Build GRCh38
Transcripts
NM_001024649.1 NM_001746.3 XM_011534664.1 XM_011534665.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6199545 c.634-4G>T p.? Chr.5: 179709869-179709869 on Build GRCh38
3011753 c.588G>A p.E196E Chr.5: 179709932-179709932 on Build GRCh38
1436542 c.657T>C p.A219A Chr.5: 179710001-179710001 on Build GRCh38
4347353 c.693G>A p.E231E Chr.5: 179709932-179709932 on Build GRCh38
5131041 c.762T>C p.A254A Chr.5: 179710001-179710001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019613285 Chr.5: 179709971-179709971 on Build GRCh38
rs768789509 Chr.5: 179709893-179709893 on Build GRCh38
rs767177938 Chr.5: 179709956-179709956 on Build GRCh38
rs772088184 Chr.5: 179710081-179710081 on Build GRCh38
rs780392943 Chr.5: 179709967-179709967 on Build GRCh38
rs752466224 Chr.5: 179709847-179709847 on Build GRCh38
rs994951211 Chr.5: 179710049-179710049 on Build GRCh38
rs759546139 Chr.5: 179709947-179709947 on Build GRCh38
rs919650527 Chr.5: 179709773-179709773 on Build GRCh38
rs748824424 Chr.5: 179709815-179709815 on Build GRCh38
rs780218950 Chr.5: 179710070-179710071 on Build GRCh38
rs769909544 Chr.5: 179709985-179709985 on Build GRCh38
rs762744899 Chr.5: 179709937-179709937 on Build GRCh38
rs747501873 Chr.5: 179709890-179709890 on Build GRCh38
rs770921171 Chr.5: 179710027-179710027 on Build GRCh38
rs777997443 Chr.5: 179710011-179710011 on Build GRCh38
rs1040946377 Chr.5: 179709898-179709898 on Build GRCh38
rs781213469 Chr.5: 179709899-179709899 on Build GRCh38
rs750752034 Chr.5: 179709863-179709863 on Build GRCh38
rs207466646 Chr.5: 179710017-179710017 on Build GRCh38
rs371318629 Chr.5: 179710043-179710043 on Build GRCh38
rs1031621798 Chr.5: 179709926-179709926 on Build GRCh38
rs931460308 Chr.5: 179709772-179709772 on Build GRCh38
rs767126942 Chr.5: 179709838-179709838 on Build GRCh38
rs908787759 Chr.5: 179709797-179709797 on Build GRCh38
rs141861672 Chr.5: 179709963-179709963 on Build GRCh38
rs965487104 Chr.5: 179709726-179709726 on Build GRCh38
rs34000275 Chr.5: 179709764-179709764 on Build GRCh38
rs779197154 Chr.5: 179709855-179709855 on Build GRCh38
rs374732152 Chr.5: 179710061-179710061 on Build GRCh38
rs749461583 Chr.5: 179710020-179710020 on Build GRCh38
rs112133623 Chr.5: 179709995-179709995 on Build GRCh38
rs748363222 Chr.5: 179709901-179709901 on Build GRCh38
rs760269082 Chr.5: 179709959-179709959 on Build GRCh38
rs202146081 Chr.5: 179710076-179710076 on Build GRCh38
rs150270438 Chr.5: 179709964-179709964 on Build GRCh38
rs1048652058 Chr.5: 179709794-179709794 on Build GRCh38
rs752230231 Chr.5: 179709957-179709957 on Build GRCh38
rs771008892 Chr.5: 179709939-179709939 on Build GRCh38
rs948199686 Chr.5: 179709978-179709978 on Build GRCh38
rs973555879 Chr.5: 179709766-179709766 on Build GRCh38
rs1056798038 Chr.5: 179709805-179709805 on Build GRCh38
rs756183761 Chr.5: 179709847-179709847 on Build GRCh38
rs780479093 Chr.5: 179709883-179709883 on Build GRCh38
rs781268361 Chr.5: 179709979-179709979 on Build GRCh38
rs921332226 Chr.5: 179709983-179709983 on Build GRCh38
rs752100105 Chr.5: 179709972-179709972 on Build GRCh38
rs754482022 Chr.5: 179709828-179709828 on Build GRCh38
rs1045472291 Chr.5: 179709980-179709980 on Build GRCh38
rs904374755 Chr.5: 179709887-179709887 on Build GRCh38
rs758832045 Chr.5: 179709881-179709881 on Build GRCh38
rs952520930 Chr.5: 179709785-179709785 on Build GRCh38
rs1044139583 Chr.5: 179709878-179709878 on Build GRCh38
rs575286152 Chr.5: 179709788-179709788 on Build GRCh38
rs192745904 Chr.5: 179709742-179709742 on Build GRCh38
rs755452646 Chr.5: 179709976-179709976 on Build GRCh38
rs142118147 Chr.5: 179709962-179709962 on Build GRCh38
rs772909275 Chr.5: 179709935-179709935 on Build GRCh38
rs768208195 Chr.5: 179709902-179709902 on Build GRCh38
rs1015102398 Chr.5: 179710015-179710015 on Build GRCh38
rs906496131 Chr.5: 179709986-179709986 on Build GRCh38
rs370218228 Chr.5: 179709849-179709849 on Build GRCh38
rs1003507982 Chr.5: 179709997-179709997 on Build GRCh38
rs985522027 Chr.5: 179709798-179709798 on Build GRCh38
rs542607097 Chr.5: 179709941-179709941 on Build GRCh38
rs755761297 Chr.5: 179709852-179709852 on Build GRCh38

View Full Product Details