Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10871355-10871934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARM1
Entrez Gene ID
10498
Gene Name
coactivator associated arginine methyltransferase 1
Gene Aliases
PRMT4
UniGene
Hs.323213
Gene Chromosome Location
Chr.19: 10871577-10923078 on Build GRCh38
Transcripts
NM_199141.1 XM_005259708.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388557 c.216C>G p.Y72* Chr.19: 10871918-10871918 on Build GRCh38
4131181 c.44C>T p.A15V Chr.19: 10871746-10871746 on Build GRCh38
5507673 c.86T>A p.V29E Chr.19: 10871788-10871788 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772075751 Chr.19: 10871831-10871831 on Build GRCh38
rs986911887 Chr.19: 10871601-10871601 on Build GRCh38
rs1002629347 Chr.19: 10871557-10871557 on Build GRCh38
rs956050539 Chr.19: 10871414-10871414 on Build GRCh38
rs1046965708 Chr.19: 10871751-10871751 on Build GRCh38
rs550869012 Chr.19: 10871708-10871709 on Build GRCh38
rs905677821 Chr.19: 10871397-10871397 on Build GRCh38
rs945196544 Chr.19: 10871584-10871607 on Build GRCh38
rs1044517097 Chr.19: 10871497-10871505 on Build GRCh38
rs919736893 Chr.19: 10871643-10871643 on Build GRCh38
rs763578324 Chr.19: 10871847-10871847 on Build GRCh38
rs938539306 Chr.19: 10871905-10871905 on Build GRCh38
rs868590477 Chr.19: 10871595-10871595 on Build GRCh38
rs913704741 Chr.19: 10871389-10871412 on Build GRCh38
rs942436996 Chr.19: 10871622-10871622 on Build GRCh38
rs865852802 Chr.19: 10871604-10871604 on Build GRCh38
rs866610548 Chr.19: 10871589-10871589 on Build GRCh38
rs967580227 Chr.19: 10871588-10871588 on Build GRCh38
rs550960247 Chr.19: 10871445-10871445 on Build GRCh38
rs375588563 Chr.19: 10871419-10871419 on Build GRCh38
rs112438892 Chr.19: 10871514-10871514 on Build GRCh38
rs1036675342 Chr.19: 10871491-10871491 on Build GRCh38
rs975219811 Chr.19: 10871434-10871434 on Build GRCh38
rs954866098 Chr.19: 10871795-10871795 on Build GRCh38
rs908090147 Chr.19: 10871479-10871479 on Build GRCh38
rs759354134 Chr.19: 10871816-10871816 on Build GRCh38
rs761503923 Chr.19: 10871886-10871886 on Build GRCh38
rs898250093 Chr.19: 10871493-10871493 on Build GRCh38
rs1022148168 Chr.19: 10871424-10871424 on Build GRCh38
rs764822141 Chr.19: 10871899-10871899 on Build GRCh38
rs865942557 Chr.19: 10871613-10871613 on Build GRCh38
rs1002766631 Chr.19: 10871407-10871408 on Build GRCh38
rs955061306 Chr.19: 10871598-10871598 on Build GRCh38
rs529298775 Chr.19: 10871426-10871426 on Build GRCh38
rs760245466 Chr.19: 10871843-10871843 on Build GRCh38
rs753316553 Chr.19: 10871866-10871866 on Build GRCh38
rs931106388 Chr.19: 10871496-10871497 on Build GRCh38
rs774334252 Chr.19: 10871804-10871804 on Build GRCh38
rs867174988 Chr.19: 10871625-10871625 on Build GRCh38
rs1021656862 Chr.19: 10871659-10871659 on Build GRCh38
rs867798542 Chr.19: 10871616-10871616 on Build GRCh38
rs867757627 Chr.19: 10871637-10871637 on Build GRCh38
rs763660570 Chr.19: 10871737-10871760 on Build GRCh38
rs933473416 Chr.19: 10871468-10871468 on Build GRCh38
rs987756515 Chr.19: 10871476-10871476 on Build GRCh38
rs988735581 Chr.19: 10871417-10871417 on Build GRCh38
rs969106951 Chr.19: 10871685-10871685 on Build GRCh38
rs775222924 Chr.19: 10871837-10871837 on Build GRCh38
rs868511606 Chr.19: 10871619-10871619 on Build GRCh38
rs1010668126 Chr.19: 10871654-10871654 on Build GRCh38
rs868171964 Chr.19: 10871607-10871607 on Build GRCh38
rs539895704 Chr.19: 10871725-10871725 on Build GRCh38
rs1029483892 Chr.19: 10871752-10871752 on Build GRCh38
rs113975921 Chr.19: 10871662-10871662 on Build GRCh38
rs1022766461 Chr.19: 10871390-10871390 on Build GRCh38
rs987598124 Chr.19: 10871800-10871800 on Build GRCh38
rs930968688 Chr.19: 10871873-10871873 on Build GRCh38
rs868352816 Chr.19: 10871638-10871638 on Build GRCh38
rs939670161 Chr.19: 10871482-10871482 on Build GRCh38
rs1035566448 Chr.19: 10871413-10871413 on Build GRCh38
rs866382016 Chr.19: 10871583-10871583 on Build GRCh38
rs868641144 Chr.19: 10871610-10871610 on Build GRCh38
rs1044184881 Chr.19: 10871887-10871887 on Build GRCh38
rs891692329 Chr.19: 10871647-10871647 on Build GRCh38
rs974893827 Chr.19: 10871738-10871738 on Build GRCh38
rs867969940 Chr.19: 10871862-10871862 on Build GRCh38
rs866968375 Chr.19: 10871640-10871640 on Build GRCh38

View Full Product Details