Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10871684-10872239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARM1
Entrez Gene ID
10498
Gene Name
coactivator associated arginine methyltransferase 1
Gene Aliases
PRMT4
UniGene
Hs.323213
Gene Chromosome Location
Chr.19: 10871577-10923078 on Build GRCh38
Transcripts
NM_199141.1 XM_005259708.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388557 c.216C>G p.Y72* Chr.19: 10871918-10871918 on Build GRCh38
4131181 c.44C>T p.A15V Chr.19: 10871746-10871746 on Build GRCh38
5507673 c.86T>A p.V29E Chr.19: 10871788-10871788 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772075751 Chr.19: 10871831-10871831 on Build GRCh38
rs927059934 Chr.19: 10872087-10872090 on Build GRCh38
rs1056865609 Chr.19: 10871931-10871931 on Build GRCh38
rs556495359 Chr.19: 10872130-10872130 on Build GRCh38
rs368657529 Chr.19: 10872097-10872097 on Build GRCh38
rs574649852 Chr.19: 10872105-10872105 on Build GRCh38
rs1046965708 Chr.19: 10871751-10871751 on Build GRCh38
rs891619017 Chr.19: 10871948-10871948 on Build GRCh38
rs563827725 Chr.19: 10872213-10872213 on Build GRCh38
rs919028369 Chr.19: 10872126-10872126 on Build GRCh38
rs913106246 Chr.19: 10872141-10872141 on Build GRCh38
rs550869012 Chr.19: 10871708-10871709 on Build GRCh38
rs907152688 Chr.19: 10871998-10871998 on Build GRCh38
rs763578324 Chr.19: 10871847-10871847 on Build GRCh38
rs374848671 Chr.19: 10872154-10872154 on Build GRCh38
rs996546704 Chr.19: 10872010-10872010 on Build GRCh38
rs938539306 Chr.19: 10871905-10871905 on Build GRCh38
rs573390096 Chr.19: 10872214-10872214 on Build GRCh38
rs1021436889 Chr.19: 10872017-10872017 on Build GRCh38
rs1029085179 Chr.19: 10872012-10872012 on Build GRCh38
rs545539245 Chr.19: 10872179-10872179 on Build GRCh38
rs1028474173 Chr.19: 10872054-10872054 on Build GRCh38
rs551902930 Chr.19: 10872016-10872016 on Build GRCh38
rs1010178253 Chr.19: 10871967-10871967 on Build GRCh38
rs954866098 Chr.19: 10871795-10871795 on Build GRCh38
rs1015129732 Chr.19: 10872031-10872031 on Build GRCh38
rs574996071 Chr.19: 10872151-10872151 on Build GRCh38
rs965588070 Chr.19: 10872034-10872034 on Build GRCh38
rs962185065 Chr.19: 10872043-10872043 on Build GRCh38
rs759354134 Chr.19: 10871816-10871816 on Build GRCh38
rs908880545 Chr.19: 10872102-10872102 on Build GRCh38
rs761503923 Chr.19: 10871886-10871886 on Build GRCh38
rs904398502 Chr.19: 10872180-10872180 on Build GRCh38
rs764822141 Chr.19: 10871899-10871899 on Build GRCh38
rs1053595316 Chr.19: 10872211-10872211 on Build GRCh38
rs760245466 Chr.19: 10871843-10871843 on Build GRCh38
rs753316553 Chr.19: 10871866-10871866 on Build GRCh38
rs774334252 Chr.19: 10871804-10871804 on Build GRCh38
rs949144142 Chr.19: 10872135-10872135 on Build GRCh38
rs954482484 Chr.19: 10872055-10872055 on Build GRCh38
rs937940415 Chr.19: 10872209-10872209 on Build GRCh38
rs111430161 Chr.19: 10871950-10871950 on Build GRCh38
rs926470941 Chr.19: 10872165-10872165 on Build GRCh38
rs763660570 Chr.19: 10871737-10871760 on Build GRCh38
rs535117761 Chr.19: 10872125-10872125 on Build GRCh38
rs775222924 Chr.19: 10871837-10871837 on Build GRCh38
rs570702117 Chr.19: 10872019-10872019 on Build GRCh38
rs539895704 Chr.19: 10871725-10871725 on Build GRCh38
rs534802737 Chr.19: 10872050-10872050 on Build GRCh38
rs1029483892 Chr.19: 10871752-10871752 on Build GRCh38
rs987598124 Chr.19: 10871800-10871800 on Build GRCh38
rs930968688 Chr.19: 10871873-10871873 on Build GRCh38
rs1043364527 Chr.19: 10871976-10871976 on Build GRCh38
rs996209103 Chr.19: 10872185-10872185 on Build GRCh38
rs890448524 Chr.19: 10872216-10872216 on Build GRCh38
rs920835219 Chr.19: 10872067-10872067 on Build GRCh38
rs1044184881 Chr.19: 10871887-10871887 on Build GRCh38
rs974893827 Chr.19: 10871738-10871738 on Build GRCh38
rs867969940 Chr.19: 10871862-10871862 on Build GRCh38
rs984337188 Chr.19: 10872095-10872095 on Build GRCh38
rs971818256 Chr.19: 10872106-10872106 on Build GRCh38
rs553023549 Chr.19: 10872086-10872086 on Build GRCh38
rs952416862 Chr.19: 10872070-10872070 on Build GRCh38
rs893830094 Chr.19: 10872212-10872212 on Build GRCh38

View Full Product Details