Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45621169-45621662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC17
Entrez Gene ID
149483
Gene Name
coiled-coil domain containing 17
Gene Aliases
-
UniGene
Hs.18912
Gene Chromosome Location
Chr.1: 45620044-45624787 on Build GRCh38
Transcripts
NM_001114938.2 NM_001190182.1 XM_011540812.1 XM_011540814.1 XM_011540820.1 XM_017000450.1 XM_017000451.1 XM_017000452.1 XM_017000453.1 XM_017000454.1 XM_017000455.1 XM_017000456.1 XM_017000457.1 XM_017000458.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6621810 c.1202C>T p.A401V Chr.1: 45621371-45621371 on Build GRCh38
1343068 c.1120C>T p.R374W Chr.1: 45621453-45621453 on Build GRCh38
4438931 c.1176T>C p.D392D Chr.1: 45621397-45621397 on Build GRCh38
4443189 c.1181G>A p.R394Q Chr.1: 45621392-45621392 on Build GRCh38
5992260 c.1171C>T p.R391C Chr.1: 45621402-45621402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751795710 Chr.1: 45621364-45621364 on Build GRCh38
rs754996329 Chr.1: 45621502-45621502 on Build GRCh38
rs1028892723 Chr.1: 45621532-45621532 on Build GRCh38
rs758716731 Chr.1: 45621261-45621261 on Build GRCh38
rs756236077 Chr.1: 45621407-45621417 on Build GRCh38
rs756073933 Chr.1: 45621392-45621392 on Build GRCh38
rs373781070 Chr.1: 45621618-45621618 on Build GRCh38
rs759025620 Chr.1: 45621319-45621319 on Build GRCh38
rs759824331 Chr.1: 45621463-45621463 on Build GRCh38
rs750724528 Chr.1: 45621257-45621257 on Build GRCh38
rs779787845 Chr.1: 45621591-45621591 on Build GRCh38
rs911720248 Chr.1: 45621280-45621280 on Build GRCh38
rs1053736618 Chr.1: 45621378-45621378 on Build GRCh38
rs748076298 Chr.1: 45621298-45621298 on Build GRCh38
rs752774794 Chr.1: 45621486-45621486 on Build GRCh38
rs978767086 Chr.1: 45621569-45621569 on Build GRCh38
rs896091694 Chr.1: 45621197-45621197 on Build GRCh38
rs773925529 Chr.1: 45621610-45621610 on Build GRCh38
rs914608420 Chr.1: 45621217-45621217 on Build GRCh38
rs967927214 Chr.1: 45621568-45621568 on Build GRCh38
rs946026422 Chr.1: 45621225-45621225 on Build GRCh38
rs777316322 Chr.1: 45621324-45621324 on Build GRCh38
rs1029970785 Chr.1: 45621343-45621343 on Build GRCh38
rs751771651 Chr.1: 45621456-45621456 on Build GRCh38
rs764126888 Chr.1: 45621628-45621628 on Build GRCh38
rs377681851 Chr.1: 45621273-45621273 on Build GRCh38
rs907267163 Chr.1: 45621260-45621260 on Build GRCh38
rs570621990 Chr.1: 45621462-45621462 on Build GRCh38
rs774807193 Chr.1: 45621614-45621614 on Build GRCh38
rs750849320 Chr.1: 45621339-45621339 on Build GRCh38
rs768272296 Chr.1: 45621599-45621599 on Build GRCh38
rs757243591 Chr.1: 45621631-45621631 on Build GRCh38
rs530006210 Chr.1: 45621204-45621204 on Build GRCh38
rs758723115 Chr.1: 45621341-45621341 on Build GRCh38
rs993559176 Chr.1: 45621331-45621331 on Build GRCh38
rs767314252 Chr.1: 45621255-45621256 on Build GRCh38
rs765668103 Chr.1: 45621256-45621256 on Build GRCh38
rs763289372 Chr.1: 45621452-45621452 on Build GRCh38
rs746532056 Chr.1: 45621594-45621594 on Build GRCh38
rs773996948 Chr.1: 45621316-45621316 on Build GRCh38
rs942930834 Chr.1: 45621190-45621190 on Build GRCh38
rs552436947 Chr.1: 45621400-45621400 on Build GRCh38
rs895946452 Chr.1: 45621385-45621385 on Build GRCh38
rs775163948 Chr.1: 45621411-45621411 on Build GRCh38
rs763488200 Chr.1: 45621612-45621612 on Build GRCh38
rs377011897 Chr.1: 45621527-45621527 on Build GRCh38
rs371774040 Chr.1: 45621453-45621453 on Build GRCh38
rs867864304 Chr.1: 45621285-45621285 on Build GRCh38
rs370281154 Chr.1: 45621531-45621531 on Build GRCh38
rs755031135 Chr.1: 45621269-45621269 on Build GRCh38
rs370532127 Chr.1: 45621256-45621257 on Build GRCh38
rs990438324 Chr.1: 45621402-45621402 on Build GRCh38
rs3014245 Chr.1: 45621241-45621241 on Build GRCh38
rs767684395 Chr.1: 45621480-45621480 on Build GRCh38
rs1022641149 Chr.1: 45621235-45621235 on Build GRCh38
rs369609741 Chr.1: 45621393-45621393 on Build GRCh38
rs748174410 Chr.1: 45621386-45621386 on Build GRCh38
rs753766451 Chr.1: 45621629-45621629 on Build GRCh38
rs749095154 Chr.1: 45621396-45621396 on Build GRCh38
rs762875576 Chr.1: 45621479-45621479 on Build GRCh38
rs752903141 Chr.1: 45621623-45621623 on Build GRCh38
rs769624054 Chr.1: 45621307-45621307 on Build GRCh38
rs755083111 Chr.1: 45621370-45621370 on Build GRCh38
rs781307741 Chr.1: 45621371-45621371 on Build GRCh38
rs571042755 Chr.1: 45621404-45621404 on Build GRCh38
rs184559988 Chr.1: 45621196-45621196 on Build GRCh38
rs190359188 Chr.1: 45621488-45621488 on Build GRCh38
rs767047954 Chr.1: 45621322-45621322 on Build GRCh38
rs770745084 Chr.1: 45621399-45621399 on Build GRCh38
rs139210753 Chr.1: 45621315-45621315 on Build GRCh38
rs201021595 Chr.1: 45621641-45621641 on Build GRCh38
rs3014244 Chr.1: 45621330-45621330 on Build GRCh38
rs928629646 Chr.1: 45621450-45621450 on Build GRCh38
rs773010574 Chr.1: 45621314-45621314 on Build GRCh38
rs772518775 Chr.1: 45621616-45621620 on Build GRCh38
rs760810886 Chr.1: 45621626-45621626 on Build GRCh38
rs747067400 Chr.1: 45621267-45621267 on Build GRCh38
rs757041758 Chr.1: 45621515-45621515 on Build GRCh38
rs777746007 Chr.1: 45621489-45621489 on Build GRCh38
rs766527900 Chr.1: 45621342-45621342 on Build GRCh38
rs754107692 Chr.1: 45621441-45621441 on Build GRCh38
rs754381858 Chr.1: 45621233-45621233 on Build GRCh38
rs200507548 Chr.1: 45621401-45621401 on Build GRCh38
rs780394352 Chr.1: 45621629-45621629 on Build GRCh38
rs759749100 Chr.1: 45621616-45621616 on Build GRCh38
rs778057210 Chr.1: 45621418-45621419 on Build GRCh38
rs938767164 Chr.1: 45621495-45621495 on Build GRCh38
rs746172322 Chr.1: 45621609-45621610 on Build GRCh38
rs771290388 Chr.1: 45621613-45621613 on Build GRCh38
rs779289692 Chr.1: 45621517-45621518 on Build GRCh38
rs778745558 Chr.1: 45621526-45621526 on Build GRCh38
rs926061422 Chr.1: 45621361-45621361 on Build GRCh38
rs1004632363 Chr.1: 45621487-45621487 on Build GRCh38
rs773800907 Chr.1: 45621440-45621440 on Build GRCh38
rs780061286 Chr.1: 45621310-45621310 on Build GRCh38
rs538469242 Chr.1: 45621420-45621420 on Build GRCh38
rs780284038 Chr.1: 45621264-45621264 on Build GRCh38

View Full Product Details