Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59526685-59527185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC175
Entrez Gene ID
729665
Gene Name
coiled-coil domain containing 175
Gene Aliases
C14orf38, c14_5395
UniGene
Hs.29706
Gene Chromosome Location
Chr.14: 59504212-59577276 on Build GRCh38
Transcripts
NM_001164399.1 XM_011537120.2 XM_011537122.2 XM_011537124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5959765 c.1800T>C p.I600I Chr.14: 59527137-59527137 on Build GRCh38
4188250 c.1837A>T p.N613Y Chr.14: 59527100-59527100 on Build GRCh38
5959501 c.1814G>C p.R605T Chr.14: 59527123-59527123 on Build GRCh38
4188251 c.1837A>T p.N613Y Chr.14: 59527100-59527100 on Build GRCh38
5959766 c.1800T>C p.I600I Chr.14: 59527137-59527137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545451874 Chr.14: 59526911-59526911 on Build GRCh38
rs748215649 Chr.14: 59527059-59527059 on Build GRCh38
rs900189362 Chr.14: 59527047-59527047 on Build GRCh38
rs574186184 Chr.14: 59526834-59526834 on Build GRCh38
rs1040447452 Chr.14: 59526843-59526843 on Build GRCh38
rs906679756 Chr.14: 59526923-59526923 on Build GRCh38
rs35564902 Chr.14: 59526744-59526745 on Build GRCh38
rs200681540 Chr.14: 59527123-59527123 on Build GRCh38
rs927944660 Chr.14: 59526749-59526749 on Build GRCh38
rs561128155 Chr.14: 59526817-59526817 on Build GRCh38
rs945626572 Chr.14: 59526920-59526920 on Build GRCh38
rs560465397 Chr.14: 59526939-59526939 on Build GRCh38
rs887543470 Chr.14: 59527085-59527085 on Build GRCh38
rs183975208 Chr.14: 59526983-59526983 on Build GRCh38
rs924072906 Chr.14: 59526980-59526980 on Build GRCh38
rs781294261 Chr.14: 59527048-59527048 on Build GRCh38
rs113032592 Chr.14: 59526785-59526785 on Build GRCh38
rs982537035 Chr.14: 59526743-59526743 on Build GRCh38
rs1004593589 Chr.14: 59527095-59527095 on Build GRCh38
rs1040626214 Chr.14: 59527039-59527039 on Build GRCh38
rs762823116 Chr.14: 59527101-59527101 on Build GRCh38
rs995441094 Chr.14: 59526818-59526818 on Build GRCh38
rs1010481351 Chr.14: 59527066-59527066 on Build GRCh38
rs531556831 Chr.14: 59527001-59527001 on Build GRCh38
rs770583079 Chr.14: 59527102-59527102 on Build GRCh38
rs1002304962 Chr.14: 59526936-59526936 on Build GRCh38
rs77868958 Chr.14: 59526888-59526888 on Build GRCh38
rs914187014 Chr.14: 59526917-59526917 on Build GRCh38
rs1046581889 Chr.14: 59526921-59526921 on Build GRCh38
rs547976594 Chr.14: 59526868-59526868 on Build GRCh38
rs573239489 Chr.14: 59526710-59526710 on Build GRCh38
rs990913858 Chr.14: 59526807-59526807 on Build GRCh38
rs915451231 Chr.14: 59526808-59526808 on Build GRCh38
rs770071340 Chr.14: 59527064-59527064 on Build GRCh38
rs770705141 Chr.14: 59527064-59527064 on Build GRCh38
rs965559240 Chr.14: 59527094-59527094 on Build GRCh38
rs887636941 Chr.14: 59526880-59526880 on Build GRCh38
rs1033713276 Chr.14: 59526816-59526816 on Build GRCh38
rs778445381 Chr.14: 59527029-59527029 on Build GRCh38
rs773244882 Chr.14: 59527079-59527079 on Build GRCh38
rs912383597 Chr.14: 59526853-59526853 on Build GRCh38
rs1055594233 Chr.14: 59526937-59526937 on Build GRCh38
rs951315790 Chr.14: 59526964-59526964 on Build GRCh38
rs755565897 Chr.14: 59526955-59526955 on Build GRCh38
rs8011271 Chr.14: 59526945-59526945 on Build GRCh38
rs372867826 Chr.14: 59527140-59527140 on Build GRCh38
rs996367185 Chr.14: 59526860-59526860 on Build GRCh38
rs936319779 Chr.14: 59526909-59526909 on Build GRCh38
rs982646999 Chr.14: 59526837-59526837 on Build GRCh38
rs527869030 Chr.14: 59526856-59526856 on Build GRCh38
rs17834184 Chr.14: 59527137-59527137 on Build GRCh38
rs756860324 Chr.14: 59526878-59526878 on Build GRCh38
rs959399714 Chr.14: 59526926-59526926 on Build GRCh38
rs533150588 Chr.14: 59526890-59526890 on Build GRCh38
rs906656471 Chr.14: 59526714-59526714 on Build GRCh38
rs538092651 Chr.14: 59526998-59526998 on Build GRCh38
rs781544018 Chr.14: 59527034-59527034 on Build GRCh38
rs569726524 Chr.14: 59526827-59526827 on Build GRCh38

View Full Product Details