Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66593541-66594046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCS
Entrez Gene ID
9973
Gene Name
copper chaperone for superoxide dismutase
Gene Aliases
-
UniGene
Hs.502917
Gene Chromosome Location
Chr.11: 66593162-66606019 on Build GRCh38
Transcripts
NM_005125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6025193 c.109G>A p.A37T Chr.11: 66593711-66593711 on Build GRCh38
5587452 c.93A>T p.K31N Chr.11: 66593695-66593695 on Build GRCh38
7360148 c.109G>T p.A37S Chr.11: 66593711-66593711 on Build GRCh38
6552313 c.89G>T p.R30L Chr.11: 66593691-66593691 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs971937350 Chr.11: 66593894-66593894 on Build GRCh38
rs902467791 Chr.11: 66593908-66593908 on Build GRCh38
rs774081476 Chr.11: 66593682-66593682 on Build GRCh38
rs1042299820 Chr.11: 66593906-66593906 on Build GRCh38
rs917381054 Chr.11: 66593826-66593826 on Build GRCh38
rs761007631 Chr.11: 66593713-66593729 on Build GRCh38
rs756233845 Chr.11: 66593727-66593727 on Build GRCh38
rs1003191553 Chr.11: 66593926-66593926 on Build GRCh38
rs371683431 Chr.11: 66593764-66593764 on Build GRCh38
rs548374979 Chr.11: 66593572-66593572 on Build GRCh38
rs754306514 Chr.11: 66593613-66593613 on Build GRCh38
rs959081345 Chr.11: 66593968-66593968 on Build GRCh38
rs368346498 Chr.11: 66593772-66593772 on Build GRCh38
rs779201305 Chr.11: 66593616-66593616 on Build GRCh38
rs940376895 Chr.11: 66594016-66594016 on Build GRCh38
rs768626429 Chr.11: 66593680-66593680 on Build GRCh38
rs757424594 Chr.11: 66593765-66593765 on Build GRCh38
rs908503027 Chr.11: 66593745-66593745 on Build GRCh38
rs755501132 Chr.11: 66593711-66593711 on Build GRCh38
rs959133362 Chr.11: 66593665-66593665 on Build GRCh38
rs1011206541 Chr.11: 66593838-66593838 on Build GRCh38
rs980306684 Chr.11: 66593946-66593946 on Build GRCh38
rs749514184 Chr.11: 66593856-66593856 on Build GRCh38
rs749979522 Chr.11: 66593595-66593595 on Build GRCh38
rs748538018 Chr.11: 66593623-66593623 on Build GRCh38
rs79152198 Chr.11: 66593740-66593740 on Build GRCh38
rs765810923 Chr.11: 66593704-66593704 on Build GRCh38
rs1049369413 Chr.11: 66594007-66594007 on Build GRCh38
rs563806954 Chr.11: 66593686-66593686 on Build GRCh38
rs927510700 Chr.11: 66593964-66593964 on Build GRCh38
rs1049527324 Chr.11: 66593816-66593816 on Build GRCh38
rs772522566 Chr.11: 66593649-66593650 on Build GRCh38
rs932753524 Chr.11: 66593790-66593790 on Build GRCh38
rs113810834 Chr.11: 66593685-66593685 on Build GRCh38
rs760212307 Chr.11: 66593691-66593691 on Build GRCh38
rs967951335 Chr.11: 66593698-66593698 on Build GRCh38
rs377031211 Chr.11: 66593705-66593705 on Build GRCh38
rs773421206 Chr.11: 66593650-66593651 on Build GRCh38
rs893688168 Chr.11: 66593589-66593589 on Build GRCh38
rs775571378 Chr.11: 66593646-66593646 on Build GRCh38
rs372515372 Chr.11: 66593638-66593638 on Build GRCh38
rs767210418 Chr.11: 66593684-66593684 on Build GRCh38
rs753372996 Chr.11: 66593709-66593709 on Build GRCh38
rs981808293 Chr.11: 66593619-66593619 on Build GRCh38
rs189468375 Chr.11: 66593842-66593842 on Build GRCh38
rs371511954 Chr.11: 66593601-66593601 on Build GRCh38
rs755376705 Chr.11: 66593614-66593614 on Build GRCh38
rs376776159 Chr.11: 66593594-66593594 on Build GRCh38
rs758584721 Chr.11: 66593632-66593632 on Build GRCh38
rs771093151 Chr.11: 66593645-66593645 on Build GRCh38
rs778171747 Chr.11: 66593759-66593759 on Build GRCh38
rs765740277 Chr.11: 66593713-66593713 on Build GRCh38
rs780188780 Chr.11: 66593771-66593771 on Build GRCh38
rs368320132 Chr.11: 66593634-66593634 on Build GRCh38
rs755599396 Chr.11: 66593597-66593597 on Build GRCh38
rs907529803 Chr.11: 66594012-66594012 on Build GRCh38
rs930861835 Chr.11: 66593982-66593982 on Build GRCh38
rs989296421 Chr.11: 66593785-66593785 on Build GRCh38
rs374487026 Chr.11: 66593605-66593605 on Build GRCh38
rs373561611 Chr.11: 66593570-66593570 on Build GRCh38
rs543925670 Chr.11: 66593683-66593683 on Build GRCh38
rs749120332 Chr.11: 66593650-66593650 on Build GRCh38

View Full Product Details