Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207322271-207322767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD55
Entrez Gene ID
1604
Gene Name
CD55 molecule (Cromer blood group)
Gene Aliases
CR, CROM, DAF, TC
UniGene
Hs.126517
Gene Chromosome Location
Chr.1: 207321472-207360966 on Build GRCh38
Transcripts
NM_000574.4 NM_001114752.2 NM_001300902.1 NM_001300903.1 NM_001300904.1 NR_125349.1 XM_017000467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4764483 c.106T>C p.C36R Chr.1: 207322387-207322387 on Build GRCh38
6237541 c.178G>T p.V60L Chr.1: 207322459-207322459 on Build GRCh38
4027596 c.277T>C p.F93L Chr.1: 207322558-207322558 on Build GRCh38
1247711 c.237G>T p.V79V Chr.1: 207322518-207322518 on Build GRCh38
4027597 c.277T>C p.F93L Chr.1: 207322558-207322558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147474393 Chr.1: 207322526-207322526 on Build GRCh38
rs756115395 Chr.1: 207322663-207322663 on Build GRCh38
rs764300881 Chr.1: 207322447-207322447 on Build GRCh38
rs751607950 Chr.1: 207322340-207322340 on Build GRCh38
rs773939798 Chr.1: 207322417-207322417 on Build GRCh38
rs758471550 Chr.1: 207322318-207322318 on Build GRCh38
rs112362936 Chr.1: 207322651-207322651 on Build GRCh38
rs913635154 Chr.1: 207322566-207322566 on Build GRCh38
rs764220870 Chr.1: 207322324-207322324 on Build GRCh38
rs992669189 Chr.1: 207322685-207322686 on Build GRCh38
rs767031474 Chr.1: 207322502-207322502 on Build GRCh38
rs754207835 Chr.1: 207322567-207322567 on Build GRCh38
rs771997746 Chr.1: 207322428-207322428 on Build GRCh38
rs745638347 Chr.1: 207322617-207322617 on Build GRCh38
rs557247015 Chr.1: 207322671-207322671 on Build GRCh38
rs200552334 Chr.1: 207322539-207322539 on Build GRCh38
rs757303688 Chr.1: 207322452-207322452 on Build GRCh38
rs776347919 Chr.1: 207322520-207322520 on Build GRCh38
rs1017610777 Chr.1: 207322503-207322503 on Build GRCh38
rs770872713 Chr.1: 207322517-207322517 on Build GRCh38
rs121909603 Chr.1: 207322542-207322542 on Build GRCh38
rs781576624 Chr.1: 207322376-207322376 on Build GRCh38
rs28371588 Chr.1: 207322436-207322436 on Build GRCh38
rs55973205 Chr.1: 207322688-207322689 on Build GRCh38
rs768043655 Chr.1: 207322455-207322455 on Build GRCh38
rs752360687 Chr.1: 207322569-207322569 on Build GRCh38
rs746529395 Chr.1: 207322580-207322580 on Build GRCh38
rs746193185 Chr.1: 207322385-207322385 on Build GRCh38
rs766996458 Chr.1: 207322565-207322565 on Build GRCh38
rs370729319 Chr.1: 207322577-207322577 on Build GRCh38
rs749682264 Chr.1: 207322474-207322474 on Build GRCh38
rs376304458 Chr.1: 207322568-207322568 on Build GRCh38
rs747673434 Chr.1: 207322492-207322492 on Build GRCh38
rs774482588 Chr.1: 207322536-207322536 on Build GRCh38
rs773709328 Chr.1: 207322548-207322548 on Build GRCh38
rs527859888 Chr.1: 207322694-207322694 on Build GRCh38
rs773074921 Chr.1: 207322429-207322429 on Build GRCh38
rs760411921 Chr.1: 207322310-207322310 on Build GRCh38
rs989033407 Chr.1: 207322538-207322538 on Build GRCh38
rs771031409 Chr.1: 207322505-207322507 on Build GRCh38
rs747643776 Chr.1: 207322419-207322419 on Build GRCh38
rs569926561 Chr.1: 207322295-207322295 on Build GRCh38
rs538797583 Chr.1: 207322307-207322307 on Build GRCh38
rs138501379 Chr.1: 207322734-207322734 on Build GRCh38
rs917883888 Chr.1: 207322693-207322693 on Build GRCh38
rs926295322 Chr.1: 207322657-207322657 on Build GRCh38
rs28371590 Chr.1: 207322737-207322737 on Build GRCh38
rs763107135 Chr.1: 207322438-207322438 on Build GRCh38
rs756591988 Chr.1: 207322396-207322396 on Build GRCh38
rs1026797430 Chr.1: 207322359-207322359 on Build GRCh38
rs750932474 Chr.1: 207322470-207322470 on Build GRCh38
rs778452813 Chr.1: 207322483-207322483 on Build GRCh38
rs780622604 Chr.1: 207322472-207322472 on Build GRCh38
rs147086040 Chr.1: 207322633-207322633 on Build GRCh38
rs759353838 Chr.1: 207322525-207322525 on Build GRCh38
rs397897243 Chr.1: 207322685-207322686 on Build GRCh38
rs373306908 Chr.1: 207322509-207322509 on Build GRCh38
rs369907962 Chr.1: 207322450-207322450 on Build GRCh38
rs757916547 Chr.1: 207322570-207322570 on Build GRCh38
rs747009122 Chr.1: 207322510-207322510 on Build GRCh38
rs761868192 Chr.1: 207322296-207322296 on Build GRCh38
rs781125021 Chr.1: 207322604-207322604 on Build GRCh38
rs186930821 Chr.1: 207322315-207322315 on Build GRCh38
rs768268102 Chr.1: 207322413-207322413 on Build GRCh38
rs767361258 Chr.1: 207322302-207322302 on Build GRCh38
rs993358787 Chr.1: 207322338-207322338 on Build GRCh38
rs28371589 Chr.1: 207322684-207322685 on Build GRCh38
rs979619069 Chr.1: 207322656-207322656 on Build GRCh38
rs749013692 Chr.1: 207322406-207322406 on Build GRCh38
rs761919258 Chr.1: 207322537-207322537 on Build GRCh38
rs768319455 Chr.1: 207322294-207322294 on Build GRCh38
rs869312818 Chr.1: 207322484-207322484 on Build GRCh38
rs766278981 Chr.1: 207322675-207322675 on Build GRCh38
rs200324555 Chr.1: 207322355-207322355 on Build GRCh38
rs750927655 Chr.1: 207322545-207322545 on Build GRCh38
rs754637367 Chr.1: 207322476-207322476 on Build GRCh38
rs929127674 Chr.1: 207322717-207322717 on Build GRCh38
rs761206859 Chr.1: 207322556-207322556 on Build GRCh38
rs770811441 Chr.1: 207322583-207322583 on Build GRCh38
rs779820703 Chr.1: 207322400-207322400 on Build GRCh38
rs896490746 Chr.1: 207322363-207322363 on Build GRCh38
rs867810442 Chr.1: 207322387-207322388 on Build GRCh38
rs926734160 Chr.1: 207322316-207322316 on Build GRCh38

View Full Product Details