Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116519144-116519557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD58
Entrez Gene ID
965
Gene Name
CD58 molecule
Gene Aliases
LFA-3, LFA3, ag3
UniGene
Hs.34341
Gene Chromosome Location
Chr.1: 116487140-116571093 on Build GRCh38
Transcripts
NM_001144822.1 NM_001779.2 NR_026665.1 XM_017002869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4506160 c.712C>T p.L238L Chr.1: 116519262-116519262 on Build GRCh38
4670519 c.707-1G>T p.? Chr.1: 116519268-116519268 on Build GRCh38
5020794 c.709A>G p.I237V Chr.1: 116519265-116519265 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1046820374 Chr.1: 116519493-116519493 on Build GRCh38
rs762356983 Chr.1: 116519348-116519348 on Build GRCh38
rs768021176 Chr.1: 116519350-116519350 on Build GRCh38
rs748132625 Chr.1: 116519227-116519227 on Build GRCh38
rs776683149 Chr.1: 116519347-116519347 on Build GRCh38
rs779126536 Chr.1: 116519223-116519223 on Build GRCh38
rs142073474 Chr.1: 116519207-116519207 on Build GRCh38
rs770492818 Chr.1: 116519303-116519303 on Build GRCh38
rs910013512 Chr.1: 116519498-116519498 on Build GRCh38
rs929675359 Chr.1: 116519468-116519468 on Build GRCh38
rs922352043 Chr.1: 116519438-116519438 on Build GRCh38
rs767838030 Chr.1: 116519424-116519424 on Build GRCh38
rs149627900 Chr.1: 116519265-116519265 on Build GRCh38
rs768834198 Chr.1: 116519200-116519200 on Build GRCh38
rs963668550 Chr.1: 116519397-116519397 on Build GRCh38
rs911158889 Chr.1: 116519392-116519392 on Build GRCh38
rs755055104 Chr.1: 116519222-116519222 on Build GRCh38
rs565866049 Chr.1: 116519384-116519384 on Build GRCh38
rs776287766 Chr.1: 116519307-116519307 on Build GRCh38
rs749336081 Chr.1: 116519221-116519221 on Build GRCh38
rs774949975 Chr.1: 116519338-116519338 on Build GRCh38
rs529692391 Chr.1: 116519526-116519526 on Build GRCh38
rs764793526 Chr.1: 116519323-116519323 on Build GRCh38
rs774606563 Chr.1: 116519210-116519210 on Build GRCh38
rs201656599 Chr.1: 116519240-116519240 on Build GRCh38
rs1003569757 Chr.1: 116519176-116519176 on Build GRCh38
rs1035092021 Chr.1: 116519185-116519185 on Build GRCh38
rs979060272 Chr.1: 116519381-116519381 on Build GRCh38
rs150868128 Chr.1: 116519251-116519251 on Build GRCh38
rs780818017 Chr.1: 116519446-116519446 on Build GRCh38
rs759174048 Chr.1: 116519316-116519316 on Build GRCh38
rs756314785 Chr.1: 116519192-116519192 on Build GRCh38
rs750778811 Chr.1: 116519189-116519189 on Build GRCh38

View Full Product Details