Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25459149-25459648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
UniGene
Hs.33366
Gene Chromosome Location
Chr.8: 25458569-25511308 on Build GRCh38
Transcripts
NM_001317906.1 NM_001317907.1 NM_152562.3 XM_006716294.2 XM_011544415.1 XM_011544416.2 XM_011544417.2 XM_011544418.1

SNP Detail

SNP ID Chromosome Location
rs1022200085 Chr.8: 25459264-25459264 on Build GRCh38
rs948901202 Chr.8: 25459332-25459332 on Build GRCh38
rs1037935230 Chr.8: 25459246-25459246 on Build GRCh38
rs1008964483 Chr.8: 25459429-25459429 on Build GRCh38
rs1022674076 Chr.8: 25459171-25459171 on Build GRCh38
rs553852298 Chr.8: 25459417-25459417 on Build GRCh38
rs902447115 Chr.8: 25459269-25459269 on Build GRCh38
rs951905736 Chr.8: 25459563-25459563 on Build GRCh38
rs535180637 Chr.8: 25459328-25459328 on Build GRCh38
rs947383063 Chr.8: 25459420-25459420 on Build GRCh38
rs999456896 Chr.8: 25459521-25459521 on Build GRCh38
rs996750419 Chr.8: 25459232-25459232 on Build GRCh38
rs3812421 Chr.8: 25459273-25459273 on Build GRCh38
rs960412538 Chr.8: 25459275-25459275 on Build GRCh38
rs932191764 Chr.8: 25459360-25459360 on Build GRCh38
rs977815174 Chr.8: 25459188-25459189 on Build GRCh38
rs990500010 Chr.8: 25459623-25459623 on Build GRCh38
rs777234420 Chr.8: 25459593-25459593 on Build GRCh38
rs930342654 Chr.8: 25459198-25459198 on Build GRCh38
rs894655475 Chr.8: 25459244-25459244 on Build GRCh38
rs759177645 Chr.8: 25459527-25459527 on Build GRCh38
rs568193537 Chr.8: 25459183-25459183 on Build GRCh38
rs7833509 Chr.8: 25459229-25459229 on Build GRCh38
rs1050515067 Chr.8: 25459363-25459363 on Build GRCh38
rs976186160 Chr.8: 25459333-25459333 on Build GRCh38
rs371661194 Chr.8: 25459395-25459395 on Build GRCh38
rs570316056 Chr.8: 25459270-25459270 on Build GRCh38
rs1049990927 Chr.8: 25459235-25459235 on Build GRCh38
rs1039151642 Chr.8: 25459453-25459453 on Build GRCh38
rs1035059520 Chr.8: 25459540-25459540 on Build GRCh38
rs1026335268 Chr.8: 25459258-25459258 on Build GRCh38
rs999452518 Chr.8: 25459458-25459458 on Build GRCh38
rs984876570 Chr.8: 25459298-25459298 on Build GRCh38
rs1042131333 Chr.8: 25459421-25459421 on Build GRCh38
rs990531070 Chr.8: 25459310-25459310 on Build GRCh38
rs908146621 Chr.8: 25459228-25459228 on Build GRCh38
rs762725048 Chr.8: 25459245-25459245 on Build GRCh38
rs141240299 Chr.8: 25459306-25459306 on Build GRCh38
rs951730728 Chr.8: 25459285-25459285 on Build GRCh38
rs927671424 Chr.8: 25459180-25459180 on Build GRCh38
rs960464932 Chr.8: 25459182-25459182 on Build GRCh38
rs879480392 Chr.8: 25459277-25459278 on Build GRCh38
rs764679542 Chr.8: 25459542-25459542 on Build GRCh38
rs1012016064 Chr.8: 25459262-25459262 on Build GRCh38
rs990447905 Chr.8: 25459187-25459187 on Build GRCh38
rs1017485060 Chr.8: 25459175-25459176 on Build GRCh38
rs967021995 Chr.8: 25459515-25459515 on Build GRCh38
rs923434264 Chr.8: 25459337-25459337 on Build GRCh38
rs893239901 Chr.8: 25459398-25459398 on Build GRCh38
rs527790792 Chr.8: 25459248-25459248 on Build GRCh38
rs1037900658 Chr.8: 25459338-25459338 on Build GRCh38
rs893134611 Chr.8: 25459261-25459261 on Build GRCh38
rs896300603 Chr.8: 25459487-25459487 on Build GRCh38
rs966551000 Chr.8: 25459178-25459178 on Build GRCh38
rs751043534 Chr.8: 25459435-25459435 on Build GRCh38
rs762756803 Chr.8: 25459335-25459336 on Build GRCh38
rs945050686 Chr.8: 25459181-25459181 on Build GRCh38
rs990581556 Chr.8: 25459276-25459276 on Build GRCh38
rs902394661 Chr.8: 25459428-25459428 on Build GRCh38
rs920824001 Chr.8: 25459378-25459378 on Build GRCh38
rs1049296889 Chr.8: 25459208-25459208 on Build GRCh38
rs1023827493 Chr.8: 25459277-25459277 on Build GRCh38
rs964591516 Chr.8: 25459176-25459176 on Build GRCh38
rs1050980264 Chr.8: 25459253-25459253 on Build GRCh38
rs549629035 Chr.8: 25459287-25459287 on Build GRCh38
rs559263896 Chr.8: 25459565-25459565 on Build GRCh38
rs916239031 Chr.8: 25459188-25459188 on Build GRCh38
rs960378398 Chr.8: 25459566-25459566 on Build GRCh38
rs761439267 Chr.8: 25459497-25459497 on Build GRCh38
rs145029103 Chr.8: 25459473-25459473 on Build GRCh38
rs1031768047 Chr.8: 25459418-25459418 on Build GRCh38
rs940948378 Chr.8: 25459325-25459325 on Build GRCh38
rs890466688 Chr.8: 25459423-25459423 on Build GRCh38
rs901895753 Chr.8: 25459247-25459247 on Build GRCh38
rs927686415 Chr.8: 25459303-25459303 on Build GRCh38
rs534496860 Chr.8: 25459543-25459543 on Build GRCh38
rs977951561 Chr.8: 25459179-25459179 on Build GRCh38

View Full Product Details