Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6850363-6850870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA3
Entrez Gene ID
83461
Gene Name
cell division cycle associated 3
Gene Aliases
GRCC8, TOME-1
UniGene
Hs.524216
Gene Chromosome Location
Chr.12: 6844796-6852066 on Build GRCh38
Transcripts
NM_001297602.1 NM_001297603.1 NM_001297604.1 NM_031299.5 NR_123727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5375847 c.118C>T p.Q40* Chr.12: 6850835-6850835 on Build GRCh38
1299826 c.234G>A p.M78I Chr.12: 6850483-6850483 on Build GRCh38
5440855 c.199C>T p.R67C Chr.12: 6850518-6850518 on Build GRCh38
6348325 c.117C>A p.I39I Chr.12: 6850836-6850836 on Build GRCh38
6374007 c.124G>T p.E42* Chr.12: 6850593-6850593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782385891 Chr.12: 6850614-6850614 on Build GRCh38
rs200560402 Chr.12: 6850620-6850620 on Build GRCh38
rs371648676 Chr.12: 6850843-6850843 on Build GRCh38
rs782413400 Chr.12: 6850819-6850819 on Build GRCh38
rs782608399 Chr.12: 6850446-6850446 on Build GRCh38
rs200061658 Chr.12: 6850600-6850600 on Build GRCh38
rs960697128 Chr.12: 6850403-6850403 on Build GRCh38
rs902306850 Chr.12: 6850645-6850645 on Build GRCh38
rs368270613 Chr.12: 6850682-6850682 on Build GRCh38
rs1043444438 Chr.12: 6850643-6850643 on Build GRCh38
rs782479855 Chr.12: 6850813-6850813 on Build GRCh38
rs782270888 Chr.12: 6850567-6850567 on Build GRCh38
rs376054935 Chr.12: 6850817-6850817 on Build GRCh38
rs367726225 Chr.12: 6850485-6850485 on Build GRCh38
rs376349479 Chr.12: 6850595-6850595 on Build GRCh38
rs907981293 Chr.12: 6850470-6850470 on Build GRCh38
rs781928783 Chr.12: 6850494-6850494 on Build GRCh38
rs753222013 Chr.12: 6850659-6850659 on Build GRCh38
rs1035342190 Chr.12: 6850394-6850394 on Build GRCh38
rs781953121 Chr.12: 6850499-6850499 on Build GRCh38
rs782009387 Chr.12: 6850476-6850476 on Build GRCh38
rs138876608 Chr.12: 6850454-6850454 on Build GRCh38
rs782212714 Chr.12: 6850448-6850448 on Build GRCh38
rs782308916 Chr.12: 6850846-6850846 on Build GRCh38
rs782668155 Chr.12: 6850566-6850566 on Build GRCh38
rs782638646 Chr.12: 6850803-6850803 on Build GRCh38
rs145639640 Chr.12: 6850647-6850647 on Build GRCh38
rs200589400 Chr.12: 6850517-6850517 on Build GRCh38
rs781886575 Chr.12: 6850502-6850502 on Build GRCh38
rs781808533 Chr.12: 6850626-6850626 on Build GRCh38
rs372106407 Chr.12: 6850796-6850796 on Build GRCh38
rs376580038 Chr.12: 6850433-6850433 on Build GRCh38
rs143204334 Chr.12: 6850563-6850563 on Build GRCh38
rs782043036 Chr.12: 6850498-6850498 on Build GRCh38
rs782463014 Chr.12: 6850548-6850548 on Build GRCh38
rs148155861 Chr.12: 6850702-6850702 on Build GRCh38
rs782140009 Chr.12: 6850602-6850602 on Build GRCh38
rs782807541 Chr.12: 6850501-6850501 on Build GRCh38
rs782380641 Chr.12: 6850570-6850570 on Build GRCh38
rs782548777 Chr.12: 6850422-6850422 on Build GRCh38
rs782051229 Chr.12: 6850644-6850644 on Build GRCh38
rs200147836 Chr.12: 6850808-6850808 on Build GRCh38
rs782281944 Chr.12: 6850818-6850818 on Build GRCh38
rs782527027 Chr.12: 6850789-6850789 on Build GRCh38
rs781871819 Chr.12: 6850578-6850578 on Build GRCh38
rs781925367 Chr.12: 6850459-6850459 on Build GRCh38
rs1012136844 Chr.12: 6850823-6850823 on Build GRCh38
rs376412771 Chr.12: 6850415-6850415 on Build GRCh38
rs150858074 Chr.12: 6850587-6850587 on Build GRCh38
rs782412281 Chr.12: 6850461-6850461 on Build GRCh38
rs781980130 Chr.12: 6850615-6850615 on Build GRCh38
rs367977966 Chr.12: 6850838-6850838 on Build GRCh38
rs782098205 Chr.12: 6850619-6850619 on Build GRCh38
rs202020864 Chr.12: 6850825-6850825 on Build GRCh38
rs373307710 Chr.12: 6850429-6850429 on Build GRCh38
rs782243097 Chr.12: 6850836-6850836 on Build GRCh38
rs782767268 Chr.12: 6850492-6850492 on Build GRCh38
rs781910283 Chr.12: 6850484-6850485 on Build GRCh38
rs781891144 Chr.12: 6850787-6850787 on Build GRCh38
rs187591628 Chr.12: 6850460-6850460 on Build GRCh38
rs782699295 Chr.12: 6850596-6850596 on Build GRCh38
rs191917652 Chr.12: 6850685-6850685 on Build GRCh38
rs782197438 Chr.12: 6850391-6850391 on Build GRCh38
rs565223479 Chr.12: 6850518-6850518 on Build GRCh38
rs782618257 Chr.12: 6850586-6850586 on Build GRCh38
rs782513128 Chr.12: 6850435-6850435 on Build GRCh38
rs371123912 Chr.12: 6850503-6850503 on Build GRCh38
rs781791457 Chr.12: 6850557-6850557 on Build GRCh38
rs782188663 Chr.12: 6850649-6850649 on Build GRCh38

View Full Product Details