Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92609068-92609581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK6
Entrez Gene ID
1021
Gene Name
cyclin dependent kinase 6
Gene Aliases
MCPH12, PLSTIRE
UniGene
Hs.119882
Gene Chromosome Location
Chr.7: 92604921-92836627 on Build GRCh38
Transcripts
NM_001145306.1 NM_001259.6 XM_006715835.2

SNP Detail

SNP ID Chromosome Location
rs535061215 Chr.7: 92609149-92609149 on Build GRCh38
rs572103922 Chr.7: 92609243-92609243 on Build GRCh38
rs10241602 Chr.7: 92609442-92609442 on Build GRCh38
rs563951956 Chr.7: 92609448-92609448 on Build GRCh38
rs1049699793 Chr.7: 92609548-92609548 on Build GRCh38
rs1047759889 Chr.7: 92609356-92609356 on Build GRCh38
rs558292271 Chr.7: 92609268-92609268 on Build GRCh38
rs1037124913 Chr.7: 92609116-92609116 on Build GRCh38
rs955228739 Chr.7: 92609168-92609168 on Build GRCh38
rs964840459 Chr.7: 92609091-92609091 on Build GRCh38
rs1037918268 Chr.7: 92609405-92609405 on Build GRCh38
rs934105255 Chr.7: 92609511-92609511 on Build GRCh38
rs937833369 Chr.7: 92609249-92609249 on Build GRCh38
rs890025609 Chr.7: 92609209-92609209 on Build GRCh38
rs997002741 Chr.7: 92609096-92609096 on Build GRCh38
rs768615372 Chr.7: 92609466-92609466 on Build GRCh38
rs939870528 Chr.7: 92609442-92609443 on Build GRCh38
rs996331425 Chr.7: 92609414-92609414 on Build GRCh38
rs900686786 Chr.7: 92609407-92609407 on Build GRCh38
rs1037498843 Chr.7: 92609468-92609468 on Build GRCh38
rs554615064 Chr.7: 92609432-92609432 on Build GRCh38
rs540774296 Chr.7: 92609247-92609247 on Build GRCh38
rs376849672 Chr.7: 92609186-92609186 on Build GRCh38
rs547545208 Chr.7: 92609164-92609164 on Build GRCh38
rs771403567 Chr.7: 92609316-92609316 on Build GRCh38
rs986464499 Chr.7: 92609176-92609176 on Build GRCh38
rs979283566 Chr.7: 92609214-92609214 on Build GRCh38
rs1028050558 Chr.7: 92609147-92609147 on Build GRCh38
rs529651816 Chr.7: 92609477-92609477 on Build GRCh38
rs886727481 Chr.7: 92609402-92609402 on Build GRCh38
rs981787131 Chr.7: 92609346-92609346 on Build GRCh38
rs949439608 Chr.7: 92609352-92609352 on Build GRCh38
rs147562791 Chr.7: 92609224-92609224 on Build GRCh38
rs760082414 Chr.7: 92609507-92609507 on Build GRCh38
rs578123537 Chr.7: 92609272-92609272 on Build GRCh38
rs774882093 Chr.7: 92609360-92609360 on Build GRCh38
rs188870214 Chr.7: 92609310-92609310 on Build GRCh38
rs866364417 Chr.7: 92609219-92609219 on Build GRCh38
rs970776784 Chr.7: 92609345-92609345 on Build GRCh38
rs966591073 Chr.7: 92609194-92609194 on Build GRCh38
rs1006488265 Chr.7: 92609404-92609404 on Build GRCh38
rs956269030 Chr.7: 92609295-92609299 on Build GRCh38
rs901127984 Chr.7: 92609141-92609141 on Build GRCh38
rs566276706 Chr.7: 92609187-92609187 on Build GRCh38
rs955147528 Chr.7: 92609500-92609500 on Build GRCh38
rs988262508 Chr.7: 92609300-92609300 on Build GRCh38
rs1020706389 Chr.7: 92609313-92609313 on Build GRCh38

View Full Product Details