Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44671107-44671630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM19
Entrez Gene ID
56971
Gene Name
carcinoembryonic antigen related cell adhesion molecule 19
Gene Aliases
CEACM19, CEAL1
UniGene
Hs.416925
Gene Chromosome Location
Chr.19: 44671452-44684355 on Build GRCh38
Transcripts
NM_001127893.1 NM_020219.3

SNP Detail

SNP ID Chromosome Location
rs1011922214 Chr.19: 44671586-44671586 on Build GRCh38
rs540940282 Chr.19: 44671355-44671355 on Build GRCh38
rs370255805 Chr.19: 44671224-44671224 on Build GRCh38
rs768977309 Chr.19: 44671400-44671400 on Build GRCh38
rs966352332 Chr.19: 44671290-44671291 on Build GRCh38
rs770632345 Chr.19: 44671291-44671291 on Build GRCh38
rs112515868 Chr.19: 44671185-44671185 on Build GRCh38
rs1048976450 Chr.19: 44671152-44671152 on Build GRCh38
rs532260790 Chr.19: 44671179-44671179 on Build GRCh38
rs1011182352 Chr.19: 44671575-44671575 on Build GRCh38
rs75232690 Chr.19: 44671138-44671138 on Build GRCh38
rs984209118 Chr.19: 44671410-44671410 on Build GRCh38
rs773550657 Chr.19: 44671182-44671182 on Build GRCh38
rs959148399 Chr.19: 44671203-44671203 on Build GRCh38
rs1050125776 Chr.19: 44671512-44671512 on Build GRCh38
rs1010630784 Chr.19: 44671279-44671279 on Build GRCh38
rs573884581 Chr.19: 44671284-44671284 on Build GRCh38
rs878895188 Chr.19: 44671553-44671557 on Build GRCh38
rs894817525 Chr.19: 44671538-44671538 on Build GRCh38
rs899444714 Chr.19: 44671446-44671446 on Build GRCh38
rs1024697571 Chr.19: 44671607-44671607 on Build GRCh38
rs998422435 Chr.19: 44671257-44671257 on Build GRCh38
rs1022475004 Chr.19: 44671280-44671280 on Build GRCh38
rs374734191 Chr.19: 44671247-44671247 on Build GRCh38
rs923905296 Chr.19: 44671265-44671265 on Build GRCh38
rs999182709 Chr.19: 44671601-44671601 on Build GRCh38
rs78585934 Chr.19: 44671140-44671140 on Build GRCh38
rs528110572 Chr.19: 44671336-44671336 on Build GRCh38
rs922010756 Chr.19: 44671302-44671302 on Build GRCh38
rs890597260 Chr.19: 44671396-44671396 on Build GRCh38
rs970645003 Chr.19: 44671409-44671409 on Build GRCh38
rs1031925728 Chr.19: 44671303-44671303 on Build GRCh38
rs896789073 Chr.19: 44671168-44671168 on Build GRCh38
rs936656326 Chr.19: 44671268-44671268 on Build GRCh38
rs922615272 Chr.19: 44671489-44671489 on Build GRCh38
rs1024723964 Chr.19: 44671238-44671238 on Build GRCh38
rs539522169 Chr.19: 44671588-44671588 on Build GRCh38
rs550777658 Chr.19: 44671481-44671481 on Build GRCh38
rs570580567 Chr.19: 44671511-44671511 on Build GRCh38
rs1043475192 Chr.19: 44671577-44671577 on Build GRCh38
rs746441517 Chr.19: 44671278-44671278 on Build GRCh38
rs113965339 Chr.19: 44671190-44671190 on Build GRCh38
rs909994576 Chr.19: 44671412-44671413 on Build GRCh38
rs1037029797 Chr.19: 44671477-44671477 on Build GRCh38
rs889621453 Chr.19: 44671270-44671270 on Build GRCh38
rs34224728 Chr.19: 44671579-44671580 on Build GRCh38
rs990610653 Chr.19: 44671206-44671206 on Build GRCh38
rs766874268 Chr.19: 44671520-44671520 on Build GRCh38
rs568662192 Chr.19: 44671405-44671405 on Build GRCh38
rs970548739 Chr.19: 44671246-44671246 on Build GRCh38
rs530968785 Chr.19: 44671445-44671445 on Build GRCh38
rs548858886 Chr.19: 44671402-44671402 on Build GRCh38
rs889597466 Chr.19: 44671535-44671535 on Build GRCh38
rs150825555 Chr.19: 44671249-44671249 on Build GRCh38
rs186386245 Chr.19: 44671145-44671145 on Build GRCh38
rs940022053 Chr.19: 44671412-44671412 on Build GRCh38

View Full Product Details