Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44505933-44506371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM20
Entrez Gene ID
125931
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Gene Aliases
UNQ9366
UniGene
Hs.689632
Gene Chromosome Location
Chr.19: 44503838-44530100 on Build GRCh38
Transcripts
NM_001102597.2 NM_001102598.2 NM_001102599.2 NM_001102600.2 XM_011526429.2 XM_011526430.2 XM_011526432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203492 c.1735-2delA p.? Chr.19: 44506216-44506216 on Build GRCh38
439754 c.1737G>C p.E579D Chr.19: 44506212-44506212 on Build GRCh38
4565150 c.1781_1782CC>TT p.S594F Chr.19: 44506167-44506168 on Build GRCh38
5043408 c.1781C>A p.S594Y Chr.19: 44506168-44506168 on Build GRCh38
5365326 c.1783G>A p.V595I Chr.19: 44506166-44506166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377019600 Chr.19: 44506163-44506163 on Build GRCh38
rs767551028 Chr.19: 44506244-44506244 on Build GRCh38
rs755024195 Chr.19: 44506114-44506114 on Build GRCh38
rs142633230 Chr.19: 44506259-44506259 on Build GRCh38
rs368316358 Chr.19: 44506243-44506243 on Build GRCh38
rs879936781 Chr.19: 44506298-44506298 on Build GRCh38
rs147391529 Chr.19: 44506167-44506167 on Build GRCh38
rs763125546 Chr.19: 44506073-44506073 on Build GRCh38
rs781266051 Chr.19: 44506119-44506119 on Build GRCh38
rs899398366 Chr.19: 44506214-44506214 on Build GRCh38
rs745625554 Chr.19: 44506166-44506166 on Build GRCh38
rs546789520 Chr.19: 44506266-44506266 on Build GRCh38
rs867408587 Chr.19: 44506172-44506172 on Build GRCh38
rs1057496931 Chr.19: 44506011-44506011 on Build GRCh38
rs531640865 Chr.19: 44506067-44506067 on Build GRCh38
rs1012904004 Chr.19: 44506050-44506050 on Build GRCh38
rs768830410 Chr.19: 44506248-44506248 on Build GRCh38
rs144260841 Chr.19: 44506053-44506053 on Build GRCh38
rs1434577 Chr.19: 44506064-44506064 on Build GRCh38
rs1051858270 Chr.19: 44506137-44506137 on Build GRCh38
rs761431435 Chr.19: 44506196-44506196 on Build GRCh38
rs766670796 Chr.19: 44506232-44506232 on Build GRCh38
rs185391004 Chr.19: 44505969-44505969 on Build GRCh38
rs769708477 Chr.19: 44506056-44506056 on Build GRCh38
rs370322358 Chr.19: 44506120-44506120 on Build GRCh38
rs13345032 Chr.19: 44506004-44506004 on Build GRCh38
rs1026623156 Chr.19: 44506258-44506258 on Build GRCh38
rs932024308 Chr.19: 44506108-44506108 on Build GRCh38
rs763340503 Chr.19: 44506215-44506215 on Build GRCh38
rs373614476 Chr.19: 44506150-44506150 on Build GRCh38
rs762389366 Chr.19: 44506068-44506068 on Build GRCh38
rs746909031 Chr.19: 44506182-44506182 on Build GRCh38
rs769414766 Chr.19: 44506202-44506202 on Build GRCh38
rs753979697 Chr.19: 44506185-44506186 on Build GRCh38
rs759619913 Chr.19: 44506226-44506227 on Build GRCh38
rs894496556 Chr.19: 44506019-44506019 on Build GRCh38
rs921947580 Chr.19: 44506096-44506096 on Build GRCh38
rs373471990 Chr.19: 44506000-44506000 on Build GRCh38
rs1012371425 Chr.19: 44506320-44506320 on Build GRCh38
rs560354313 Chr.19: 44505974-44505974 on Build GRCh38
rs995107035 Chr.19: 44506237-44506237 on Build GRCh38
rs779881184 Chr.19: 44506170-44506170 on Build GRCh38
rs773608197 Chr.19: 44506206-44506206 on Build GRCh38
rs778169781 Chr.19: 44506262-44506262 on Build GRCh38
rs781548043 Chr.19: 44505995-44505995 on Build GRCh38
rs768060519 Chr.19: 44506185-44506185 on Build GRCh38
rs947592301 Chr.19: 44505992-44505992 on Build GRCh38
rs970590199 Chr.19: 44506037-44506037 on Build GRCh38
rs959087969 Chr.19: 44505978-44505978 on Build GRCh38
rs566407088 Chr.19: 44506303-44506303 on Build GRCh38
rs112372603 Chr.19: 44506188-44506188 on Build GRCh38
rs1053690999 Chr.19: 44506093-44506093 on Build GRCh38
rs752796436 Chr.19: 44506252-44506252 on Build GRCh38

View Full Product Details