Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101102216-101102679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPI
Entrez Gene ID
2491
Gene Name
centromere protein I
Gene Aliases
CENP-I, FSHPRH1, LRPR1
UniGene
Hs.348920
Gene Chromosome Location
Chr.X: 101098159-101181847 on Build GRCh38
Transcripts
NM_001318521.1 NM_001318523.1 NM_006733.3 XM_005262111.2 XM_011530894.2 XM_011530895.2 XM_011530897.2 XM_011530899.2 XM_017029383.1

SNP Detail

SNP ID Chromosome Location
rs867650649 Chr.X: 101102498-101102498 on Build GRCh38
rs769793208 Chr.X: 101102518-101102518 on Build GRCh38
rs765733690 Chr.X: 101102494-101102494 on Build GRCh38
rs765817106 Chr.X: 101102321-101102321 on Build GRCh38
rs745463299 Chr.X: 101102360-101102360 on Build GRCh38
rs776207901 Chr.X: 101102490-101102491 on Build GRCh38
rs796841886 Chr.X: 101102437-101102437 on Build GRCh38
rs771844571 Chr.X: 101102400-101102400 on Build GRCh38
rs12011012 Chr.X: 101102516-101102516 on Build GRCh38
rs767702335 Chr.X: 101102244-101102244 on Build GRCh38
rs758784281 Chr.X: 101102338-101102338 on Build GRCh38
rs780710759 Chr.X: 101102345-101102345 on Build GRCh38
rs767759498 Chr.X: 101102650-101102650 on Build GRCh38
rs59141949 Chr.X: 101102429-101102429 on Build GRCh38
rs760940794 Chr.X: 101102424-101102424 on Build GRCh38
rs200797522 Chr.X: 101102497-101102498 on Build GRCh38
rs755640134 Chr.X: 101102369-101102369 on Build GRCh38
rs60464423 Chr.X: 101102517-101102518 on Build GRCh38
rs776899461 Chr.X: 101102441-101102441 on Build GRCh38
rs908104557 Chr.X: 101102587-101102587 on Build GRCh38
rs756424575 Chr.X: 101102462-101102462 on Build GRCh38
rs866630410 Chr.X: 101102333-101102333 on Build GRCh38
rs868342659 Chr.X: 101102522-101102522 on Build GRCh38
rs976461004 Chr.X: 101102566-101102566 on Build GRCh38
rs759674999 Chr.X: 101102459-101102459 on Build GRCh38
rs148497497 Chr.X: 101102514-101102515 on Build GRCh38
rs190903759 Chr.X: 101102496-101102496 on Build GRCh38
rs773419547 Chr.X: 101102250-101102250 on Build GRCh38
rs35162075 Chr.X: 101102496-101102497 on Build GRCh38
rs779494823 Chr.X: 101102379-101102379 on Build GRCh38
rs865935590 Chr.X: 101102437-101102437 on Build GRCh38
rs187409283 Chr.X: 101102302-101102302 on Build GRCh38
rs766678253 Chr.X: 101102426-101102426 on Build GRCh38
rs377659499 Chr.X: 101102264-101102264 on Build GRCh38
rs942215119 Chr.X: 101102605-101102605 on Build GRCh38
rs55716641 Chr.X: 101102573-101102573 on Build GRCh38
rs374865533 Chr.X: 101102486-101102491 on Build GRCh38
rs12833675 Chr.X: 101102575-101102575 on Build GRCh38
rs75488341 Chr.X: 101102439-101102439 on Build GRCh38
rs773141456 Chr.X: 101102547-101102547 on Build GRCh38
rs1035674465 Chr.X: 101102506-101102506 on Build GRCh38
rs747902723 Chr.X: 101102394-101102394 on Build GRCh38
rs759847027 Chr.X: 101102607-101102607 on Build GRCh38
rs767600505 Chr.X: 101102428-101102429 on Build GRCh38
rs141102097 Chr.X: 101102391-101102391 on Build GRCh38
rs756254011 Chr.X: 101102485-101102486 on Build GRCh38
rs765282648 Chr.X: 101102307-101102307 on Build GRCh38
rs773340500 Chr.X: 101102337-101102337 on Build GRCh38
rs199833748 Chr.X: 101102494-101102495 on Build GRCh38
rs750578447 Chr.X: 101102461-101102461 on Build GRCh38
rs199742109 Chr.X: 101102492-101102493 on Build GRCh38
rs755581582 Chr.X: 101102318-101102318 on Build GRCh38
rs763160317 Chr.X: 101102380-101102380 on Build GRCh38
rs778560144 Chr.X: 101102516-101102517 on Build GRCh38
rs751826366 Chr.X: 101102470-101102470 on Build GRCh38
rs745821824 Chr.X: 101102491-101102492 on Build GRCh38
rs867622085 Chr.X: 101102520-101102520 on Build GRCh38
rs773330852 Chr.X: 101102418-101102418 on Build GRCh38
rs187429448 Chr.X: 101102582-101102582 on Build GRCh38
rs147421602 Chr.X: 101102492-101102492 on Build GRCh38
rs768642126 Chr.X: 101102387-101102387 on Build GRCh38

View Full Product Details