Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32971735-32972231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP89
Entrez Gene ID
84902
Gene Name
centrosomal protein 89
Gene Aliases
CCDC123, CEP123
UniGene
Hs.599703
Gene Chromosome Location
Chr.19: 32875925-32972029 on Build GRCh38
Transcripts
NM_032816.4 XM_005259344.3 XM_017027398.1 NM_001300978.1 NM_152266.4 XM_005259393.3

Gene Symbol
FAAP24
Entrez Gene ID
91442
Gene Name
Fanconi anemia core complex associated protein 24
Gene Aliases
C19orf40
UniGene
Hs.579899
Gene Chromosome Location
Chr.19: 32972212-32977495 on Build GRCh38
Transcripts
NM_032816.4 XM_005259344.3 XM_017027398.1 NM_001300978.1 NM_152266.4 XM_005259393.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6498113 c.30G>A p.R10R Chr.19: 32971845-32971845 on Build GRCh38
5109145 c.39+6T>C p.? Chr.19: 32971830-32971830 on Build GRCh38
5000076 c.34C>G p.H12D Chr.19: 32971841-32971841 on Build GRCh38
5000075 c.34C>G p.H12D Chr.19: 32971841-32971841 on Build GRCh38
994640 c.4C>A p.L2I Chr.19: 32971871-32971871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767261902 Chr.19: 32971804-32971804 on Build GRCh38
rs980561895 Chr.19: 32971904-32971904 on Build GRCh38
rs8109774 Chr.19: 32971930-32971930 on Build GRCh38
rs896990957 Chr.19: 32971945-32971945 on Build GRCh38
rs747086646 Chr.19: 32971843-32971843 on Build GRCh38
rs532038729 Chr.19: 32972094-32972094 on Build GRCh38
rs769043551 Chr.19: 32971882-32971882 on Build GRCh38
rs1050554387 Chr.19: 32971810-32971810 on Build GRCh38
rs1041056986 Chr.19: 32972067-32972067 on Build GRCh38
rs926074903 Chr.19: 32971806-32971806 on Build GRCh38
rs575794232 Chr.19: 32971971-32971971 on Build GRCh38
rs896237424 Chr.19: 32972138-32972138 on Build GRCh38
rs919636424 Chr.19: 32971972-32971972 on Build GRCh38
rs926151453 Chr.19: 32972018-32972018 on Build GRCh38
rs545596625 Chr.19: 32972103-32972103 on Build GRCh38
rs775697744 Chr.19: 32971875-32971875 on Build GRCh38
rs766045245 Chr.19: 32971815-32971815 on Build GRCh38
rs960705504 Chr.19: 32971976-32971976 on Build GRCh38
rs574377970 Chr.19: 32972048-32972048 on Build GRCh38
rs897008444 Chr.19: 32972107-32972110 on Build GRCh38
rs747278441 Chr.19: 32971765-32971765 on Build GRCh38
rs559831280 Chr.19: 32972086-32972086 on Build GRCh38
rs755008843 Chr.19: 32971922-32971922 on Build GRCh38
rs141271133 Chr.19: 32972033-32972033 on Build GRCh38
rs908188482 Chr.19: 32971951-32971951 on Build GRCh38
rs1023337856 Chr.19: 32972197-32972197 on Build GRCh38
rs767065939 Chr.19: 32971894-32971894 on Build GRCh38
rs548468487 Chr.19: 32972106-32972106 on Build GRCh38
rs766154576 Chr.19: 32971768-32971768 on Build GRCh38
rs1005386327 Chr.19: 32971941-32971941 on Build GRCh38
rs973783593 Chr.19: 32971959-32971959 on Build GRCh38
rs769395900 Chr.19: 32971963-32971963 on Build GRCh38
rs766244413 Chr.19: 32971907-32971907 on Build GRCh38
rs1022362841 Chr.19: 32971901-32971901 on Build GRCh38
rs750033831 Chr.19: 32971811-32971811 on Build GRCh38
rs761417999 Chr.19: 32971888-32971888 on Build GRCh38
rs944959531 Chr.19: 32972066-32972066 on Build GRCh38
rs751731 Chr.19: 32972152-32972152 on Build GRCh38
rs377061550 Chr.19: 32971816-32971816 on Build GRCh38
rs994027581 Chr.19: 32972121-32972121 on Build GRCh38
rs773027488 Chr.19: 32971903-32971903 on Build GRCh38
rs994345941 Chr.19: 32971956-32971956 on Build GRCh38
rs770219740 Chr.19: 32971872-32971872 on Build GRCh38
rs754847206 Chr.19: 32971821-32971821 on Build GRCh38
rs768999273 Chr.19: 32971793-32971793 on Build GRCh38
rs903906072 Chr.19: 32972122-32972122 on Build GRCh38
rs181519439 Chr.19: 32972058-32972058 on Build GRCh38
rs934829105 Chr.19: 32972031-32972031 on Build GRCh38
rs1033493568 Chr.19: 32972145-32972145 on Build GRCh38
rs746052213 Chr.19: 32971870-32971870 on Build GRCh38
rs979003954 Chr.19: 32972014-32972014 on Build GRCh38
rs960606493 Chr.19: 32971937-32971937 on Build GRCh38
rs1009595335 Chr.19: 32971814-32971814 on Build GRCh38
rs3764636 Chr.19: 32971942-32971942 on Build GRCh38
rs376784638 Chr.19: 32972120-32972120 on Build GRCh38
rs774059616 Chr.19: 32971886-32971886 on Build GRCh38
rs1002324061 Chr.19: 32972144-32972144 on Build GRCh38
rs373123128 Chr.19: 32971812-32971812 on Build GRCh38
rs934047508 Chr.19: 32972037-32972037 on Build GRCh38
rs892486079 Chr.19: 32971818-32971818 on Build GRCh38
rs1049689826 Chr.19: 32971928-32971928 on Build GRCh38
rs757271183 Chr.19: 32971858-32971858 on Build GRCh38
rs990517522 Chr.19: 32971988-32971988 on Build GRCh38
rs190784937 Chr.19: 32972162-32972162 on Build GRCh38
rs1023264220 Chr.19: 32972006-32972011 on Build GRCh38
rs186165828 Chr.19: 32972087-32972087 on Build GRCh38
rs866040615 Chr.19: 32971906-32971906 on Build GRCh38
rs1038555837 Chr.19: 32971759-32971759 on Build GRCh38
rs879258435 Chr.19: 32971760-32971760 on Build GRCh38
rs753546073 Chr.19: 32971921-32971921 on Build GRCh38
rs562388904 Chr.19: 32972140-32972140 on Build GRCh38
rs749846338 Chr.19: 32971877-32971877 on Build GRCh38
rs765323475 Chr.19: 32972130-32972130 on Build GRCh38
rs1053208703 Chr.19: 32971820-32971820 on Build GRCh38
rs781400043 Chr.19: 32971864-32971864 on Build GRCh38
rs933994895 Chr.19: 32971886-32971888 on Build GRCh38
rs1043749521 Chr.19: 32972108-32972108 on Build GRCh38
rs939654101 Chr.19: 32971958-32971958 on Build GRCh38
rs1049720007 Chr.19: 32972061-32972061 on Build GRCh38
rs777948531 Chr.19: 32971823-32971823 on Build GRCh38
rs566940218 Chr.19: 32971925-32971925 on Build GRCh38
rs970304204 Chr.19: 32972012-32972012 on Build GRCh38
rs1013433241 Chr.19: 32972206-32972206 on Build GRCh38
rs751732 Chr.19: 32972032-32972032 on Build GRCh38

View Full Product Details