Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23246245-23246780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP7
Entrez Gene ID
91782
Gene Name
charged multivesicular body protein 7
Gene Aliases
-
UniGene
Hs.5019
Gene Chromosome Location
Chr.8: 23243637-23262000 on Build GRCh38
Transcripts
NM_001317899.1 NM_152272.4 XM_006716415.2 XM_017013961.1 XM_017013962.1 XM_017013963.1 XM_017013964.1 XM_017013965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6394436 c.37G>A p.G13R Chr.8: 23246732-23246732 on Build GRCh38
6041512 c.61_62insC p.E23fs*115 Chr.8: 23246756-23246757 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369859458 Chr.8: 23246673-23246673 on Build GRCh38
rs779077485 Chr.8: 23246283-23246283 on Build GRCh38
rs188498740 Chr.8: 23246532-23246532 on Build GRCh38
rs956490205 Chr.8: 23246570-23246570 on Build GRCh38
rs994868131 Chr.8: 23246300-23246300 on Build GRCh38
rs545719362 Chr.8: 23246595-23246595 on Build GRCh38
rs768966961 Chr.8: 23246597-23246597 on Build GRCh38
rs1008524507 Chr.8: 23246486-23246486 on Build GRCh38
rs79284930 Chr.8: 23246495-23246495 on Build GRCh38
rs1049053126 Chr.8: 23246568-23246568 on Build GRCh38
rs2294123 Chr.8: 23246682-23246682 on Build GRCh38
rs927165576 Chr.8: 23246654-23246654 on Build GRCh38
rs1019048580 Chr.8: 23246587-23246587 on Build GRCh38
rs769012960 Chr.8: 23246493-23246494 on Build GRCh38
rs369322622 Chr.8: 23246686-23246686 on Build GRCh38
rs564055720 Chr.8: 23246611-23246611 on Build GRCh38
rs776284298 Chr.8: 23246657-23246657 on Build GRCh38
rs746427690 Chr.8: 23246288-23246288 on Build GRCh38
rs537853691 Chr.8: 23246432-23246432 on Build GRCh38
rs1052119696 Chr.8: 23246352-23246352 on Build GRCh38
rs555855778 Chr.8: 23246447-23246447 on Build GRCh38
rs866030507 Chr.8: 23246667-23246667 on Build GRCh38
rs994548860 Chr.8: 23246453-23246453 on Build GRCh38
rs867159578 Chr.8: 23246712-23246712 on Build GRCh38
rs749206784 Chr.8: 23246735-23246735 on Build GRCh38
rs574475394 Chr.8: 23246516-23246516 on Build GRCh38
rs371585622 Chr.8: 23246694-23246694 on Build GRCh38
rs759678274 Chr.8: 23246741-23246741 on Build GRCh38
rs781718333 Chr.8: 23246752-23246752 on Build GRCh38
rs773334747 Chr.8: 23246731-23246731 on Build GRCh38
rs1047171106 Chr.8: 23246746-23246746 on Build GRCh38
rs1018513611 Chr.8: 23246655-23246655 on Build GRCh38
rs966202938 Chr.8: 23246388-23246388 on Build GRCh38
rs905771217 Chr.8: 23246547-23246547 on Build GRCh38
rs901914209 Chr.8: 23246692-23246692 on Build GRCh38
rs988172611 Chr.8: 23246588-23246588 on Build GRCh38
rs768597444 Chr.8: 23246755-23246756 on Build GRCh38
rs4563894 Chr.8: 23246360-23246360 on Build GRCh38
rs891103085 Chr.8: 23246749-23246749 on Build GRCh38
rs941847584 Chr.8: 23246601-23246601 on Build GRCh38
rs571180587 Chr.8: 23246719-23246719 on Build GRCh38
rs760790780 Chr.8: 23246760-23246760 on Build GRCh38
rs769791976 Chr.8: 23246650-23246650 on Build GRCh38
rs1025653421 Chr.8: 23246465-23246465 on Build GRCh38
rs554862627 Chr.8: 23246392-23246392 on Build GRCh38
rs917791564 Chr.8: 23246358-23246358 on Build GRCh38
rs964234820 Chr.8: 23246520-23246520 on Build GRCh38
rs1028096621 Chr.8: 23246342-23246342 on Build GRCh38
rs1038443706 Chr.8: 23246399-23246399 on Build GRCh38
rs754602844 Chr.8: 23246705-23246705 on Build GRCh38
rs560767327 Chr.8: 23246680-23246680 on Build GRCh38
rs1044269231 Chr.8: 23246508-23246508 on Build GRCh38
rs557607369 Chr.8: 23246376-23246376 on Build GRCh38
rs1001088708 Chr.8: 23246730-23246730 on Build GRCh38
rs35764309 Chr.8: 23246495-23246497 on Build GRCh38
rs968312091 Chr.8: 23246395-23246395 on Build GRCh38
rs548392575 Chr.8: 23246678-23246678 on Build GRCh38
rs988548433 Chr.8: 23246612-23246612 on Build GRCh38
rs979210169 Chr.8: 23246324-23246324 on Build GRCh38
rs534548346 Chr.8: 23246387-23246387 on Build GRCh38
rs67349869 Chr.8: 23246494-23246496 on Build GRCh38
rs891161305 Chr.8: 23246647-23246647 on Build GRCh38
rs764402077 Chr.8: 23246494-23246499 on Build GRCh38
rs111683156 Chr.8: 23246272-23246272 on Build GRCh38
rs934967548 Chr.8: 23246347-23246347 on Build GRCh38
rs368371553 Chr.8: 23246621-23246621 on Build GRCh38
rs1018446459 Chr.8: 23246502-23246502 on Build GRCh38
rs988966692 Chr.8: 23246431-23246431 on Build GRCh38
rs544203300 Chr.8: 23246639-23246639 on Build GRCh38
rs752703084 Chr.8: 23246675-23246675 on Build GRCh38
rs926442529 Chr.8: 23246600-23246600 on Build GRCh38
rs112810868 Chr.8: 23246708-23246708 on Build GRCh38
rs757590012 Chr.8: 23246593-23246593 on Build GRCh38
rs1044872583 Chr.8: 23246392-23246392 on Build GRCh38
rs772778321 Chr.8: 23246706-23246706 on Build GRCh38
rs973607314 Chr.8: 23246736-23246736 on Build GRCh38
rs748110164 Chr.8: 23246649-23246649 on Build GRCh38
rs537986788 Chr.8: 23246653-23246653 on Build GRCh38
rs1029262097 Chr.8: 23246392-23246393 on Build GRCh38
rs747366955 Chr.8: 23246442-23246442 on Build GRCh38
rs111600590 Chr.8: 23246396-23246396 on Build GRCh38
rs767793008 Chr.8: 23246750-23246750 on Build GRCh38
rs924923202 Chr.8: 23246334-23246334 on Build GRCh38
rs898542648 Chr.8: 23246614-23246614 on Build GRCh38
rs769418317 Chr.8: 23246723-23246723 on Build GRCh38
rs934422633 Chr.8: 23246406-23246406 on Build GRCh38
rs774133883 Chr.8: 23246739-23246739 on Build GRCh38
rs979614423 Chr.8: 23246531-23246531 on Build GRCh38
rs576863357 Chr.8: 23246457-23246457 on Build GRCh38
rs114517772 Chr.8: 23246590-23246590 on Build GRCh38
rs4581062 Chr.8: 23246398-23246398 on Build GRCh38
rs375307979 Chr.8: 23246554-23246554 on Build GRCh38
rs113024402 Chr.8: 23246711-23246711 on Build GRCh38
rs955775027 Chr.8: 23246483-23246483 on Build GRCh38
rs562699014 Chr.8: 23246646-23246646 on Build GRCh38
rs1019121922 Chr.8: 23246386-23246386 on Build GRCh38
rs1004930153 Chr.8: 23246572-23246572 on Build GRCh38
rs917045501 Chr.8: 23246491-23246491 on Build GRCh38
rs779298604 Chr.8: 23246691-23246691 on Build GRCh38
rs747731611 Chr.8: 23246720-23246720 on Build GRCh38
rs375262054 Chr.8: 23246737-23246737 on Build GRCh38
rs766177063 Chr.8: 23246656-23246656 on Build GRCh38
rs1047504309 Chr.8: 23246622-23246622 on Build GRCh38

View Full Product Details