Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78595671-78596172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA3
Entrez Gene ID
1136
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
Gene Aliases
LNCR2, NACHRA3, PAOD2
UniGene
Hs.89605
Gene Chromosome Location
Chr.15: 78593052-78621295 on Build GRCh38
Transcripts
NM_000743.4 NM_001166694.1 NR_046313.1 XM_006720382.2

SNP Detail

SNP ID Chromosome Location
rs10637216 Chr.15: 78595895-78595896 on Build GRCh38
rs958450480 Chr.15: 78596115-78596115 on Build GRCh38
rs56113144 Chr.15: 78596009-78596009 on Build GRCh38
rs12101928 Chr.15: 78595838-78595838 on Build GRCh38
rs79150488 Chr.15: 78595997-78595997 on Build GRCh38
rs968435724 Chr.15: 78595740-78595740 on Build GRCh38
rs915491226 Chr.15: 78595703-78595703 on Build GRCh38
rs386383550 Chr.15: 78595893-78595894 on Build GRCh38
rs201550331 Chr.15: 78596138-78596138 on Build GRCh38
rs199926717 Chr.15: 78596001-78596001 on Build GRCh38
rs201185018 Chr.15: 78595993-78595993 on Build GRCh38
rs958417908 Chr.15: 78595843-78595844 on Build GRCh38
rs760490495 Chr.15: 78596061-78596061 on Build GRCh38
rs77726924 Chr.15: 78596043-78596043 on Build GRCh38
rs1031387154 Chr.15: 78595785-78595785 on Build GRCh38
rs79043459 Chr.15: 78596086-78596086 on Build GRCh38
rs992688546 Chr.15: 78595797-78595797 on Build GRCh38
rs78211178 Chr.15: 78595944-78595944 on Build GRCh38
rs774510192 Chr.15: 78595766-78595766 on Build GRCh38
rs375922184 Chr.15: 78595895-78595895 on Build GRCh38
rs186138817 Chr.15: 78595950-78595950 on Build GRCh38
rs1037078324 Chr.15: 78595934-78595934 on Build GRCh38
rs79564225 Chr.15: 78596074-78596074 on Build GRCh38
rs71581744 Chr.15: 78595892-78595893 on Build GRCh38
rs999869535 Chr.15: 78595744-78595744 on Build GRCh38
rs980509216 Chr.15: 78595825-78595825 on Build GRCh38
rs971036364 Chr.15: 78595793-78595794 on Build GRCh38
rs978358471 Chr.15: 78595923-78595923 on Build GRCh38
rs80119127 Chr.15: 78596030-78596030 on Build GRCh38
rs146990002 Chr.15: 78595867-78595867 on Build GRCh38
rs200493956 Chr.15: 78595725-78595725 on Build GRCh38
rs970025828 Chr.15: 78595821-78595821 on Build GRCh38
rs962676543 Chr.15: 78596147-78596147 on Build GRCh38
rs200335947 Chr.15: 78596110-78596110 on Build GRCh38
rs946965681 Chr.15: 78595911-78595911 on Build GRCh38
rs76953355 Chr.15: 78595707-78595707 on Build GRCh38
rs201555090 Chr.15: 78595737-78595737 on Build GRCh38
rs999836946 Chr.15: 78596049-78596049 on Build GRCh38
rs199707098 Chr.15: 78596107-78596107 on Build GRCh38
rs981182156 Chr.15: 78595810-78595810 on Build GRCh38
rs199832016 Chr.15: 78595892-78595892 on Build GRCh38
rs560568806 Chr.15: 78595917-78595917 on Build GRCh38
rs8029939 Chr.15: 78596007-78596007 on Build GRCh38
rs201569881 Chr.15: 78595874-78595874 on Build GRCh38
rs1015041040 Chr.15: 78595765-78595765 on Build GRCh38
rs201775345 Chr.15: 78595999-78595999 on Build GRCh38
rs952058570 Chr.15: 78595755-78595758 on Build GRCh38
rs202206429 Chr.15: 78595718-78595718 on Build GRCh38
rs541584973 Chr.15: 78595754-78595755 on Build GRCh38
rs56148181 Chr.15: 78595816-78595816 on Build GRCh38
rs1040369819 Chr.15: 78595985-78595985 on Build GRCh38
rs989879894 Chr.15: 78596126-78596126 on Build GRCh38
rs900360208 Chr.15: 78595745-78595746 on Build GRCh38
rs1014488469 Chr.15: 78596070-78596070 on Build GRCh38
rs955849480 Chr.15: 78595791-78595791 on Build GRCh38
rs12904278 Chr.15: 78595896-78595896 on Build GRCh38
rs201179655 Chr.15: 78595826-78595826 on Build GRCh38
rs1051299934 Chr.15: 78595994-78595994 on Build GRCh38
rs199846852 Chr.15: 78595987-78595987 on Build GRCh38
rs200202079 Chr.15: 78596087-78596087 on Build GRCh38
rs578776 Chr.15: 78596058-78596058 on Build GRCh38
rs1031354545 Chr.15: 78596055-78596055 on Build GRCh38
rs747089373 Chr.15: 78596047-78596047 on Build GRCh38
rs201135151 Chr.15: 78596037-78596037 on Build GRCh38
rs199551739 Chr.15: 78595761-78595761 on Build GRCh38
rs980008832 Chr.15: 78596137-78596137 on Build GRCh38
rs201238950 Chr.15: 78596106-78596106 on Build GRCh38
rs996024616 Chr.15: 78595748-78595748 on Build GRCh38
rs549732833 Chr.15: 78596092-78596092 on Build GRCh38
rs941347623 Chr.15: 78595888-78595888 on Build GRCh38
rs75860163 Chr.15: 78595871-78595871 on Build GRCh38

View Full Product Details